Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 4487951-4488449 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C12orf4
Entrez Gene ID
57102
Gene Name
chromosome 12 open reading frame 4
Gene Aliases
-
UniGene
Hs.302977
Gene Chromosome Location
Chr.12: 4487730-4538508 on Build GRCh38
Transcripts
NM_001304811.1 NM_020374.3 XM_005253716.1

SNP Detail

SNP ID Chromosome Location
rs980259374 Chr.12: 4488247-4488247 on Build GRCh38
rs917307879 Chr.12: 4488205-4488205 on Build GRCh38
rs773130179 Chr.12: 4488115-4488115 on Build GRCh38
rs751454442 Chr.12: 4488137-4488137 on Build GRCh38
rs1045898750 Chr.12: 4488048-4488048 on Build GRCh38
rs957253027 Chr.12: 4488327-4488327 on Build GRCh38
rs1026229675 Chr.12: 4488157-4488157 on Build GRCh38
rs1031613112 Chr.12: 4488135-4488135 on Build GRCh38
rs370679663 Chr.12: 4488305-4488305 on Build GRCh38
rs762992874 Chr.12: 4488122-4488122 on Build GRCh38
rs978451945 Chr.12: 4488134-4488134 on Build GRCh38
rs78885223 Chr.12: 4488163-4488163 on Build GRCh38
rs905931542 Chr.12: 4488083-4488083 on Build GRCh38
rs913518879 Chr.12: 4488000-4488000 on Build GRCh38
rs140936339 Chr.12: 4488307-4488307 on Build GRCh38
rs139647281 Chr.12: 4488032-4488032 on Build GRCh38
rs1018735478 Chr.12: 4488096-4488096 on Build GRCh38
rs189339235 Chr.12: 4488243-4488243 on Build GRCh38
rs185418758 Chr.12: 4488111-4488111 on Build GRCh38
rs900264716 Chr.12: 4488097-4488097 on Build GRCh38
rs10849081 Chr.12: 4488315-4488315 on Build GRCh38
rs1022552483 Chr.12: 4488119-4488119 on Build GRCh38
rs536616060 Chr.12: 4488397-4488397 on Build GRCh38
rs899374404 Chr.12: 4487988-4487988 on Build GRCh38
rs1008633937 Chr.12: 4488089-4488089 on Build GRCh38
rs766358268 Chr.12: 4488127-4488127 on Build GRCh38
rs930898295 Chr.12: 4487993-4487993 on Build GRCh38
rs142706796 Chr.12: 4488213-4488213 on Build GRCh38
rs944328115 Chr.12: 4488423-4488423 on Build GRCh38
rs1032769669 Chr.12: 4488316-4488316 on Build GRCh38
rs1038474133 Chr.12: 4488114-4488114 on Build GRCh38
rs752223032 Chr.12: 4488193-4488193 on Build GRCh38
rs181876363 Chr.12: 4487997-4487997 on Build GRCh38
rs956066975 Chr.12: 4488151-4488151 on Build GRCh38
rs1018819391 Chr.12: 4488300-4488300 on Build GRCh38
rs762616709 Chr.12: 4488292-4488292 on Build GRCh38

View Full Product Details