Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 6248418-6248911 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR153
Entrez Gene ID
387509
Gene Name
G protein-coupled receptor 153
Gene Aliases
PGR1
UniGene
Hs.531581
Gene Chromosome Location
Chr.1: 6247346-6260975 on Build GRCh38
Transcripts
NM_207370.2 XM_011541434.2 XM_017001250.1

SNP Detail

SNP ID Chromosome Location
rs1056077412 Chr.1: 6248669-6248669 on Build GRCh38
rs949435858 Chr.1: 6248648-6248649 on Build GRCh38
rs113114824 Chr.1: 6248830-6248830 on Build GRCh38
rs975612211 Chr.1: 6248744-6248744 on Build GRCh38
rs565151549 Chr.1: 6248813-6248813 on Build GRCh38
rs548671805 Chr.1: 6248876-6248876 on Build GRCh38
rs893070712 Chr.1: 6248688-6248688 on Build GRCh38
rs559843177 Chr.1: 6248874-6248874 on Build GRCh38
rs902878440 Chr.1: 6248701-6248701 on Build GRCh38
rs975626659 Chr.1: 6248588-6248588 on Build GRCh38
rs959552299 Chr.1: 6248463-6248463 on Build GRCh38
rs1034310118 Chr.1: 6248477-6248477 on Build GRCh38
rs527734492 Chr.1: 6248502-6248502 on Build GRCh38
rs989994137 Chr.1: 6248885-6248885 on Build GRCh38
rs569105259 Chr.1: 6248665-6248665 on Build GRCh38
rs879760205 Chr.1: 6248873-6248874 on Build GRCh38
rs74049514 Chr.1: 6248717-6248717 on Build GRCh38
rs975945945 Chr.1: 6248755-6248755 on Build GRCh38
rs576713113 Chr.1: 6248618-6248618 on Build GRCh38
rs545453190 Chr.1: 6248803-6248803 on Build GRCh38
rs983052699 Chr.1: 6248871-6248872 on Build GRCh38
rs931833384 Chr.1: 6248549-6248549 on Build GRCh38
rs12744452 Chr.1: 6248635-6248635 on Build GRCh38
rs907034015 Chr.1: 6248655-6248655 on Build GRCh38
rs542752243 Chr.1: 6248732-6248732 on Build GRCh38
rs1003833232 Chr.1: 6248453-6248453 on Build GRCh38
rs531582259 Chr.1: 6248565-6248565 on Build GRCh38
rs143241502 Chr.1: 6248888-6248891 on Build GRCh38
rs1056698088 Chr.1: 6248865-6248865 on Build GRCh38
rs141090221 Chr.1: 6248611-6248611 on Build GRCh38
rs914406690 Chr.1: 6248886-6248886 on Build GRCh38
rs12138468 Chr.1: 6248476-6248476 on Build GRCh38
rs968419132 Chr.1: 6248496-6248496 on Build GRCh38
rs560778020 Chr.1: 6248745-6248745 on Build GRCh38
rs1022393181 Chr.1: 6248473-6248473 on Build GRCh38
rs895403999 Chr.1: 6248866-6248866 on Build GRCh38
rs560126469 Chr.1: 6248700-6248700 on Build GRCh38
rs185873685 Chr.1: 6248863-6248863 on Build GRCh38
rs369336095 Chr.1: 6248873-6248873 on Build GRCh38
rs547359717 Chr.1: 6248868-6248868 on Build GRCh38
rs901821992 Chr.1: 6248872-6248872 on Build GRCh38
rs537601099 Chr.1: 6248628-6248628 on Build GRCh38
rs921747923 Chr.1: 6248541-6248541 on Build GRCh38
rs990317389 Chr.1: 6248880-6248880 on Build GRCh38
rs948342754 Chr.1: 6248867-6248867 on Build GRCh38
rs928262337 Chr.1: 6248833-6248833 on Build GRCh38
rs892988585 Chr.1: 6248864-6248864 on Build GRCh38
rs12568183 Chr.1: 6248869-6248869 on Build GRCh38
rs940476610 Chr.1: 6248589-6248589 on Build GRCh38
rs1012313975 Chr.1: 6248467-6248467 on Build GRCh38
rs953180188 Chr.1: 6248807-6248807 on Build GRCh38
rs564859655 Chr.1: 6248828-6248828 on Build GRCh38
rs1037653305 Chr.1: 6248447-6248447 on Build GRCh38
rs1031520079 Chr.1: 6248871-6248871 on Build GRCh38
rs377238774 Chr.1: 6248460-6248460 on Build GRCh38
rs897405039 Chr.1: 6248450-6248450 on Build GRCh38
rs964144822 Chr.1: 6248735-6248735 on Build GRCh38
rs909074520 Chr.1: 6248822-6248822 on Build GRCh38
rs965926017 Chr.1: 6248513-6248513 on Build GRCh38
rs573258020 Chr.1: 6248648-6248648 on Build GRCh38
rs964217694 Chr.1: 6248581-6248581 on Build GRCh38
rs937134197 Chr.1: 6248841-6248841 on Build GRCh38
rs972249877 Chr.1: 6248832-6248832 on Build GRCh38
rs145452747 Chr.1: 6248528-6248528 on Build GRCh38
rs190745061 Chr.1: 6248870-6248870 on Build GRCh38
rs1055644894 Chr.1: 6248847-6248847 on Build GRCh38
rs966038418 Chr.1: 6248750-6248750 on Build GRCh38
rs76534458 Chr.1: 6248510-6248510 on Build GRCh38
rs1019355241 Chr.1: 6248728-6248728 on Build GRCh38
rs953006585 Chr.1: 6248769-6248769 on Build GRCh38
rs986539715 Chr.1: 6248533-6248533 on Build GRCh38
rs908966947 Chr.1: 6248570-6248570 on Build GRCh38
rs949405318 Chr.1: 6248817-6248817 on Build GRCh38
rs938341276 Chr.1: 6248855-6248855 on Build GRCh38
rs1056751032 Chr.1: 6248639-6248639 on Build GRCh38
rs1049257228 Chr.1: 6248554-6248554 on Build GRCh38
rs536787678 Chr.1: 6248572-6248572 on Build GRCh38
rs879305968 Chr.1: 6248863-6248864 on Build GRCh38

View Full Product Details