Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 102958640-102959147 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASCL1
Entrez Gene ID
429
Gene Name
achaete-scute family bHLH transcription factor 1
Gene Aliases
ASH1, HASH1, MASH1, bHLHa46
UniGene
Hs.703025
Gene Chromosome Location
Chr.12: 102957674-102960516 on Build GRCh38
Transcripts
NM_004316.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1358241 c.447G>A p.A149A Chr.12: 102958691-102958691 on Build GRCh38
1358240 c.441C>T p.N147N Chr.12: 102958685-102958685 on Build GRCh38
1245550 c.445G>A p.A149T Chr.12: 102958689-102958689 on Build GRCh38
4549403 c.471G>A p.V157V Chr.12: 102958715-102958715 on Build GRCh38
4759621 c.566T>A p.L189Q Chr.12: 102958810-102958810 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779740401 Chr.12: 102958676-102958676 on Build GRCh38
rs760961127 Chr.12: 102958860-102958860 on Build GRCh38
rs754619890 Chr.12: 102958934-102958934 on Build GRCh38
rs906747146 Chr.12: 102958661-102958661 on Build GRCh38
rs762296939 Chr.12: 102958882-102958882 on Build GRCh38
rs776432175 Chr.12: 102958685-102958685 on Build GRCh38
rs757679028 Chr.12: 102958736-102958736 on Build GRCh38
rs748011544 Chr.12: 102958690-102958690 on Build GRCh38
rs995997230 Chr.12: 102958962-102958962 on Build GRCh38
rs373926773 Chr.12: 102958999-102958999 on Build GRCh38
rs752301449 Chr.12: 102958945-102958945 on Build GRCh38
rs748326871 Chr.12: 102958989-102958990 on Build GRCh38
rs761946538 Chr.12: 102958976-102958976 on Build GRCh38
rs751856889 Chr.12: 102958963-102958963 on Build GRCh38
rs754782502 Chr.12: 102958698-102958700 on Build GRCh38
rs771043819 Chr.12: 102958956-102958956 on Build GRCh38
rs1055293109 Chr.12: 102958725-102958725 on Build GRCh38
rs751056404 Chr.12: 102959005-102959005 on Build GRCh38
rs781591287 Chr.12: 102958943-102958944 on Build GRCh38
rs1009016672 Chr.12: 102959092-102959092 on Build GRCh38
rs765842571 Chr.12: 102958755-102958755 on Build GRCh38
rs759716455 Chr.12: 102958700-102958700 on Build GRCh38
rs1803157 Chr.12: 102958717-102958717 on Build GRCh38
rs749184658 Chr.12: 102958832-102958832 on Build GRCh38
rs770156395 Chr.12: 102958977-102958977 on Build GRCh38
rs1042537846 Chr.12: 102959036-102959036 on Build GRCh38
rs750876856 Chr.12: 102958916-102958916 on Build GRCh38
rs112254340 Chr.12: 102958790-102958790 on Build GRCh38
rs148322943 Chr.12: 102958710-102958710 on Build GRCh38
rs1055324639 Chr.12: 102959020-102959020 on Build GRCh38
rs961679419 Chr.12: 102958988-102958988 on Build GRCh38
rs755868552 Chr.12: 102958946-102958946 on Build GRCh38
rs546616125 Chr.12: 102959019-102959019 on Build GRCh38
rs1020678605 Chr.12: 102958823-102958823 on Build GRCh38
rs749198459 Chr.12: 102958951-102958951 on Build GRCh38
rs775736920 Chr.12: 102958859-102958859 on Build GRCh38
rs774289730 Chr.12: 102958697-102958697 on Build GRCh38
rs768991058 Chr.12: 102958972-102958972 on Build GRCh38
rs763417772 Chr.12: 102958982-102958982 on Build GRCh38
rs777456615 Chr.12: 102958949-102958949 on Build GRCh38
rs780252827 Chr.12: 102958959-102958959 on Build GRCh38
rs957409606 Chr.12: 102959084-102959084 on Build GRCh38
rs779545788 Chr.12: 102958788-102958788 on Build GRCh38
rs780932953 Chr.12: 102958813-102958813 on Build GRCh38
rs764454598 Chr.12: 102958868-102958868 on Build GRCh38
rs731682 Chr.12: 102958871-102958871 on Build GRCh38
rs766603626 Chr.12: 102958879-102958879 on Build GRCh38
rs938231545 Chr.12: 102959008-102959008 on Build GRCh38
rs1040029442 Chr.12: 102959109-102959109 on Build GRCh38
rs915429685 Chr.12: 102959029-102959029 on Build GRCh38
rs372191664 Chr.12: 102958917-102958917 on Build GRCh38
rs946838143 Chr.12: 102959032-102959032 on Build GRCh38
rs769602096 Chr.12: 102958826-102958826 on Build GRCh38
rs984185259 Chr.12: 102958979-102958979 on Build GRCh38
rs754672998 Chr.12: 102958791-102958791 on Build GRCh38
rs768374125 Chr.12: 102958684-102958684 on Build GRCh38
rs746680272 Chr.12: 102958679-102958679 on Build GRCh38
rs748042985 Chr.12: 102958818-102958818 on Build GRCh38
rs373158202 Chr.12: 102958692-102958692 on Build GRCh38
rs1019280002 Chr.12: 102958880-102958880 on Build GRCh38
rs765851489 Chr.12: 102958922-102958922 on Build GRCh38
rs761006723 Chr.12: 102958727-102958727 on Build GRCh38
rs757867727 Chr.12: 102958771-102958771 on Build GRCh38
rs765533945 Chr.12: 102958889-102958889 on Build GRCh38
rs776855019 Chr.12: 102958974-102958974 on Build GRCh38
rs754306112 Chr.12: 102958732-102958732 on Build GRCh38
rs34123200 Chr.12: 102959072-102959073 on Build GRCh38
rs902219061 Chr.12: 102959075-102959075 on Build GRCh38
rs544415662 Chr.12: 102958943-102958943 on Build GRCh38
rs147504896 Chr.12: 102958827-102958827 on Build GRCh38
rs924608992 Chr.12: 102959069-102959069 on Build GRCh38
rs562900183 Chr.12: 102958978-102958978 on Build GRCh38
rs886801322 Chr.12: 102958781-102958781 on Build GRCh38
rs751107874 Chr.12: 102958930-102958930 on Build GRCh38
rs775785911 Chr.12: 102958715-102958715 on Build GRCh38
rs143148107 Chr.12: 102958730-102958730 on Build GRCh38
rs749888158 Chr.12: 102958769-102958769 on Build GRCh38
rs11111420 Chr.12: 102958952-102958952 on Build GRCh38
rs772235674 Chr.12: 102958834-102958834 on Build GRCh38
rs562680434 Chr.12: 102958718-102958718 on Build GRCh38

View Full Product Details