Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 103339354-103339853 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF5
Entrez Gene ID
1983
Gene Name
eukaryotic translation initiation factor 5
Gene Aliases
EIF-5, EIF-5A
UniGene
Hs.433702
Gene Chromosome Location
Chr.14: 103334002-103345025 on Build GRCh38
Transcripts
NM_001969.4 NM_183004.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3936423 c.1000C>G p.P334A Chr.14: 103339732-103339732 on Build GRCh38
3744229 c.1021T>A p.Y341N Chr.14: 103339753-103339753 on Build GRCh38
1707902 c.946C>T p.H316Y Chr.14: 103339678-103339678 on Build GRCh38
7337701 c.1052T>C p.I351T Chr.14: 103339784-103339784 on Build GRCh38
5729902 c.911G>A p.C304Y Chr.14: 103339643-103339643 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748987642 Chr.14: 103339622-103339622 on Build GRCh38
rs974319330 Chr.14: 103339695-103339695 on Build GRCh38
rs574739499 Chr.14: 103339538-103339538 on Build GRCh38
rs544139996 Chr.14: 103339552-103339552 on Build GRCh38
rs769380604 Chr.14: 103339612-103339612 on Build GRCh38
rs772677637 Chr.14: 103339532-103339532 on Build GRCh38
rs532642425 Chr.14: 103339783-103339783 on Build GRCh38
rs1001280375 Chr.14: 103339620-103339620 on Build GRCh38
rs767435217 Chr.14: 103339385-103339385 on Build GRCh38
rs760489465 Chr.14: 103339654-103339654 on Build GRCh38
rs149375790 Chr.14: 103339668-103339668 on Build GRCh38
rs200636039 Chr.14: 103339789-103339789 on Build GRCh38
rs766954577 Chr.14: 103339652-103339652 on Build GRCh38
rs1048175276 Chr.14: 103339651-103339651 on Build GRCh38
rs531495028 Chr.14: 103339578-103339578 on Build GRCh38
rs190274777 Chr.14: 103339523-103339523 on Build GRCh38
rs781658262 Chr.14: 103339727-103339727 on Build GRCh38
rs961192955 Chr.14: 103339499-103339499 on Build GRCh38
rs371151308 Chr.14: 103339703-103339703 on Build GRCh38
rs768280061 Chr.14: 103339624-103339624 on Build GRCh38
rs1011207321 Chr.14: 103339404-103339404 on Build GRCh38
rs1031502036 Chr.14: 103339534-103339534 on Build GRCh38
rs1026683830 Chr.14: 103339443-103339443 on Build GRCh38
rs368646581 Chr.14: 103339478-103339478 on Build GRCh38
rs757721525 Chr.14: 103339722-103339722 on Build GRCh38
rs547626594 Chr.14: 103339746-103339746 on Build GRCh38
rs746456095 Chr.14: 103339558-103339558 on Build GRCh38
rs878889812 Chr.14: 103339554-103339554 on Build GRCh38
rs768335016 Chr.14: 103339758-103339758 on Build GRCh38
rs562540355 Chr.14: 103339577-103339577 on Build GRCh38
rs1033835888 Chr.14: 103339448-103339448 on Build GRCh38
rs1036904175 Chr.14: 103339582-103339582 on Build GRCh38
rs865866202 Chr.14: 103339603-103339603 on Build GRCh38
rs1037177511 Chr.14: 103339416-103339416 on Build GRCh38
rs535384715 Chr.14: 103339566-103339566 on Build GRCh38
rs181476713 Chr.14: 103339570-103339570 on Build GRCh38
rs17853111 Chr.14: 103339663-103339663 on Build GRCh38
rs1011513768 Chr.14: 103339599-103339599 on Build GRCh38
rs752994678 Chr.14: 103339592-103339592 on Build GRCh38
rs765363519 Chr.14: 103339656-103339656 on Build GRCh38
rs752706175 Chr.14: 103339665-103339665 on Build GRCh38
rs778513411 Chr.14: 103339770-103339770 on Build GRCh38
rs370322854 Chr.14: 103339613-103339615 on Build GRCh38
rs72708831 Chr.14: 103339633-103339633 on Build GRCh38
rs758482988 Chr.14: 103339593-103339593 on Build GRCh38
rs770769193 Chr.14: 103339810-103339810 on Build GRCh38
rs753956619 Chr.14: 103339588-103339588 on Build GRCh38
rs576890269 Chr.14: 103339565-103339565 on Build GRCh38
rs370607296 Chr.14: 103339629-103339629 on Build GRCh38
rs763105240 Chr.14: 103339829-103339829 on Build GRCh38
rs757880965 Chr.14: 103339601-103339601 on Build GRCh38
rs542189573 Chr.14: 103339602-103339602 on Build GRCh38
rs766542614 Chr.14: 103339646-103339648 on Build GRCh38
rs774069710 Chr.14: 103339380-103339380 on Build GRCh38
rs995166203 Chr.14: 103339430-103339430 on Build GRCh38
rs778339636 Chr.14: 103339595-103339595 on Build GRCh38
rs765181042 Chr.14: 103339590-103339590 on Build GRCh38
rs1052943691 Chr.14: 103339580-103339580 on Build GRCh38
rs747748708 Chr.14: 103339778-103339778 on Build GRCh38
rs779368912 Chr.14: 103339621-103339621 on Build GRCh38
rs760674708 Chr.14: 103339797-103339797 on Build GRCh38
rs375443615 Chr.14: 103339755-103339755 on Build GRCh38
rs539920522 Chr.14: 103339543-103339543 on Build GRCh38
rs139943036 Chr.14: 103339419-103339419 on Build GRCh38
rs974013952 Chr.14: 103339549-103339549 on Build GRCh38
rs755061763 Chr.14: 103339589-103339589 on Build GRCh38
rs1012464421 Chr.14: 103339502-103339502 on Build GRCh38
rs752020951 Chr.14: 103339712-103339712 on Build GRCh38
rs971274840 Chr.14: 103339528-103339528 on Build GRCh38
rs894819860 Chr.14: 103339484-103339484 on Build GRCh38
rs919914233 Chr.14: 103339828-103339828 on Build GRCh38
rs746314881 Chr.14: 103339610-103339610 on Build GRCh38
rs377592907 Chr.14: 103339598-103339598 on Build GRCh38
rs553487061 Chr.14: 103339547-103339547 on Build GRCh38
rs906824293 Chr.14: 103339796-103339796 on Build GRCh38
rs377276266 Chr.14: 103339674-103339674 on Build GRCh38
rs371198700 Chr.14: 103339692-103339692 on Build GRCh38
rs750948249 Chr.14: 103339740-103339740 on Build GRCh38
rs775680600 Chr.14: 103339823-103339823 on Build GRCh38
rs761794084 Chr.14: 103339384-103339384 on Build GRCh38
rs777906217 Chr.14: 103339610-103339611 on Build GRCh38
rs201789959 Chr.14: 103339820-103339820 on Build GRCh38
rs34222936 Chr.14: 103339611-103339613 on Build GRCh38
rs751821459 Chr.14: 103339828-103339829 on Build GRCh38
rs773222627 Chr.14: 103339649-103339649 on Build GRCh38
rs772044086 Chr.14: 103339636-103339636 on Build GRCh38
rs116795967 Chr.14: 103339415-103339415 on Build GRCh38
rs566823311 Chr.14: 103339780-103339780 on Build GRCh38
rs1000094663 Chr.14: 103339807-103339807 on Build GRCh38
rs756525202 Chr.14: 103339741-103339741 on Build GRCh38
rs149719688 Chr.14: 103339487-103339487 on Build GRCh38

View Full Product Details