Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 27460521-27461016 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTF3C1
Entrez Gene ID
2975
Gene Name
general transcription factor IIIC subunit 1
Gene Aliases
TFIIIC, TFIIIC220, TFIIICalpha
UniGene
Hs.371718
Gene Chromosome Location
Chr.16: 27460613-27549930 on Build GRCh38
Transcripts
NM_001286242.1 NM_001520.3 XM_017023188.1

SNP Detail

SNP ID Chromosome Location
rs920179721 Chr.16: 27460978-27460978 on Build GRCh38
rs957320958 Chr.16: 27460898-27460898 on Build GRCh38
rs376126286 Chr.16: 27460615-27460615 on Build GRCh38
rs1025277892 Chr.16: 27460647-27460647 on Build GRCh38
rs990210882 Chr.16: 27460665-27460665 on Build GRCh38
rs940204035 Chr.16: 27460937-27460937 on Build GRCh38
rs999416302 Chr.16: 27460889-27460889 on Build GRCh38
rs973818643 Chr.16: 27460910-27460910 on Build GRCh38
rs926893783 Chr.16: 27460890-27460890 on Build GRCh38
rs34851727 Chr.16: 27460676-27460677 on Build GRCh38
rs990158564 Chr.16: 27460899-27460899 on Build GRCh38
rs552116752 Chr.16: 27460813-27460813 on Build GRCh38
rs147677668 Chr.16: 27460838-27460838 on Build GRCh38
rs894903025 Chr.16: 27460853-27460853 on Build GRCh38
rs1046931591 Chr.16: 27460586-27460586 on Build GRCh38
rs921363063 Chr.16: 27460734-27460735 on Build GRCh38
rs79460981 Chr.16: 27460763-27460763 on Build GRCh38
rs929392710 Chr.16: 27460920-27460920 on Build GRCh38
rs972934934 Chr.16: 27460965-27460965 on Build GRCh38
rs996956829 Chr.16: 27460732-27460732 on Build GRCh38
rs943874471 Chr.16: 27460944-27460944 on Build GRCh38
rs1046901632 Chr.16: 27460929-27460929 on Build GRCh38
rs958035835 Chr.16: 27460781-27460781 on Build GRCh38
rs1024791557 Chr.16: 27460884-27460884 on Build GRCh38
rs920648027 Chr.16: 27460915-27460915 on Build GRCh38
rs982317857 Chr.16: 27460840-27460840 on Build GRCh38
rs184889434 Chr.16: 27460542-27460542 on Build GRCh38
rs754252364 Chr.16: 27460971-27460971 on Build GRCh38
rs932780638 Chr.16: 27460735-27460737 on Build GRCh38
rs1019571068 Chr.16: 27460684-27460684 on Build GRCh38
rs957371892 Chr.16: 27460664-27460665 on Build GRCh38
rs1013422047 Chr.16: 27460641-27460641 on Build GRCh38
rs988230605 Chr.16: 27460782-27460782 on Build GRCh38
rs899479035 Chr.16: 27460685-27460685 on Build GRCh38
rs1021099615 Chr.16: 27460786-27460786 on Build GRCh38
rs12922976 Chr.16: 27460543-27460543 on Build GRCh38
rs543752848 Chr.16: 27460662-27460662 on Build GRCh38
rs75173566 Chr.16: 27460657-27460657 on Build GRCh38
rs548382705 Chr.16: 27460926-27460926 on Build GRCh38
rs938016632 Chr.16: 27460897-27460897 on Build GRCh38
rs886998766 Chr.16: 27460593-27460593 on Build GRCh38
rs147025942 Chr.16: 27460663-27460663 on Build GRCh38
rs902374660 Chr.16: 27460885-27460885 on Build GRCh38
rs559596915 Chr.16: 27460837-27460837 on Build GRCh38
rs568293080 Chr.16: 27460871-27460871 on Build GRCh38
rs1037273009 Chr.16: 27460799-27460799 on Build GRCh38
rs760161820 Chr.16: 27460675-27460676 on Build GRCh38
rs1040963680 Chr.16: 27460955-27460955 on Build GRCh38
rs548384236 Chr.16: 27460842-27460842 on Build GRCh38
rs189400069 Chr.16: 27460570-27460570 on Build GRCh38
rs1033401519 Chr.16: 27460564-27460564 on Build GRCh38
rs114173854 Chr.16: 27460936-27460936 on Build GRCh38

View Full Product Details