Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 27464569-27465105 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTF3C1
Entrez Gene ID
2975
Gene Name
general transcription factor IIIC subunit 1
Gene Aliases
TFIIIC, TFIIIC220, TFIIICalpha
UniGene
Hs.371718
Gene Chromosome Location
Chr.16: 27460613-27549930 on Build GRCh38
Transcripts
NM_001286242.1 NM_001520.3 XM_017023188.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6676772 c.5370G>T p.Q1790H Chr.16: 27464822-27464822 on Build GRCh38
4185714 c.5602A>G p.S1868G Chr.16: 27464590-27464590 on Build GRCh38
4059640 c.5474C>T p.A1825V Chr.16: 27464718-27464718 on Build GRCh38
5384610 c.5586G>A p.R1862R Chr.16: 27464606-27464606 on Build GRCh38
4059638 c.5587C>T p.R1863C Chr.16: 27464605-27464605 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751246377 Chr.16: 27464699-27464719 on Build GRCh38
rs750972619 Chr.16: 27464630-27464632 on Build GRCh38
rs375324231 Chr.16: 27464859-27464859 on Build GRCh38
rs554509437 Chr.16: 27464649-27464649 on Build GRCh38
rs200654354 Chr.16: 27464783-27464783 on Build GRCh38
rs942975330 Chr.16: 27464924-27464924 on Build GRCh38
rs912786760 Chr.16: 27464911-27464911 on Build GRCh38
rs561383320 Chr.16: 27464837-27464837 on Build GRCh38
rs763161381 Chr.16: 27464650-27464651 on Build GRCh38
rs774131442 Chr.16: 27464714-27464714 on Build GRCh38
rs772166492 Chr.16: 27464603-27464603 on Build GRCh38
rs899579907 Chr.16: 27464722-27464722 on Build GRCh38
rs755178488 Chr.16: 27464858-27464858 on Build GRCh38
rs201456699 Chr.16: 27464657-27464657 on Build GRCh38
rs35870192 Chr.16: 27464717-27464717 on Build GRCh38
rs751045906 Chr.16: 27464639-27464639 on Build GRCh38
rs187541487 Chr.16: 27465052-27465052 on Build GRCh38
rs778419217 Chr.16: 27464767-27464767 on Build GRCh38
rs971084579 Chr.16: 27464615-27464615 on Build GRCh38
rs199681752 Chr.16: 27464853-27464853 on Build GRCh38
rs750603013 Chr.16: 27464650-27464650 on Build GRCh38
rs368203794 Chr.16: 27464789-27464789 on Build GRCh38
rs118005982 Chr.16: 27465011-27465011 on Build GRCh38
rs374523613 Chr.16: 27464930-27464930 on Build GRCh38
rs926995584 Chr.16: 27464624-27464624 on Build GRCh38
rs777054416 Chr.16: 27464662-27464662 on Build GRCh38
rs771574952 Chr.16: 27465060-27465060 on Build GRCh38
rs975661157 Chr.16: 27464949-27464949 on Build GRCh38
rs377528153 Chr.16: 27464718-27464718 on Build GRCh38
rs777194031 Chr.16: 27464619-27464619 on Build GRCh38
rs111579786 Chr.16: 27465039-27465039 on Build GRCh38
rs549894790 Chr.16: 27464919-27464919 on Build GRCh38
rs766201057 Chr.16: 27464705-27464705 on Build GRCh38
rs769089553 Chr.16: 27464618-27464618 on Build GRCh38
rs917401218 Chr.16: 27464867-27464867 on Build GRCh38
rs940704778 Chr.16: 27464637-27464637 on Build GRCh38
rs751813220 Chr.16: 27464857-27464857 on Build GRCh38
rs374226840 Chr.16: 27464653-27464653 on Build GRCh38
rs552301601 Chr.16: 27464985-27464985 on Build GRCh38
rs903428916 Chr.16: 27464921-27464921 on Build GRCh38
rs34737721 Chr.16: 27464620-27464620 on Build GRCh38
rs760530604 Chr.16: 27464821-27464821 on Build GRCh38
rs370190480 Chr.16: 27464604-27464604 on Build GRCh38
rs936709197 Chr.16: 27464969-27464969 on Build GRCh38
rs763806529 Chr.16: 27464720-27464720 on Build GRCh38
rs950202632 Chr.16: 27464896-27464896 on Build GRCh38
rs993065252 Chr.16: 27464933-27464933 on Build GRCh38
rs201376754 Chr.16: 27464602-27464602 on Build GRCh38
rs1003992604 Chr.16: 27465040-27465040 on Build GRCh38
rs780116256 Chr.16: 27464654-27464654 on Build GRCh38
rs545590934 Chr.16: 27464739-27464739 on Build GRCh38
rs764986583 Chr.16: 27464856-27464856 on Build GRCh38
rs766782270 Chr.16: 27464695-27464695 on Build GRCh38
rs759165012 Chr.16: 27464716-27464716 on Build GRCh38
rs372263308 Chr.16: 27464855-27464855 on Build GRCh38
rs749728897 Chr.16: 27464771-27464771 on Build GRCh38
rs892548069 Chr.16: 27465006-27465006 on Build GRCh38
rs149738247 Chr.16: 27464740-27464740 on Build GRCh38
rs534956383 Chr.16: 27465023-27465023 on Build GRCh38
rs145463437 Chr.16: 27464744-27464744 on Build GRCh38
rs569776788 Chr.16: 27464961-27464961 on Build GRCh38
rs547765738 Chr.16: 27464804-27464804 on Build GRCh38
rs774186409 Chr.16: 27464776-27464776 on Build GRCh38
rs762698678 Chr.16: 27464699-27464699 on Build GRCh38
rs767108822 Chr.16: 27464838-27464839 on Build GRCh38
rs749780130 Chr.16: 27464888-27464888 on Build GRCh38
rs962982039 Chr.16: 27464836-27464836 on Build GRCh38
rs368155594 Chr.16: 27464667-27464667 on Build GRCh38
rs748624940 Chr.16: 27464747-27464747 on Build GRCh38
rs753566678 Chr.16: 27464727-27464727 on Build GRCh38
rs757450992 Chr.16: 27464903-27464903 on Build GRCh38
rs9888849 Chr.16: 27464978-27464978 on Build GRCh38
rs756134739 Chr.16: 27464875-27464875 on Build GRCh38
rs747530911 Chr.16: 27464607-27464607 on Build GRCh38
rs951574793 Chr.16: 27464899-27464899 on Build GRCh38
rs762653996 Chr.16: 27464632-27464632 on Build GRCh38
rs201674362 Chr.16: 27464652-27464652 on Build GRCh38
rs371093619 Chr.16: 27464846-27464846 on Build GRCh38
rs779758810 Chr.16: 27464719-27464719 on Build GRCh38
rs754747703 Chr.16: 27464759-27464760 on Build GRCh38
rs752888384 Chr.16: 27464864-27464864 on Build GRCh38
rs773577095 Chr.16: 27464668-27464668 on Build GRCh38
rs778470418 Chr.16: 27464887-27464887 on Build GRCh38
rs752200937 Chr.16: 27464881-27464881 on Build GRCh38
rs554563678 Chr.16: 27465043-27465043 on Build GRCh38
rs748660296 Chr.16: 27464665-27464665 on Build GRCh38
rs768711825 Chr.16: 27464660-27464660 on Build GRCh38
rs766010926 Chr.16: 27464633-27464633 on Build GRCh38
rs920680662 Chr.16: 27464641-27464641 on Build GRCh38
rs975209611 Chr.16: 27464617-27464617 on Build GRCh38
rs779049860 Chr.16: 27464595-27464595 on Build GRCh38
rs772781415 Chr.16: 27464628-27464628 on Build GRCh38
rs962392935 Chr.16: 27464876-27464876 on Build GRCh38
rs565442635 Chr.16: 27464775-27464775 on Build GRCh38
rs530012376 Chr.16: 27464897-27464897 on Build GRCh38
rs200275527 Chr.16: 27464651-27464651 on Build GRCh38
rs925274705 Chr.16: 27464960-27464960 on Build GRCh38
rs1010525713 Chr.16: 27464844-27464844 on Build GRCh38
rs890925406 Chr.16: 27465015-27465015 on Build GRCh38
rs962465379 Chr.16: 27464625-27464625 on Build GRCh38
rs201678843 Chr.16: 27464743-27464743 on Build GRCh38
rs372063061 Chr.16: 27464671-27464671 on Build GRCh38
rs60215987 Chr.16: 27464708-27464708 on Build GRCh38
rs770278888 Chr.16: 27464757-27464757 on Build GRCh38

View Full Product Details