Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7114461-7114972 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASGR2
Entrez Gene ID
433
Gene Name
asialoglycoprotein receptor 2
Gene Aliases
ASGP-R2, ASGPR2, CLEC4H2, HBXBP, HL-2
UniGene
Hs.654440
Gene Chromosome Location
Chr.17: 7101322-7115522 on Build GRCh38
Transcripts
NM_001181.4 NM_001201352.1 NM_080912.3 NM_080913.3 XM_005256648.2 XM_006721524.2 XM_006721526.1 XM_011523862.2 XM_011523863.2 XM_011523864.2 XM_011523865.1 XM_011523866.1 XM_017024651.1 XM_017024652.1 XM_017024653.1

SNP Detail

SNP ID Chromosome Location
rs954378255 Chr.17: 7114570-7114570 on Build GRCh38
rs988033343 Chr.17: 7114921-7114921 on Build GRCh38
rs979227106 Chr.17: 7114937-7114937 on Build GRCh38
rs960029088 Chr.17: 7114876-7114876 on Build GRCh38
rs937304144 Chr.17: 7114946-7114946 on Build GRCh38
rs951524128 Chr.17: 7114641-7114641 on Build GRCh38
rs996147224 Chr.17: 7114783-7114783 on Build GRCh38
rs1018501464 Chr.17: 7114755-7114755 on Build GRCh38
rs536249977 Chr.17: 7114740-7114740 on Build GRCh38
rs2289644 Chr.17: 7114903-7114903 on Build GRCh38
rs1001724586 Chr.17: 7114638-7114638 on Build GRCh38
rs984447409 Chr.17: 7114499-7114499 on Build GRCh38
rs1011379200 Chr.17: 7114518-7114518 on Build GRCh38
rs1051823000 Chr.17: 7114613-7114613 on Build GRCh38
rs920763 Chr.17: 7114647-7114647 on Build GRCh38
rs548443821 Chr.17: 7114826-7114826 on Build GRCh38
rs1030024341 Chr.17: 7114553-7114553 on Build GRCh38
rs1013978143 Chr.17: 7114649-7114649 on Build GRCh38
rs928659453 Chr.17: 7114665-7114665 on Build GRCh38
rs34624974 Chr.17: 7114679-7114679 on Build GRCh38
rs530114773 Chr.17: 7114791-7114791 on Build GRCh38
rs947351422 Chr.17: 7114910-7114910 on Build GRCh38
rs938603166 Chr.17: 7114484-7114484 on Build GRCh38
rs1055716731 Chr.17: 7114503-7114503 on Build GRCh38
rs866606572 Chr.17: 7114606-7114606 on Build GRCh38
rs1007141338 Chr.17: 7114748-7114748 on Build GRCh38
rs201738518 Chr.17: 7114514-7114515 on Build GRCh38
rs982921819 Chr.17: 7114832-7114832 on Build GRCh38
rs1016612459 Chr.17: 7114856-7114856 on Build GRCh38
rs967518714 Chr.17: 7114924-7114924 on Build GRCh38
rs57814106 Chr.17: 7114519-7114519 on Build GRCh38
rs964184111 Chr.17: 7114591-7114591 on Build GRCh38
rs920764 Chr.17: 7114528-7114528 on Build GRCh38
rs3214729 Chr.17: 7114940-7114941 on Build GRCh38
rs75501022 Chr.17: 7114941-7114942 on Build GRCh38
rs781229955 Chr.17: 7114725-7114725 on Build GRCh38
rs559386164 Chr.17: 7114633-7114633 on Build GRCh38
rs934454803 Chr.17: 7114944-7114944 on Build GRCh38
rs1005699887 Chr.17: 7114845-7114845 on Build GRCh38
rs991410077 Chr.17: 7114677-7114677 on Build GRCh38
rs1042924848 Chr.17: 7114695-7114695 on Build GRCh38
rs1029549892 Chr.17: 7114904-7114905 on Build GRCh38
rs937608499 Chr.17: 7114763-7114763 on Build GRCh38
rs886886103 Chr.17: 7114835-7114836 on Build GRCh38
rs902999204 Chr.17: 7114710-7114710 on Build GRCh38
rs78971871 Chr.17: 7114747-7114747 on Build GRCh38
rs117563700 Chr.17: 7114686-7114686 on Build GRCh38
rs924505573 Chr.17: 7114932-7114932 on Build GRCh38
rs951197419 Chr.17: 7114659-7114659 on Build GRCh38
rs998999984 Chr.17: 7114539-7114539 on Build GRCh38
rs370857015 Chr.17: 7114662-7114662 on Build GRCh38
rs529237913 Chr.17: 7114574-7114574 on Build GRCh38
rs946032490 Chr.17: 7114634-7114634 on Build GRCh38
rs1016992137 Chr.17: 7114663-7114663 on Build GRCh38
rs1025353898 Chr.17: 7114652-7114652 on Build GRCh38
rs900067025 Chr.17: 7114782-7114782 on Build GRCh38
rs4796386 Chr.17: 7114675-7114675 on Build GRCh38
rs2289645 Chr.17: 7114639-7114639 on Build GRCh38
rs183301562 Chr.17: 7114806-7114806 on Build GRCh38
rs558243279 Chr.17: 7114488-7114488 on Build GRCh38
rs542950667 Chr.17: 7114586-7114586 on Build GRCh38
rs568291160 Chr.17: 7114923-7114923 on Build GRCh38
rs934468891 Chr.17: 7114739-7114739 on Build GRCh38
rs1035741048 Chr.17: 7114864-7114864 on Build GRCh38
rs991462432 Chr.17: 7114892-7114892 on Build GRCh38
rs920765 Chr.17: 7114515-7114515 on Build GRCh38
rs2289646 Chr.17: 7114635-7114635 on Build GRCh38
rs891791091 Chr.17: 7114628-7114628 on Build GRCh38

View Full Product Details