Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 18958897-18959408 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC5A10
Entrez Gene ID
125206
Gene Name
solute carrier family 5 member 10
Gene Aliases
SGLT-5, SGLT5
UniGene
Hs.462418
Gene Chromosome Location
Chr.17: 18950676-19020691 on Build GRCh38
Transcripts
NM_001042450.2 NM_001270648.1 NM_001270649.1 NM_001282417.1 NM_152351.4 XM_011523652.1 XM_011523653.2 XM_017024191.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6783026 c.71C>T p.A24V Chr.17: 18959190-18959190 on Build GRCh38
5182877 c.266C>T p.A89V Chr.17: 18959217-18959217 on Build GRCh38
976366 c.191C>T p.A64V Chr.17: 18959142-18959142 on Build GRCh38
2741338 c.259G>T p.G87C Chr.17: 18959210-18959210 on Build GRCh38
6783032 c.111C>T p.F37F Chr.17: 18959230-18959230 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1020274741 Chr.17: 18959305-18959305 on Build GRCh38
rs547355040 Chr.17: 18958961-18958961 on Build GRCh38
rs1035312968 Chr.17: 18959336-18959336 on Build GRCh38
rs950916468 Chr.17: 18959288-18959288 on Build GRCh38
rs868185880 Chr.17: 18959219-18959219 on Build GRCh38
rs748208181 Chr.17: 18959131-18959131 on Build GRCh38
rs1054758188 Chr.17: 18959003-18959003 on Build GRCh38
rs141010615 Chr.17: 18959353-18959353 on Build GRCh38
rs752398263 Chr.17: 18959090-18959090 on Build GRCh38
rs776188825 Chr.17: 18959233-18959233 on Build GRCh38
rs752166223 Chr.17: 18959181-18959181 on Build GRCh38
rs749349168 Chr.17: 18959135-18959135 on Build GRCh38
rs775126438 Chr.17: 18959260-18959260 on Build GRCh38
rs148178887 Chr.17: 18959238-18959238 on Build GRCh38
rs753485335 Chr.17: 18959161-18959161 on Build GRCh38
rs371199217 Chr.17: 18959216-18959216 on Build GRCh38
rs982052590 Chr.17: 18959321-18959321 on Build GRCh38
rs929593302 Chr.17: 18959142-18959142 on Build GRCh38
rs909660019 Chr.17: 18959369-18959369 on Build GRCh38
rs773257179 Chr.17: 18959215-18959215 on Build GRCh38
rs755752701 Chr.17: 18959091-18959091 on Build GRCh38
rs368023740 Chr.17: 18959218-18959218 on Build GRCh38
rs368871920 Chr.17: 18959262-18959262 on Build GRCh38
rs184713738 Chr.17: 18958996-18958996 on Build GRCh38
rs779038083 Chr.17: 18959203-18959203 on Build GRCh38
rs145851589 Chr.17: 18959143-18959143 on Build GRCh38
rs982386577 Chr.17: 18959173-18959173 on Build GRCh38
rs755437279 Chr.17: 18959113-18959113 on Build GRCh38
rs910633466 Chr.17: 18959069-18959069 on Build GRCh38
rs759451010 Chr.17: 18959126-18959126 on Build GRCh38
rs897382180 Chr.17: 18959088-18959088 on Build GRCh38
rs770103114 Chr.17: 18959198-18959202 on Build GRCh38
rs920130667 Chr.17: 18959328-18959328 on Build GRCh38
rs529587659 Chr.17: 18959038-18959038 on Build GRCh38
rs964881666 Chr.17: 18959075-18959075 on Build GRCh38
rs762490779 Chr.17: 18959246-18959246 on Build GRCh38
rs375900675 Chr.17: 18959235-18959235 on Build GRCh38
rs779042953 Chr.17: 18959265-18959265 on Build GRCh38
rs780002209 Chr.17: 18959284-18959284 on Build GRCh38
rs36021973 Chr.17: 18959033-18959034 on Build GRCh38
rs974286619 Chr.17: 18959212-18959212 on Build GRCh38
rs759445784 Chr.17: 18959147-18959147 on Build GRCh38
rs757499110 Chr.17: 18959263-18959263 on Build GRCh38
rs773421754 Chr.17: 18959128-18959128 on Build GRCh38
rs181066022 Chr.17: 18959190-18959190 on Build GRCh38
rs747480723 Chr.17: 18959107-18959107 on Build GRCh38
rs756474448 Chr.17: 18959257-18959257 on Build GRCh38
rs780451515 Chr.17: 18959106-18959106 on Build GRCh38
rs888115704 Chr.17: 18959025-18959025 on Build GRCh38
rs570382583 Chr.17: 18958994-18958994 on Build GRCh38
rs569744995 Chr.17: 18959308-18959308 on Build GRCh38
rs777948863 Chr.17: 18958947-18958947 on Build GRCh38
rs756736600 Chr.17: 18959099-18959099 on Build GRCh38
rs923477417 Chr.17: 18959285-18959285 on Build GRCh38
rs386795995 Chr.17: 18959352-18959353 on Build GRCh38
rs867128152 Chr.17: 18959110-18959110 on Build GRCh38
rs945220180 Chr.17: 18958958-18958958 on Build GRCh38
rs776564750 Chr.17: 18959092-18959092 on Build GRCh38
rs202110338 Chr.17: 18959280-18959280 on Build GRCh38
rs761263626 Chr.17: 18959166-18959166 on Build GRCh38
rs368342002 Chr.17: 18959152-18959152 on Build GRCh38
rs1031682828 Chr.17: 18959017-18959017 on Build GRCh38
rs556143819 Chr.17: 18959380-18959380 on Build GRCh38
rs753214605 Chr.17: 18959252-18959252 on Build GRCh38
rs115522833 Chr.17: 18958988-18958988 on Build GRCh38
rs544704182 Chr.17: 18959146-18959146 on Build GRCh38
rs201103784 Chr.17: 18959153-18959153 on Build GRCh38
rs61740520 Chr.17: 18959251-18959251 on Build GRCh38
rs767267599 Chr.17: 18959089-18959089 on Build GRCh38
rs749493783 Chr.17: 18959202-18959202 on Build GRCh38
rs376732499 Chr.17: 18959201-18959201 on Build GRCh38
rs745960581 Chr.17: 18959279-18959279 on Build GRCh38
rs141133983 Chr.17: 18959223-18959223 on Build GRCh38
rs909463182 Chr.17: 18959318-18959318 on Build GRCh38
rs913788288 Chr.17: 18958932-18958932 on Build GRCh38
rs1005140778 Chr.17: 18959032-18959032 on Build GRCh38
rs764776829 Chr.17: 18959174-18959174 on Build GRCh38
rs966632788 Chr.17: 18958995-18958995 on Build GRCh38
rs763780929 Chr.17: 18959373-18959373 on Build GRCh38
rs748445402 Chr.17: 18959118-18959118 on Build GRCh38
rs867712861 Chr.17: 18959193-18959197 on Build GRCh38
rs1006979022 Chr.17: 18959123-18959123 on Build GRCh38
rs1013424642 Chr.17: 18958963-18958963 on Build GRCh38
rs999823568 Chr.17: 18959008-18959008 on Build GRCh38
rs375990555 Chr.17: 18959114-18959114 on Build GRCh38
rs768462645 Chr.17: 18959231-18959231 on Build GRCh38
rs151061894 Chr.17: 18959352-18959352 on Build GRCh38
rs767359533 Chr.17: 18959150-18959150 on Build GRCh38
rs549400455 Chr.17: 18959039-18959039 on Build GRCh38
rs776839845 Chr.17: 18959379-18959379 on Build GRCh38
rs1010267805 Chr.17: 18959294-18959294 on Build GRCh38
rs767481861 Chr.17: 18959005-18959005 on Build GRCh38
rs749955837 Chr.17: 18959244-18959244 on Build GRCh38
rs370923912 Chr.17: 18959200-18959200 on Build GRCh38
rs146918144 Chr.17: 18959230-18959230 on Build GRCh38
rs753292893 Chr.17: 18959097-18959097 on Build GRCh38
rs147790895 Chr.17: 18959191-18959191 on Build GRCh38
rs770140535 Chr.17: 18959119-18959119 on Build GRCh38

View Full Product Details