Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 55353524-55354016 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COX6B2
Entrez Gene ID
125965
Gene Name
cytochrome c oxidase subunit 6B2
Gene Aliases
COXVIB2, CT59
UniGene
Hs.550544
Gene Chromosome Location
Chr.19: 55349702-55354814 on Build GRCh38
Transcripts
NM_144613.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1001484 c.216G>A p.V72V Chr.19: 55353772-55353772 on Build GRCh38
248863 c.210C>A p.S70R Chr.19: 55353869-55353869 on Build GRCh38
6239877 c.113-6C>A p.? Chr.19: 55353972-55353972 on Build GRCh38
6319186 c.120C>A p.H40Q Chr.19: 55353959-55353959 on Build GRCh38
6331368 c.231G>A p.E77E Chr.19: 55353757-55353757 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749917626 Chr.19: 55353782-55353782 on Build GRCh38
rs532782350 Chr.19: 55353970-55353970 on Build GRCh38
rs373724976 Chr.19: 55353834-55353834 on Build GRCh38
rs1021177174 Chr.19: 55353889-55353889 on Build GRCh38
rs967488344 Chr.19: 55353602-55353602 on Build GRCh38
rs573751549 Chr.19: 55353652-55353652 on Build GRCh38
rs758667483 Chr.19: 55353829-55353829 on Build GRCh38
rs919394325 Chr.19: 55353983-55353983 on Build GRCh38
rs144967211 Chr.19: 55353694-55353694 on Build GRCh38
rs766964948 Chr.19: 55353760-55353760 on Build GRCh38
rs368616257 Chr.19: 55353973-55353973 on Build GRCh38
rs768310323 Chr.19: 55353742-55353742 on Build GRCh38
rs1000147287 Chr.19: 55353753-55353753 on Build GRCh38
rs376652873 Chr.19: 55353985-55353985 on Build GRCh38
rs748245172 Chr.19: 55353726-55353726 on Build GRCh38
rs375106737 Chr.19: 55353758-55353758 on Build GRCh38
rs757743813 Chr.19: 55353546-55353546 on Build GRCh38
rs754403498 Chr.19: 55353794-55353795 on Build GRCh38
rs949187460 Chr.19: 55353854-55353854 on Build GRCh38
rs772656738 Chr.19: 55353896-55353896 on Build GRCh38
rs755214225 Chr.19: 55353717-55353717 on Build GRCh38
rs751035825 Chr.19: 55353583-55353583 on Build GRCh38
rs747838819 Chr.19: 55353885-55353885 on Build GRCh38
rs751601816 Chr.19: 55353921-55353921 on Build GRCh38
rs576407591 Chr.19: 55353884-55353885 on Build GRCh38
rs759928619 Chr.19: 55353739-55353739 on Build GRCh38
rs540732232 Chr.19: 55353936-55353936 on Build GRCh38
rs747105382 Chr.19: 55353838-55353838 on Build GRCh38
rs767447246 Chr.19: 55353709-55353709 on Build GRCh38
rs963574618 Chr.19: 55353977-55353977 on Build GRCh38
rs756591149 Chr.19: 55353939-55353939 on Build GRCh38
rs199757828 Chr.19: 55353961-55353961 on Build GRCh38
rs981811985 Chr.19: 55353567-55353567 on Build GRCh38
rs959006077 Chr.19: 55353580-55353580 on Build GRCh38
rs771727073 Chr.19: 55353952-55353952 on Build GRCh38
rs750319351 Chr.19: 55353711-55353711 on Build GRCh38
rs781681152 Chr.19: 55353852-55353852 on Build GRCh38
rs772243483 Chr.19: 55353731-55353731 on Build GRCh38
rs746336473 Chr.19: 55353937-55353937 on Build GRCh38
rs372455839 Chr.19: 55353919-55353919 on Build GRCh38
rs757292951 Chr.19: 55353935-55353935 on Build GRCh38
rs375973787 Chr.19: 55353681-55353681 on Build GRCh38
rs753591686 Chr.19: 55353807-55353807 on Build GRCh38
rs951004902 Chr.19: 55353956-55353956 on Build GRCh38
rs12461665 Chr.19: 55353982-55353982 on Build GRCh38
rs753665213 Chr.19: 55353716-55353716 on Build GRCh38
rs779754280 Chr.19: 55353715-55353715 on Build GRCh38
rs780464482 Chr.19: 55353945-55353945 on Build GRCh38
rs752922477 Chr.19: 55353824-55353824 on Build GRCh38
rs756056464 Chr.19: 55353712-55353712 on Build GRCh38
rs368349487 Chr.19: 55353887-55353887 on Build GRCh38
rs749640568 Chr.19: 55353947-55353947 on Build GRCh38
rs776594364 Chr.19: 55353899-55353899 on Build GRCh38
rs765159935 Chr.19: 55353577-55353577 on Build GRCh38
rs754739644 Chr.19: 55353811-55353811 on Build GRCh38
rs80221452 Chr.19: 55353689-55353689 on Build GRCh38
rs970572235 Chr.19: 55353891-55353891 on Build GRCh38
rs371151549 Chr.19: 55353951-55353951 on Build GRCh38
rs377408076 Chr.19: 55353920-55353920 on Build GRCh38
rs1056084533 Chr.19: 55353544-55353544 on Build GRCh38
rs970858597 Chr.19: 55353558-55353558 on Build GRCh38
rs537414842 Chr.19: 55353548-55353548 on Build GRCh38
rs375975717 Chr.19: 55353978-55353978 on Build GRCh38
rs893686488 Chr.19: 55353793-55353793 on Build GRCh38
rs771433803 Chr.19: 55353734-55353734 on Build GRCh38
rs866542245 Chr.19: 55353769-55353769 on Build GRCh38
rs557384845 Chr.19: 55353650-55353650 on Build GRCh38
rs369466477 Chr.19: 55353791-55353791 on Build GRCh38
rs373469041 Chr.19: 55353706-55353706 on Build GRCh38
rs796742615 Chr.19: 55353960-55353960 on Build GRCh38
rs775481185 Chr.19: 55353981-55353981 on Build GRCh38
rs776925334 Chr.19: 55353735-55353735 on Build GRCh38
rs777028454 Chr.19: 55353678-55353678 on Build GRCh38
rs763794883 Chr.19: 55353687-55353687 on Build GRCh38
rs746221230 Chr.19: 55353860-55353860 on Build GRCh38
rs774014388 Chr.19: 55353878-55353878 on Build GRCh38
rs1048321097 Chr.19: 55353653-55353653 on Build GRCh38
rs760175987 Chr.19: 55353897-55353897 on Build GRCh38
rs983708538 Chr.19: 55353957-55353957 on Build GRCh38
rs1039130868 Chr.19: 55353964-55353964 on Build GRCh38
rs765177838 Chr.19: 55353914-55353914 on Build GRCh38
rs771396653 Chr.19: 55353850-55353850 on Build GRCh38
rs749268309 Chr.19: 55353708-55353709 on Build GRCh38
rs760596207 Chr.19: 55353789-55353789 on Build GRCh38
rs779222457 Chr.19: 55353724-55353724 on Build GRCh38
rs868054358 Chr.19: 55353917-55353917 on Build GRCh38
rs770103934 Chr.19: 55353865-55353865 on Build GRCh38
rs778449941 Chr.19: 55353732-55353732 on Build GRCh38
rs747515071 Chr.19: 55353733-55353733 on Build GRCh38
rs138900187 Chr.19: 55353911-55353911 on Build GRCh38
rs751764711 Chr.19: 55353988-55353988 on Build GRCh38
rs908846239 Chr.19: 55353752-55353752 on Build GRCh38
rs559887981 Chr.19: 55353898-55353898 on Build GRCh38
rs546237576 Chr.19: 55353615-55353615 on Build GRCh38
rs773924881 Chr.19: 55353754-55353754 on Build GRCh38
rs542900165 Chr.19: 55353822-55353822 on Build GRCh38
rs1041937201 Chr.19: 55353943-55353943 on Build GRCh38
rs944890987 Chr.19: 55353963-55353963 on Build GRCh38
rs963011053 Chr.19: 55353974-55353974 on Build GRCh38
rs750498393 Chr.19: 55353991-55353991 on Build GRCh38
rs903384132 Chr.19: 55353944-55353944 on Build GRCh38
rs757305730 Chr.19: 55353990-55353990 on Build GRCh38
rs920838959 Chr.19: 55353645-55353645 on Build GRCh38

View Full Product Details