Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 69460065-69460575 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AAK1
Entrez Gene ID
22848
Gene Name
AP2 associated kinase 1
Gene Aliases
-
UniGene
Hs.468878
Gene Chromosome Location
Chr.2: 69457995-69643845 on Build GRCh38
Transcripts
NM_014911.3

SNP Detail

SNP ID Chromosome Location
rs980336576 Chr.2: 69460546-69460546 on Build GRCh38
rs983242070 Chr.2: 69460210-69460210 on Build GRCh38
rs1021815254 Chr.2: 69460528-69460528 on Build GRCh38
rs112483904 Chr.2: 69460541-69460541 on Build GRCh38
rs569084898 Chr.2: 69460138-69460138 on Build GRCh38
rs755622707 Chr.2: 69460176-69460176 on Build GRCh38
rs76651100 Chr.2: 69460243-69460243 on Build GRCh38
rs758244813 Chr.2: 69460490-69460490 on Build GRCh38
rs763182103 Chr.2: 69460447-69460447 on Build GRCh38
rs114823244 Chr.2: 69460122-69460122 on Build GRCh38
rs537795841 Chr.2: 69460188-69460188 on Build GRCh38
rs926681717 Chr.2: 69460221-69460221 on Build GRCh38
rs1038404897 Chr.2: 69460217-69460217 on Build GRCh38
rs904004964 Chr.2: 69460268-69460268 on Build GRCh38
rs73934943 Chr.2: 69460454-69460454 on Build GRCh38
rs139493216 Chr.2: 69460113-69460113 on Build GRCh38
rs962615867 Chr.2: 69460494-69460494 on Build GRCh38
rs926462380 Chr.2: 69460547-69460547 on Build GRCh38
rs1049785501 Chr.2: 69460426-69460426 on Build GRCh38
rs553876991 Chr.2: 69460365-69460365 on Build GRCh38
rs891951082 Chr.2: 69460474-69460474 on Build GRCh38
rs1009815823 Chr.2: 69460248-69460248 on Build GRCh38
rs538410318 Chr.2: 69460119-69460119 on Build GRCh38
rs955284153 Chr.2: 69460168-69460168 on Build GRCh38
rs753778827 Chr.2: 69460177-69460177 on Build GRCh38
rs149696109 Chr.2: 69460234-69460234 on Build GRCh38
rs1032226667 Chr.2: 69460117-69460117 on Build GRCh38
rs932620164 Chr.2: 69460422-69460422 on Build GRCh38
rs1032788159 Chr.2: 69460121-69460121 on Build GRCh38
rs376867774 Chr.2: 69460341-69460345 on Build GRCh38
rs1001004501 Chr.2: 69460276-69460276 on Build GRCh38
rs533853842 Chr.2: 69460255-69460255 on Build GRCh38
rs4441525 Chr.2: 69460092-69460092 on Build GRCh38
rs955255230 Chr.2: 69460451-69460451 on Build GRCh38
rs1017804265 Chr.2: 69460310-69460310 on Build GRCh38
rs1056353456 Chr.2: 69460236-69460236 on Build GRCh38
rs969694266 Chr.2: 69460157-69460157 on Build GRCh38
rs975051160 Chr.2: 69460136-69460136 on Build GRCh38
rs1031764662 Chr.2: 69460470-69460470 on Build GRCh38
rs996605980 Chr.2: 69460353-69460353 on Build GRCh38
rs990773935 Chr.2: 69460132-69460132 on Build GRCh38
rs891145487 Chr.2: 69460427-69460427 on Build GRCh38

View Full Product Details