Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 1206818-1207350 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C20orf202
Entrez Gene ID
400831
Gene Name
chromosome 20 open reading frame 202
Gene Aliases
-
UniGene
Hs.148503
Gene Chromosome Location
Chr.20: 1203454-1208274 on Build GRCh38
Transcripts
NM_001009612.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7358382 c.266G>T p.R89I Chr.20: 1206999-1206999 on Build GRCh38
6869768 c.222C>A p.A74A Chr.20: 1206955-1206955 on Build GRCh38
6336042 c.252G>A p.G84G Chr.20: 1206985-1206985 on Build GRCh38
6293311 c.235G>A p.A79T Chr.20: 1206968-1206968 on Build GRCh38
3414131 c.221C>A p.A74D Chr.20: 1206954-1206954 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776325462 Chr.20: 1207139-1207139 on Build GRCh38
rs369917157 Chr.20: 1207019-1207019 on Build GRCh38
rs957069521 Chr.20: 1207070-1207070 on Build GRCh38
rs546383083 Chr.20: 1207068-1207068 on Build GRCh38
rs1005048564 Chr.20: 1207050-1207050 on Build GRCh38
rs151145673 Chr.20: 1206901-1206901 on Build GRCh38
rs772688457 Chr.20: 1207236-1207236 on Build GRCh38
rs886526907 Chr.20: 1207005-1207005 on Build GRCh38
rs772155605 Chr.20: 1206867-1206867 on Build GRCh38
rs372546105 Chr.20: 1206946-1206946 on Build GRCh38
rs925817178 Chr.20: 1206935-1206935 on Build GRCh38
rs1014309242 Chr.20: 1207008-1207008 on Build GRCh38
rs950199204 Chr.20: 1207297-1207297 on Build GRCh38
rs529757520 Chr.20: 1206891-1206892 on Build GRCh38
rs576260774 Chr.20: 1207036-1207036 on Build GRCh38
rs185116681 Chr.20: 1207096-1207096 on Build GRCh38
rs760277903 Chr.20: 1206878-1206878 on Build GRCh38
rs558875651 Chr.20: 1207177-1207177 on Build GRCh38
rs770841361 Chr.20: 1206905-1206905 on Build GRCh38
rs943501470 Chr.20: 1207159-1207159 on Build GRCh38
rs980011046 Chr.20: 1207121-1207121 on Build GRCh38
rs1052370873 Chr.20: 1206903-1206903 on Build GRCh38
rs930842200 Chr.20: 1207223-1207223 on Build GRCh38
rs866769135 Chr.20: 1207004-1207004 on Build GRCh38
rs774628902 Chr.20: 1207168-1207168 on Build GRCh38
rs376822625 Chr.20: 1206959-1206959 on Build GRCh38
rs764590838 Chr.20: 1207009-1207009 on Build GRCh38
rs181425626 Chr.20: 1207085-1207085 on Build GRCh38
rs564190235 Chr.20: 1207130-1207130 on Build GRCh38
rs920842357 Chr.20: 1207190-1207190 on Build GRCh38
rs1036549085 Chr.20: 1207318-1207318 on Build GRCh38
rs749344982 Chr.20: 1206848-1206848 on Build GRCh38
rs912862930 Chr.20: 1207104-1207104 on Build GRCh38
rs781386665 Chr.20: 1207041-1207041 on Build GRCh38
rs769846460 Chr.20: 1207033-1207033 on Build GRCh38
rs556149890 Chr.20: 1206932-1206932 on Build GRCh38
rs940881309 Chr.20: 1207306-1207306 on Build GRCh38
rs80125338 Chr.20: 1207108-1207108 on Build GRCh38
rs542103133 Chr.20: 1207081-1207081 on Build GRCh38
rs994829830 Chr.20: 1206973-1206973 on Build GRCh38
rs979959180 Chr.20: 1206926-1206926 on Build GRCh38
rs375391788 Chr.20: 1206892-1206892 on Build GRCh38
rs79521191 Chr.20: 1207234-1207234 on Build GRCh38
rs969964510 Chr.20: 1206894-1206894 on Build GRCh38
rs1049377354 Chr.20: 1207239-1207239 on Build GRCh38
rs753555889 Chr.20: 1207026-1207026 on Build GRCh38
rs912966128 Chr.20: 1206842-1206842 on Build GRCh38
rs746924826 Chr.20: 1207119-1207119 on Build GRCh38
rs759328192 Chr.20: 1207142-1207142 on Build GRCh38
rs78087104 Chr.20: 1206983-1206983 on Build GRCh38
rs774853040 Chr.20: 1206855-1206855 on Build GRCh38
rs769020106 Chr.20: 1207128-1207128 on Build GRCh38
rs1032878402 Chr.20: 1207035-1207035 on Build GRCh38
rs921980623 Chr.20: 1207233-1207233 on Build GRCh38
rs1040394919 Chr.20: 1206998-1206998 on Build GRCh38
rs888039993 Chr.20: 1207258-1207258 on Build GRCh38
rs912035516 Chr.20: 1206923-1206923 on Build GRCh38
rs750696862 Chr.20: 1207074-1207074 on Build GRCh38
rs975023503 Chr.20: 1207170-1207170 on Build GRCh38
rs899191175 Chr.20: 1206953-1206953 on Build GRCh38
rs961370266 Chr.20: 1206868-1206868 on Build GRCh38
rs944822532 Chr.20: 1207172-1207172 on Build GRCh38
rs768786582 Chr.20: 1207103-1207103 on Build GRCh38
rs887987547 Chr.20: 1207037-1207037 on Build GRCh38
rs79380554 Chr.20: 1206942-1206942 on Build GRCh38
rs548549645 Chr.20: 1206876-1206876 on Build GRCh38
rs912070700 Chr.20: 1207150-1207150 on Build GRCh38

View Full Product Details