Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 86425260-86425769 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLCA2
Entrez Gene ID
9635
Gene Name
chloride channel accessory 2
Gene Aliases
CACC, CACC3, CLCRG2, CaCC-3
UniGene
Hs.241551
Gene Chromosome Location
Chr.1: 86424086-86456558 on Build GRCh38
Transcripts
NM_006536.5 XM_011542448.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4010339 c.292A>G p.N98D Chr.1: 86425444-86425444 on Build GRCh38
6127982 c.233G>T p.R78I Chr.1: 86425385-86425385 on Build GRCh38
5649058 c.310G>C p.E104Q Chr.1: 86425462-86425462 on Build GRCh38
2244324 c.189A>C p.E63D Chr.1: 86425341-86425341 on Build GRCh38
6629773 c.306A>G p.K102K Chr.1: 86425458-86425458 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764516893 Chr.1: 86425433-86425433 on Build GRCh38
rs780523591 Chr.1: 86425342-86425344 on Build GRCh38
rs941891098 Chr.1: 86425673-86425673 on Build GRCh38
rs988356549 Chr.1: 86425552-86425552 on Build GRCh38
rs769191745 Chr.1: 86425474-86425481 on Build GRCh38
rs759235548 Chr.1: 86425393-86425393 on Build GRCh38
rs984794866 Chr.1: 86425655-86425655 on Build GRCh38
rs937414727 Chr.1: 86425407-86425407 on Build GRCh38
rs757643348 Chr.1: 86425448-86425448 on Build GRCh38
rs769327909 Chr.1: 86425516-86425516 on Build GRCh38
rs758489245 Chr.1: 86425313-86425325 on Build GRCh38
rs757555301 Chr.1: 86425316-86425316 on Build GRCh38
rs779973019 Chr.1: 86425354-86425354 on Build GRCh38
rs764614832 Chr.1: 86425300-86425300 on Build GRCh38
rs752095761 Chr.1: 86425523-86425523 on Build GRCh38
rs780415016 Chr.1: 86425500-86425500 on Build GRCh38
rs759291413 Chr.1: 86425515-86425515 on Build GRCh38
rs145716190 Chr.1: 86425422-86425422 on Build GRCh38
rs368536348 Chr.1: 86425637-86425637 on Build GRCh38
rs756044978 Chr.1: 86425384-86425384 on Build GRCh38
rs763548529 Chr.1: 86425522-86425522 on Build GRCh38
rs35688971 Chr.1: 86425513-86425514 on Build GRCh38
rs908878787 Chr.1: 86425670-86425670 on Build GRCh38
rs965511661 Chr.1: 86425591-86425591 on Build GRCh38
rs1025477246 Chr.1: 86425420-86425420 on Build GRCh38
rs762921669 Chr.1: 86425303-86425303 on Build GRCh38
rs11580625 Chr.1: 86425390-86425390 on Build GRCh38
rs367906804 Chr.1: 86425488-86425488 on Build GRCh38
rs753289103 Chr.1: 86425301-86425301 on Build GRCh38
rs34861824 Chr.1: 86425397-86425397 on Build GRCh38
rs748601846 Chr.1: 86425514-86425514 on Build GRCh38
rs772327653 Chr.1: 86425293-86425295 on Build GRCh38
rs770892431 Chr.1: 86425501-86425501 on Build GRCh38
rs897669169 Chr.1: 86425341-86425341 on Build GRCh38
rs775144819 Chr.1: 86425517-86425517 on Build GRCh38
rs2210641 Chr.1: 86425496-86425496 on Build GRCh38
rs1158506 Chr.1: 86425322-86425322 on Build GRCh38
rs745582468 Chr.1: 86425566-86425566 on Build GRCh38
rs751723158 Chr.1: 86425370-86425370 on Build GRCh38
rs1019353352 Chr.1: 86425530-86425530 on Build GRCh38
rs80126918 Chr.1: 86425607-86425607 on Build GRCh38
rs752007799 Chr.1: 86425443-86425443 on Build GRCh38
rs1034501930 Chr.1: 86425675-86425675 on Build GRCh38
rs752135917 Chr.1: 86425419-86425419 on Build GRCh38
rs758068496 Chr.1: 86425644-86425644 on Build GRCh38
rs762387832 Chr.1: 86425521-86425521 on Build GRCh38
rs547013179 Chr.1: 86425576-86425576 on Build GRCh38
rs748171859 Chr.1: 86425355-86425355 on Build GRCh38
rs751921670 Chr.1: 86425313-86425313 on Build GRCh38
rs781502557 Chr.1: 86425454-86425454 on Build GRCh38
rs931760720 Chr.1: 86425630-86425630 on Build GRCh38
rs758809611 Chr.1: 86425311-86425311 on Build GRCh38
rs756273740 Chr.1: 86425460-86425460 on Build GRCh38
rs1036156349 Chr.1: 86425325-86425325 on Build GRCh38
rs976930114 Chr.1: 86425602-86425602 on Build GRCh38
rs750690143 Chr.1: 86425459-86425459 on Build GRCh38
rs773217484 Chr.1: 86425380-86425380 on Build GRCh38
rs781410301 Chr.1: 86425317-86425317 on Build GRCh38
rs754555450 Chr.1: 86425288-86425288 on Build GRCh38
rs946039768 Chr.1: 86425423-86425423 on Build GRCh38
rs887660032 Chr.1: 86425292-86425292 on Build GRCh38
rs764852305 Chr.1: 86425410-86425410 on Build GRCh38
rs747368671 Chr.1: 86425439-86425440 on Build GRCh38
rs778397588 Chr.1: 86425545-86425545 on Build GRCh38
rs768722488 Chr.1: 86425489-86425489 on Build GRCh38
rs538983082 Chr.1: 86425735-86425735 on Build GRCh38
rs780243230 Chr.1: 86425470-86425470 on Build GRCh38
rs1041735101 Chr.1: 86425469-86425469 on Build GRCh38
rs1036271934 Chr.1: 86425698-86425698 on Build GRCh38
rs548308822 Chr.1: 86425335-86425335 on Build GRCh38
rs776546003 Chr.1: 86425504-86425504 on Build GRCh38

View Full Product Details