Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 100192525-100193020 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DBT
Entrez Gene ID
1629
Gene Name
dihydrolipoamide branched chain transacylase E2
Gene Aliases
BCATE2, BCKAD-E2, BCKADE2, BCOADC-E2, E2, E2B
UniGene
Hs.709187
Gene Chromosome Location
Chr.1: 100186922-100249864 on Build GRCh38
Transcripts
NM_001918.3

SNP Detail

SNP ID Chromosome Location
rs189112426 Chr.1: 100192853-100192853 on Build GRCh38
rs940693198 Chr.1: 100192856-100192856 on Build GRCh38
rs1040388116 Chr.1: 100192887-100192887 on Build GRCh38
rs886044949 Chr.1: 100192800-100192800 on Build GRCh38
rs958295647 Chr.1: 100192633-100192633 on Build GRCh38
rs758437531 Chr.1: 100192885-100192887 on Build GRCh38
rs779084294 Chr.1: 100192821-100192821 on Build GRCh38
rs549556884 Chr.1: 100192755-100192755 on Build GRCh38
rs971713115 Chr.1: 100192707-100192707 on Build GRCh38
rs1039501817 Chr.1: 100192849-100192849 on Build GRCh38
rs1032690307 Chr.1: 100192634-100192634 on Build GRCh38
rs35766227 Chr.1: 100192840-100192840 on Build GRCh38
rs951086952 Chr.1: 100192742-100192742 on Build GRCh38
rs917188058 Chr.1: 100192679-100192679 on Build GRCh38
rs532058526 Chr.1: 100192794-100192794 on Build GRCh38
rs979999111 Chr.1: 100192722-100192722 on Build GRCh38
rs886044950 Chr.1: 100192916-100192917 on Build GRCh38
rs904028975 Chr.1: 100192978-100192978 on Build GRCh38
rs999540297 Chr.1: 100192598-100192599 on Build GRCh38
rs903456830 Chr.1: 100192573-100192585 on Build GRCh38
rs999909646 Chr.1: 100192956-100192956 on Build GRCh38
rs755229964 Chr.1: 100192603-100192603 on Build GRCh38
rs930457587 Chr.1: 100192783-100192783 on Build GRCh38
rs551877714 Chr.1: 100192941-100192941 on Build GRCh38
rs1052460576 Chr.1: 100192966-100192966 on Build GRCh38
rs754461561 Chr.1: 100192734-100192734 on Build GRCh38
rs1050661116 Chr.1: 100192959-100192959 on Build GRCh38
rs921132757 Chr.1: 100192935-100192935 on Build GRCh38
rs541279304 Chr.1: 100192669-100192669 on Build GRCh38
rs1011197393 Chr.1: 100192996-100192996 on Build GRCh38
rs894050029 Chr.1: 100192990-100192990 on Build GRCh38
rs890558109 Chr.1: 100192946-100192946 on Build GRCh38
rs376749142 Chr.1: 100192777-100192777 on Build GRCh38
rs746277434 Chr.1: 100192943-100192943 on Build GRCh38
rs996831131 Chr.1: 100192920-100192920 on Build GRCh38
rs1029570357 Chr.1: 100192565-100192565 on Build GRCh38
rs74465832 Chr.1: 100192716-100192716 on Build GRCh38
rs902128917 Chr.1: 100192893-100192893 on Build GRCh38
rs530051114 Chr.1: 100192731-100192731 on Build GRCh38
rs1011061582 Chr.1: 100192645-100192645 on Build GRCh38
rs953860619 Chr.1: 100192581-100192581 on Build GRCh38
rs955719854 Chr.1: 100192665-100192665 on Build GRCh38

View Full Product Details