Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 58094751-58095217 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLNB
Entrez Gene ID
2317
Gene Name
filamin B
Gene Aliases
ABP-278, ABP-280, AOI, FH1, FLN-B, FLN1L, LRS1, SCT, TABP, TAP
UniGene
Hs.476448
Gene Chromosome Location
Chr.3: 58008400-58172255 on Build GRCh38
Transcripts
NM_001164317.1 NM_001164318.1 NM_001164319.1 NM_001457.3 XM_005264978.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7133381 c.793G>A p.E265K Chr.3: 58094841-58094841 on Build GRCh38
6991164 c.891A>G p.E297E Chr.3: 58094939-58094939 on Build GRCh38
6901062 c.851C>T p.A284V Chr.3: 58094899-58094899 on Build GRCh38
3974539 c.849C>T p.S283S Chr.3: 58094897-58094897 on Build GRCh38
6664980 c.850G>A p.A284T Chr.3: 58094898-58094898 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs971791969 Chr.3: 58095196-58095196 on Build GRCh38
rs764368324 Chr.3: 58094913-58094913 on Build GRCh38
rs75470177 Chr.3: 58094837-58094837 on Build GRCh38
rs780494761 Chr.3: 58094867-58094867 on Build GRCh38
rs887536271 Chr.3: 58095080-58095080 on Build GRCh38
rs184295638 Chr.3: 58094798-58094798 on Build GRCh38
rs771766815 Chr.3: 58094892-58094892 on Build GRCh38
rs753798350 Chr.3: 58094795-58094795 on Build GRCh38
rs886058760 Chr.3: 58094901-58094901 on Build GRCh38
rs760304957 Chr.3: 58094896-58094896 on Build GRCh38
rs149185745 Chr.3: 58095123-58095123 on Build GRCh38
rs7623314 Chr.3: 58094840-58094840 on Build GRCh38
rs771534892 Chr.3: 58095029-58095029 on Build GRCh38
rs758651741 Chr.3: 58094819-58094819 on Build GRCh38
rs1040277720 Chr.3: 58095014-58095014 on Build GRCh38
rs749717676 Chr.3: 58094989-58094989 on Build GRCh38
rs901727042 Chr.3: 58095017-58095017 on Build GRCh38
rs757322226 Chr.3: 58094845-58094845 on Build GRCh38
rs772982798 Chr.3: 58094895-58094895 on Build GRCh38
rs545783631 Chr.3: 58095099-58095099 on Build GRCh38
rs150052174 Chr.3: 58095007-58095007 on Build GRCh38
rs776285380 Chr.3: 58095005-58095005 on Build GRCh38
rs770546915 Chr.3: 58094897-58094897 on Build GRCh38
rs771580553 Chr.3: 58094787-58094787 on Build GRCh38
rs751698758 Chr.3: 58094844-58094844 on Build GRCh38
rs780363553 Chr.3: 58094967-58094967 on Build GRCh38
rs1031667584 Chr.3: 58095164-58095164 on Build GRCh38
rs35462228 Chr.3: 58095165-58095166 on Build GRCh38
rs767605044 Chr.3: 58094929-58094929 on Build GRCh38
rs756510662 Chr.3: 58095081-58095081 on Build GRCh38
rs971618722 Chr.3: 58095143-58095143 on Build GRCh38
rs750107682 Chr.3: 58094844-58094844 on Build GRCh38
rs765107865 Chr.3: 58094799-58094799 on Build GRCh38
rs576908333 Chr.3: 58095097-58095097 on Build GRCh38
rs145036794 Chr.3: 58094856-58094856 on Build GRCh38
rs147844614 Chr.3: 58094898-58094898 on Build GRCh38
rs749545528 Chr.3: 58094873-58094873 on Build GRCh38
rs751747906 Chr.3: 58094919-58094919 on Build GRCh38
rs999400916 Chr.3: 58095112-58095112 on Build GRCh38
rs757375169 Chr.3: 58094920-58094920 on Build GRCh38
rs940499590 Chr.3: 58094934-58094934 on Build GRCh38
rs750807613 Chr.3: 58094781-58094781 on Build GRCh38
rs879503470 Chr.3: 58095122-58095122 on Build GRCh38
rs765017696 Chr.3: 58094900-58094900 on Build GRCh38
rs367916093 Chr.3: 58095126-58095126 on Build GRCh38
rs572680271 Chr.3: 58095162-58095162 on Build GRCh38
rs533627376 Chr.3: 58095114-58095114 on Build GRCh38
rs928992134 Chr.3: 58095018-58095018 on Build GRCh38
rs1005678949 Chr.3: 58094777-58094777 on Build GRCh38
rs770194540 Chr.3: 58094794-58094794 on Build GRCh38
rs1024574496 Chr.3: 58095138-58095138 on Build GRCh38
rs761549302 Chr.3: 58094785-58094785 on Build GRCh38
rs377452206 Chr.3: 58094998-58094998 on Build GRCh38
rs141151998 Chr.3: 58094912-58094912 on Build GRCh38
rs112902485 Chr.3: 58094833-58094833 on Build GRCh38
rs114460813 Chr.3: 58095060-58095060 on Build GRCh38
rs570016927 Chr.3: 58094791-58094791 on Build GRCh38
rs750429255 Chr.3: 58094957-58094957 on Build GRCh38
rs1012954255 Chr.3: 58095139-58095139 on Build GRCh38
rs369429040 Chr.3: 58094990-58094990 on Build GRCh38
rs756644134 Chr.3: 58094960-58094960 on Build GRCh38
rs777620894 Chr.3: 58094993-58094993 on Build GRCh38
rs7644723 Chr.3: 58094778-58094778 on Build GRCh38
rs201495856 Chr.3: 58094792-58094792 on Build GRCh38
rs770671839 Chr.3: 58094999-58094999 on Build GRCh38
rs752395354 Chr.3: 58094810-58094810 on Build GRCh38
rs746351342 Chr.3: 58094854-58094854 on Build GRCh38

View Full Product Details