Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 117627806-117628322 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM46C
Entrez Gene ID
54855
Gene Name
family with sequence similarity 46 member C
Gene Aliases
-
UniGene
Hs.356216
Gene Chromosome Location
Chr.1: 117605982-117628389 on Build GRCh38
Transcripts
NM_017709.3

SNP Detail

SNP ID Chromosome Location
rs939683873 Chr.1: 117628076-117628076 on Build GRCh38
rs1042711436 Chr.1: 117628196-117628196 on Build GRCh38
rs1032506181 Chr.1: 117628015-117628015 on Build GRCh38
rs1038203758 Chr.1: 117628122-117628122 on Build GRCh38
rs778620117 Chr.1: 117628273-117628274 on Build GRCh38
rs900074376 Chr.1: 117628237-117628237 on Build GRCh38
rs756463446 Chr.1: 117627926-117627926 on Build GRCh38
rs1016281766 Chr.1: 117628266-117628266 on Build GRCh38
rs398053386 Chr.1: 117627863-117627864 on Build GRCh38
rs1025744562 Chr.1: 117627892-117627892 on Build GRCh38
rs927215352 Chr.1: 117628074-117628074 on Build GRCh38
rs919705502 Chr.1: 117628083-117628083 on Build GRCh38
rs193094433 Chr.1: 117628233-117628233 on Build GRCh38
rs1009704607 Chr.1: 117628283-117628283 on Build GRCh38
rs937178846 Chr.1: 117627851-117627851 on Build GRCh38
rs564022764 Chr.1: 117628098-117628098 on Build GRCh38
rs754985412 Chr.1: 117627847-117627848 on Build GRCh38
rs988480364 Chr.1: 117628037-117628037 on Build GRCh38
rs546472977 Chr.1: 117628265-117628265 on Build GRCh38
rs1002062485 Chr.1: 117627846-117627846 on Build GRCh38
rs956875989 Chr.1: 117628020-117628020 on Build GRCh38
rs1008656467 Chr.1: 117627908-117627909 on Build GRCh38
rs536265001 Chr.1: 117628168-117628168 on Build GRCh38
rs937297063 Chr.1: 117628109-117628109 on Build GRCh38
rs937738012 Chr.1: 117628090-117628090 on Build GRCh38
rs1033076804 Chr.1: 117627953-117627953 on Build GRCh38
rs994288873 Chr.1: 117627866-117627866 on Build GRCh38
rs891232273 Chr.1: 117628189-117628189 on Build GRCh38
rs973713839 Chr.1: 117628080-117628080 on Build GRCh38
rs955740127 Chr.1: 117628264-117628264 on Build GRCh38
rs975585698 Chr.1: 117627971-117627971 on Build GRCh38
rs778258294 Chr.1: 117628023-117628023 on Build GRCh38
rs898581456 Chr.1: 117627865-117627866 on Build GRCh38
rs71766466 Chr.1: 117627845-117627846 on Build GRCh38
rs1055009252 Chr.1: 117628108-117628108 on Build GRCh38
rs898300293 Chr.1: 117628147-117628147 on Build GRCh38
rs1028371359 Chr.1: 117628262-117628262 on Build GRCh38
rs1008209222 Chr.1: 117628204-117628204 on Build GRCh38
rs998837040 Chr.1: 117628242-117628242 on Build GRCh38
rs746150388 Chr.1: 117628294-117628294 on Build GRCh38
rs189068201 Chr.1: 117628066-117628066 on Build GRCh38
rs865964653 Chr.1: 117628078-117628078 on Build GRCh38
rs370700940 Chr.1: 117628167-117628167 on Build GRCh38
rs562495138 Chr.1: 117627980-117627980 on Build GRCh38
rs949754483 Chr.1: 117627845-117627850 on Build GRCh38
rs189796564 Chr.1: 117627863-117627863 on Build GRCh38
rs996889246 Chr.1: 117628256-117628256 on Build GRCh38
rs1013920513 Chr.1: 117627865-117627865 on Build GRCh38
rs182091336 Chr.1: 117627887-117627887 on Build GRCh38
rs955527653 Chr.1: 117628026-117628026 on Build GRCh38
rs753447359 Chr.1: 117628146-117628146 on Build GRCh38
rs912865767 Chr.1: 117627844-117627845 on Build GRCh38
rs1047146890 Chr.1: 117628170-117628170 on Build GRCh38
rs967790747 Chr.1: 117627901-117627901 on Build GRCh38
rs569195195 Chr.1: 117628068-117628072 on Build GRCh38
rs778932719 Chr.1: 117628253-117628253 on Build GRCh38
rs979619825 Chr.1: 117627976-117627976 on Build GRCh38
rs1045627809 Chr.1: 117627847-117627848 on Build GRCh38
rs1054954442 Chr.1: 117627848-117627848 on Build GRCh38
rs575186177 Chr.1: 117628063-117628063 on Build GRCh38
rs544239656 Chr.1: 117627964-117627964 on Build GRCh38
rs893260938 Chr.1: 117628255-117628255 on Build GRCh38
rs573352828 Chr.1: 117627835-117627835 on Build GRCh38
rs778517212 Chr.1: 117627845-117627848 on Build GRCh38
rs555858305 Chr.1: 117627883-117627883 on Build GRCh38
rs749781442 Chr.1: 117628046-117628046 on Build GRCh38
rs949719200 Chr.1: 117628062-117628063 on Build GRCh38

View Full Product Details