Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 69960739-69961249 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSN2
Entrez Gene ID
1447
Gene Name
casein beta
Gene Aliases
CASB
UniGene
Hs.2242
Gene Chromosome Location
Chr.4: 69955256-69965762 on Build GRCh38
Transcripts
NM_001302770.1 NM_001891.3 XM_017007760.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1694476 c.49G>A p.E17K Chr.4: 69960947-69960947 on Build GRCh38
734862 c.41T>C p.L14P Chr.4: 69960955-69960955 on Build GRCh38
5060439 c.6G>T p.K2N Chr.4: 69960990-69960990 on Build GRCh38
1328428 c.45A>C p.A15A Chr.4: 69960951-69960951 on Build GRCh38
5763049 c.12C>T p.L4L Chr.4: 69960984-69960984 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs950224956 Chr.4: 69961184-69961184 on Build GRCh38
rs1006189006 Chr.4: 69960935-69960935 on Build GRCh38
rs201625212 Chr.4: 69960985-69960985 on Build GRCh38
rs768004133 Chr.4: 69960963-69960963 on Build GRCh38
rs527415329 Chr.4: 69961208-69961208 on Build GRCh38
rs1015952297 Chr.4: 69960958-69960958 on Build GRCh38
rs542438639 Chr.4: 69961092-69961092 on Build GRCh38
rs1028053998 Chr.4: 69961106-69961106 on Build GRCh38
rs369659969 Chr.4: 69960986-69960986 on Build GRCh38
rs371872668 Chr.4: 69960917-69960917 on Build GRCh38
rs372623101 Chr.4: 69961023-69961023 on Build GRCh38
rs760790800 Chr.4: 69961019-69961020 on Build GRCh38
rs942736502 Chr.4: 69961067-69961067 on Build GRCh38
rs764524368 Chr.4: 69960940-69960940 on Build GRCh38
rs193156979 Chr.4: 69961088-69961088 on Build GRCh38
rs1002557809 Chr.4: 69960826-69960826 on Build GRCh38
rs747892438 Chr.4: 69960981-69960981 on Build GRCh38
rs941625713 Chr.4: 69961207-69961207 on Build GRCh38
rs758287932 Chr.4: 69961027-69961027 on Build GRCh38
rs761107327 Chr.4: 69961003-69961003 on Build GRCh38
rs753274466 Chr.4: 69961009-69961009 on Build GRCh38
rs1023291714 Chr.4: 69961225-69961225 on Build GRCh38
rs893732101 Chr.4: 69961041-69961041 on Build GRCh38
rs879303166 Chr.4: 69961092-69961093 on Build GRCh38
rs778827215 Chr.4: 69960979-69960979 on Build GRCh38
rs554980000 Chr.4: 69960838-69960838 on Build GRCh38
rs984396000 Chr.4: 69960976-69960976 on Build GRCh38
rs562253705 Chr.4: 69960996-69960996 on Build GRCh38
rs371660589 Chr.4: 69960987-69960987 on Build GRCh38
rs745913926 Chr.4: 69960925-69960925 on Build GRCh38
rs200475419 Chr.4: 69960930-69960930 on Build GRCh38
rs981937515 Chr.4: 69960768-69960768 on Build GRCh38
rs544047853 Chr.4: 69960989-69960989 on Build GRCh38
rs752588082 Chr.4: 69961026-69961026 on Build GRCh38
rs755646278 Chr.4: 69960969-69960969 on Build GRCh38
rs777669886 Chr.4: 69961033-69961033 on Build GRCh38
rs374478021 Chr.4: 69960923-69960923 on Build GRCh38
rs898789406 Chr.4: 69960959-69960959 on Build GRCh38
rs749973270 Chr.4: 69960966-69960966 on Build GRCh38
rs1009398639 Chr.4: 69961062-69961062 on Build GRCh38
rs981968616 Chr.4: 69961010-69961010 on Build GRCh38
rs577733993 Chr.4: 69960926-69960926 on Build GRCh38
rs915203518 Chr.4: 69960787-69960787 on Build GRCh38
rs957923741 Chr.4: 69960883-69960883 on Build GRCh38
rs922713171 Chr.4: 69961096-69961096 on Build GRCh38
rs915252164 Chr.4: 69961058-69961058 on Build GRCh38
rs774517687 Chr.4: 69960941-69960941 on Build GRCh38
rs566368562 Chr.4: 69961101-69961102 on Build GRCh38
rs753536657 Chr.4: 69960977-69960977 on Build GRCh38
rs116737887 Chr.4: 69960775-69960775 on Build GRCh38
rs545973689 Chr.4: 69960913-69960913 on Build GRCh38
rs186903559 Chr.4: 69960907-69960907 on Build GRCh38
rs771765527 Chr.4: 69960889-69960889 on Build GRCh38
rs1044933623 Chr.4: 69961194-69961194 on Build GRCh38
rs1009472326 Chr.4: 69961223-69961223 on Build GRCh38
rs1031220827 Chr.4: 69961016-69961016 on Build GRCh38
rs115503476 Chr.4: 69961133-69961133 on Build GRCh38
rs909515527 Chr.4: 69961135-69961135 on Build GRCh38
rs775364490 Chr.4: 69960990-69960990 on Build GRCh38
rs560266090 Chr.4: 69961181-69961181 on Build GRCh38
rs901257581 Chr.4: 69960892-69960892 on Build GRCh38
rs201010990 Chr.4: 69961006-69961006 on Build GRCh38
rs894723581 Chr.4: 69961220-69961220 on Build GRCh38
rs748130959 Chr.4: 69960911-69960911 on Build GRCh38
rs989764782 Chr.4: 69961043-69961043 on Build GRCh38
rs1020782359 Chr.4: 69961066-69961066 on Build GRCh38
rs961628479 Chr.4: 69961171-69961171 on Build GRCh38
rs768059121 Chr.4: 69961001-69961001 on Build GRCh38
rs768895212 Chr.4: 69961162-69961162 on Build GRCh38
rs934064476 Chr.4: 69961115-69961115 on Build GRCh38
rs1047200707 Chr.4: 69960908-69960908 on Build GRCh38
rs576584582 Chr.4: 69960972-69960972 on Build GRCh38
rs1037212150 Chr.4: 69960993-69960993 on Build GRCh38
rs749538846 Chr.4: 69960995-69960995 on Build GRCh38
rs773742805 Chr.4: 69961002-69961002 on Build GRCh38
rs920964836 Chr.4: 69960948-69960948 on Build GRCh38
rs953590187 Chr.4: 69960991-69960991 on Build GRCh38
rs542569883 Chr.4: 69961172-69961172 on Build GRCh38
rs754534286 Chr.4: 69960978-69960978 on Build GRCh38
rs923902079 Chr.4: 69960770-69960770 on Build GRCh38
rs762131883 Chr.4: 69960998-69960998 on Build GRCh38
rs371790632 Chr.4: 69960945-69960945 on Build GRCh38
rs746947012 Chr.4: 69961045-69961045 on Build GRCh38
rs1053802150 Chr.4: 69960784-69960784 on Build GRCh38
rs1011941347 Chr.4: 69961221-69961221 on Build GRCh38
rs972980233 Chr.4: 69961173-69961173 on Build GRCh38
rs975383303 Chr.4: 69961081-69961081 on Build GRCh38
rs942790673 Chr.4: 69960799-69960799 on Build GRCh38
rs768530446 Chr.4: 69960905-69960905 on Build GRCh38
rs113735079 Chr.4: 69961019-69961019 on Build GRCh38

View Full Product Details