Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 112186603-112187119 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C4orf32
Entrez Gene ID
132720
Gene Name
chromosome 4 open reading frame 32
Gene Aliases
-
UniGene
Hs.23439
Gene Chromosome Location
Chr.4: 112145397-112189081 on Build GRCh38
Transcripts
NM_152400.2 XM_017007732.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5763545 c.157G>T p.V53F Chr.4: 112186696-112186696 on Build GRCh38
5455254 c.154-1G>T p.? Chr.4: 112186692-112186692 on Build GRCh38
4957684 c.160G>T p.E54* Chr.4: 112186699-112186699 on Build GRCh38
4328804 c.154-4A>T p.? Chr.4: 112186689-112186689 on Build GRCh38
4187138 c.270G>A p.M90I Chr.4: 112186809-112186809 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768942742 Chr.4: 112186965-112186965 on Build GRCh38
rs1002832619 Chr.4: 112187041-112187041 on Build GRCh38
rs777153892 Chr.4: 112186970-112186970 on Build GRCh38
rs200706775 Chr.4: 112186727-112186727 on Build GRCh38
rs779586463 Chr.4: 112186716-112186716 on Build GRCh38
rs751196818 Chr.4: 112186881-112186881 on Build GRCh38
rs536393611 Chr.4: 112187077-112187077 on Build GRCh38
rs201899963 Chr.4: 112186987-112186987 on Build GRCh38
rs757892606 Chr.4: 112186709-112186709 on Build GRCh38
rs764560269 Chr.4: 112186683-112186683 on Build GRCh38
rs199842368 Chr.4: 112186911-112186911 on Build GRCh38
rs1055215116 Chr.4: 112187070-112187070 on Build GRCh38
rs761025644 Chr.4: 112186817-112186817 on Build GRCh38
rs755242403 Chr.4: 112186653-112186657 on Build GRCh38
rs749027811 Chr.4: 112186674-112186676 on Build GRCh38
rs144235488 Chr.4: 112186909-112186909 on Build GRCh38
rs1050714162 Chr.4: 112186912-112186912 on Build GRCh38
rs1026262414 Chr.4: 112186761-112186761 on Build GRCh38
rs754286283 Chr.4: 112186830-112186830 on Build GRCh38
rs767851700 Chr.4: 112186654-112186654 on Build GRCh38
rs571639524 Chr.4: 112186934-112186934 on Build GRCh38
rs1034406130 Chr.4: 112186763-112186763 on Build GRCh38
rs11548409 Chr.4: 112186733-112186733 on Build GRCh38
rs866694073 Chr.4: 112186776-112186776 on Build GRCh38
rs914714979 Chr.4: 112186802-112186802 on Build GRCh38
rs889540759 Chr.4: 112186915-112186915 on Build GRCh38
rs753172124 Chr.4: 112186663-112186663 on Build GRCh38
rs866555738 Chr.4: 112186823-112186823 on Build GRCh38
rs371585740 Chr.4: 112186674-112186674 on Build GRCh38
rs750004005 Chr.4: 112186688-112186688 on Build GRCh38
rs199993966 Chr.4: 112187061-112187061 on Build GRCh38
rs762336096 Chr.4: 112186973-112186973 on Build GRCh38
rs774419672 Chr.4: 112186994-112186994 on Build GRCh38
rs200998231 Chr.4: 112186851-112186851 on Build GRCh38
rs775678102 Chr.4: 112186954-112186954 on Build GRCh38
rs755957136 Chr.4: 112186929-112186929 on Build GRCh38
rs549594524 Chr.4: 112186680-112186680 on Build GRCh38
rs774436785 Chr.4: 112186778-112186778 on Build GRCh38
rs770981967 Chr.4: 112186771-112186771 on Build GRCh38
rs146826929 Chr.4: 112186757-112186757 on Build GRCh38
rs866170231 Chr.4: 112186694-112186694 on Build GRCh38
rs754765327 Chr.4: 112186720-112186720 on Build GRCh38
rs113499240 Chr.4: 112187023-112187023 on Build GRCh38
rs759831425 Chr.4: 112186650-112186650 on Build GRCh38
rs977682754 Chr.4: 112186839-112186839 on Build GRCh38
rs1026092236 Chr.4: 112186638-112186638 on Build GRCh38
rs923640375 Chr.4: 112186866-112186866 on Build GRCh38
rs777862360 Chr.4: 112186931-112186931 on Build GRCh38
rs772344788 Chr.4: 112186803-112186803 on Build GRCh38
rs770833534 Chr.4: 112186674-112186677 on Build GRCh38
rs536135180 Chr.4: 112187067-112187067 on Build GRCh38
rs201921265 Chr.4: 112186978-112186978 on Build GRCh38
rs191854126 Chr.4: 112187008-112187008 on Build GRCh38
rs958745483 Chr.4: 112186773-112186773 on Build GRCh38
rs751157231 Chr.4: 112186719-112186719 on Build GRCh38
rs765877248 Chr.4: 112186869-112186869 on Build GRCh38
rs903992048 Chr.4: 112186645-112186645 on Build GRCh38
rs538521310 Chr.4: 112186814-112186814 on Build GRCh38
rs777845898 Chr.4: 112186740-112186740 on Build GRCh38
rs772376933 Chr.4: 112186953-112186953 on Build GRCh38
rs761042195 Chr.4: 112186958-112186958 on Build GRCh38
rs779001227 Chr.4: 112186948-112186948 on Build GRCh38
rs376066122 Chr.4: 112186780-112186780 on Build GRCh38
rs1034291316 Chr.4: 112187067-112187067 on Build GRCh38
rs764644257 Chr.4: 112186818-112186818 on Build GRCh38
rs755960529 Chr.4: 112186673-112186674 on Build GRCh38
rs80269620 Chr.4: 112186674-112186674 on Build GRCh38
rs527843136 Chr.4: 112186652-112186652 on Build GRCh38
rs564831717 Chr.4: 112187001-112187001 on Build GRCh38
rs769053933 Chr.4: 112186947-112186947 on Build GRCh38
rs139612441 Chr.4: 112186908-112186908 on Build GRCh38
rs771783437 Chr.4: 112186978-112186979 on Build GRCh38
rs745482731 Chr.4: 112186956-112186959 on Build GRCh38
rs138369870 Chr.4: 112186637-112186637 on Build GRCh38
rs781101378 Chr.4: 112186674-112186675 on Build GRCh38
rs373004886 Chr.4: 112186644-112186644 on Build GRCh38
rs754711443 Chr.4: 112186884-112186884 on Build GRCh38
rs898312653 Chr.4: 112186980-112186980 on Build GRCh38
rs747936613 Chr.4: 112186728-112186728 on Build GRCh38
rs774334194 Chr.4: 112186787-112186787 on Build GRCh38
rs572504068 Chr.4: 112187019-112187019 on Build GRCh38
rs749433746 Chr.4: 112186760-112186760 on Build GRCh38

View Full Product Details