Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 42938461-42938964 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNPY3
Entrez Gene ID
10695
Gene Name
canopy FGF signaling regulator 3
Gene Aliases
CAG4A, ERDA5, PRAT4A, TNRC5
UniGene
Hs.414099
Gene Chromosome Location
Chr.6: 42928002-42939287 on Build GRCh38
Transcripts
NM_001318842.1 NM_001318845.1 NM_006586.4 NR_134880.1 NR_134881.1 NR_134882.1 NR_134888.1 NM_001318856.1 NM_001318857.1 NM_001318858.1 NR_134890.1 NR_134891.1 NR_134892.1

Gene Symbol
CNPY3-GNMT
Entrez Gene ID
107080644
Gene Name
CNPY3-GNMT readthrough
Gene Aliases
-
UniGene
Hs.144914
Gene Chromosome Location
Chr.6: 42929130-42963880 on Build GRCh38
Transcripts
NM_001318842.1 NM_001318845.1 NM_006586.4 NR_134880.1 NR_134881.1 NR_134882.1 NR_134888.1 NM_001318856.1 NM_001318857.1 NM_001318858.1 NR_134890.1 NR_134891.1 NR_134892.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1444595 c.619C>A p.L207M Chr.6: 42938573-42938573 on Build GRCh38
3627438 c.773C>T p.P258L Chr.6: 42938727-42938727 on Build GRCh38
1444596 c.622G>A p.A208T Chr.6: 42938576-42938576 on Build GRCh38
5416563 c.680A>G p.K227R Chr.6: 42938634-42938634 on Build GRCh38
4186137 c.671_672delAG p.K225fs*16 Chr.6: 42938625-42938626 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369994608 Chr.6: 42938505-42938506 on Build GRCh38
rs761175533 Chr.6: 42938563-42938563 on Build GRCh38
rs759180897 Chr.6: 42938626-42938626 on Build GRCh38
rs745314983 Chr.6: 42938844-42938844 on Build GRCh38
rs766741461 Chr.6: 42938777-42938777 on Build GRCh38
rs200953616 Chr.6: 42938665-42938665 on Build GRCh38
rs954399876 Chr.6: 42938923-42938923 on Build GRCh38
rs886596058 Chr.6: 42938817-42938817 on Build GRCh38
rs572963128 Chr.6: 42938866-42938866 on Build GRCh38
rs750565283 Chr.6: 42938844-42938845 on Build GRCh38
rs193021383 Chr.6: 42938630-42938630 on Build GRCh38
rs778069363 Chr.6: 42938811-42938811 on Build GRCh38
rs776350885 Chr.6: 42938745-42938745 on Build GRCh38
rs559424919 Chr.6: 42938849-42938849 on Build GRCh38
rs374869606 Chr.6: 42938721-42938721 on Build GRCh38
rs375246952 Chr.6: 42938591-42938591 on Build GRCh38
rs116255295 Chr.6: 42938503-42938503 on Build GRCh38
rs770826260 Chr.6: 42938843-42938843 on Build GRCh38
rs1014922602 Chr.6: 42938912-42938912 on Build GRCh38
rs768042941 Chr.6: 42938645-42938645 on Build GRCh38
rs563479822 Chr.6: 42938749-42938749 on Build GRCh38
rs764184789 Chr.6: 42938534-42938535 on Build GRCh38
rs151243509 Chr.6: 42938566-42938566 on Build GRCh38
rs1010647785 Chr.6: 42938671-42938671 on Build GRCh38
rs372206190 Chr.6: 42938554-42938554 on Build GRCh38
rs776041923 Chr.6: 42938558-42938558 on Build GRCh38
rs371606876 Chr.6: 42938666-42938666 on Build GRCh38
rs750935131 Chr.6: 42938766-42938766 on Build GRCh38
rs900212242 Chr.6: 42938904-42938904 on Build GRCh38
rs140628883 Chr.6: 42938604-42938604 on Build GRCh38
rs1055503092 Chr.6: 42938760-42938760 on Build GRCh38
rs999323110 Chr.6: 42938743-42938743 on Build GRCh38
rs1037892509 Chr.6: 42938867-42938867 on Build GRCh38
rs776354473 Chr.6: 42938623-42938623 on Build GRCh38
rs746956278 Chr.6: 42938837-42938837 on Build GRCh38
rs557897346 Chr.6: 42938484-42938484 on Build GRCh38
rs567190082 Chr.6: 42938738-42938738 on Build GRCh38
rs542436560 Chr.6: 42938534-42938534 on Build GRCh38
rs577921355 Chr.6: 42938932-42938932 on Build GRCh38
rs1016456544 Chr.6: 42938490-42938490 on Build GRCh38
rs751395318 Chr.6: 42938633-42938635 on Build GRCh38
rs779104003 Chr.6: 42938683-42938683 on Build GRCh38
rs752257311 Chr.6: 42938631-42938631 on Build GRCh38
rs756251267 Chr.6: 42938587-42938587 on Build GRCh38
rs765887645 Chr.6: 42938622-42938622 on Build GRCh38
rs748265663 Chr.6: 42938699-42938699 on Build GRCh38
rs137924295 Chr.6: 42938806-42938807 on Build GRCh38
rs146664776 Chr.6: 42938729-42938729 on Build GRCh38
rs200549681 Chr.6: 42938770-42938770 on Build GRCh38
rs115953673 Chr.6: 42938505-42938505 on Build GRCh38
rs540868724 Chr.6: 42938916-42938916 on Build GRCh38
rs984951273 Chr.6: 42938584-42938584 on Build GRCh38
rs147735655 Chr.6: 42938612-42938612 on Build GRCh38
rs371860465 Chr.6: 42938590-42938590 on Build GRCh38
rs746693743 Chr.6: 42938675-42938675 on Build GRCh38
rs949637249 Chr.6: 42938797-42938797 on Build GRCh38
rs868395570 Chr.6: 42938719-42938719 on Build GRCh38
rs538003610 Chr.6: 42938855-42938855 on Build GRCh38
rs9471969 Chr.6: 42938646-42938646 on Build GRCh38
rs549629737 Chr.6: 42938778-42938778 on Build GRCh38
rs1034954308 Chr.6: 42938785-42938785 on Build GRCh38
rs768775544 Chr.6: 42938600-42938600 on Build GRCh38
rs952016367 Chr.6: 42938898-42938898 on Build GRCh38
rs984856530 Chr.6: 42938910-42938910 on Build GRCh38
rs781518711 Chr.6: 42938560-42938560 on Build GRCh38
rs759209443 Chr.6: 42938746-42938746 on Build GRCh38
rs752764710 Chr.6: 42938803-42938803 on Build GRCh38
rs767956827 Chr.6: 42938762-42938762 on Build GRCh38
rs761738758 Chr.6: 42938764-42938767 on Build GRCh38
rs537322844 Chr.6: 42938617-42938617 on Build GRCh38
rs756925011 Chr.6: 42938705-42938705 on Build GRCh38
rs577182193 Chr.6: 42938649-42938649 on Build GRCh38
rs533937818 Chr.6: 42938933-42938933 on Build GRCh38
rs760537930 Chr.6: 42938621-42938621 on Build GRCh38
rs769480238 Chr.6: 42938753-42938753 on Build GRCh38
rs577867594 Chr.6: 42938488-42938488 on Build GRCh38
rs773097165 Chr.6: 42938620-42938620 on Build GRCh38
rs762473290 Chr.6: 42938756-42938756 on Build GRCh38
rs1024041684 Chr.6: 42938876-42938876 on Build GRCh38
rs752059295 Chr.6: 42938539-42938539 on Build GRCh38
rs77807794 Chr.6: 42938504-42938504 on Build GRCh38
rs774941846 Chr.6: 42938755-42938755 on Build GRCh38
rs569310782 Chr.6: 42938805-42938805 on Build GRCh38
rs71547825 Chr.6: 42938503-42938505 on Build GRCh38

View Full Product Details