Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 17334709-17335236 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AHR
Entrez Gene ID
196
Gene Name
aryl hydrocarbon receptor
Gene Aliases
bHLHe76
UniGene
Hs.171189
Gene Chromosome Location
Chr.7: 17298652-17346151 on Build GRCh38
Transcripts
NM_001621.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3879945 c.928T>C p.Y310H Chr.7: 17334906-17334906 on Build GRCh38
6597050 c.1016G>A p.R339Q Chr.7: 17334994-17334994 on Build GRCh38
3306704 c.951G>A p.T317T Chr.7: 17334929-17334929 on Build GRCh38
3769998 c.989T>C p.M330T Chr.7: 17334967-17334967 on Build GRCh38
5894859 c.932C>T p.T311I Chr.7: 17334910-17334910 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747383234 Chr.7: 17334862-17334862 on Build GRCh38
rs758945560 Chr.7: 17334923-17334923 on Build GRCh38
rs964471158 Chr.7: 17335112-17335112 on Build GRCh38
rs543567818 Chr.7: 17335068-17335068 on Build GRCh38
rs892723975 Chr.7: 17334761-17334761 on Build GRCh38
rs937141789 Chr.7: 17334902-17334902 on Build GRCh38
rs915478762 Chr.7: 17335167-17335167 on Build GRCh38
rs577789382 Chr.7: 17335115-17335115 on Build GRCh38
rs922832794 Chr.7: 17334734-17334734 on Build GRCh38
rs369808533 Chr.7: 17334845-17334845 on Build GRCh38
rs527328319 Chr.7: 17334749-17334749 on Build GRCh38
rs753084329 Chr.7: 17334901-17334901 on Build GRCh38
rs776833733 Chr.7: 17334871-17334871 on Build GRCh38
rs1029948307 Chr.7: 17335170-17335170 on Build GRCh38
rs1029545383 Chr.7: 17334826-17334826 on Build GRCh38
rs771252130 Chr.7: 17335007-17335007 on Build GRCh38
rs759679049 Chr.7: 17335020-17335020 on Build GRCh38
rs904228566 Chr.7: 17335119-17335119 on Build GRCh38
rs186170100 Chr.7: 17335051-17335051 on Build GRCh38
rs999835149 Chr.7: 17335125-17335125 on Build GRCh38
rs951320946 Chr.7: 17335189-17335189 on Build GRCh38
rs1005494075 Chr.7: 17335083-17335087 on Build GRCh38
rs748008207 Chr.7: 17335000-17335000 on Build GRCh38
rs1057043375 Chr.7: 17335071-17335071 on Build GRCh38
rs762230170 Chr.7: 17335044-17335044 on Build GRCh38
rs764709620 Chr.7: 17335036-17335036 on Build GRCh38
rs888385964 Chr.7: 17335069-17335069 on Build GRCh38
rs760768902 Chr.7: 17334855-17334855 on Build GRCh38
rs552120377 Chr.7: 17334799-17334799 on Build GRCh38
rs997933221 Chr.7: 17335145-17335145 on Build GRCh38
rs769847453 Chr.7: 17334892-17334892 on Build GRCh38
rs926713019 Chr.7: 17335052-17335052 on Build GRCh38
rs777062899 Chr.7: 17335014-17335014 on Build GRCh38
rs199987821 Chr.7: 17334838-17334838 on Build GRCh38
rs571785328 Chr.7: 17334867-17334867 on Build GRCh38
rs548579736 Chr.7: 17334786-17334786 on Build GRCh38
rs765837917 Chr.7: 17334897-17334897 on Build GRCh38
rs1009873070 Chr.7: 17334778-17334778 on Build GRCh38
rs771391655 Chr.7: 17334868-17334869 on Build GRCh38
rs201375419 Chr.7: 17335034-17335034 on Build GRCh38
rs537797986 Chr.7: 17335129-17335129 on Build GRCh38
rs756880752 Chr.7: 17334944-17334944 on Build GRCh38
rs763316804 Chr.7: 17335035-17335035 on Build GRCh38
rs13223444 Chr.7: 17335096-17335096 on Build GRCh38
rs147123367 Chr.7: 17335120-17335121 on Build GRCh38
rs768669484 Chr.7: 17334983-17334983 on Build GRCh38
rs1004566449 Chr.7: 17335197-17335197 on Build GRCh38
rs531329767 Chr.7: 17334822-17334822 on Build GRCh38
rs994419821 Chr.7: 17334991-17334991 on Build GRCh38
rs892681071 Chr.7: 17335075-17335075 on Build GRCh38
rs963821484 Chr.7: 17334809-17334809 on Build GRCh38
rs775877254 Chr.7: 17335060-17335060 on Build GRCh38
rs905388665 Chr.7: 17334741-17334741 on Build GRCh38
rs899597756 Chr.7: 17335131-17335131 on Build GRCh38
rs1056695952 Chr.7: 17334754-17334754 on Build GRCh38
rs114930376 Chr.7: 17334802-17334802 on Build GRCh38
rs566364753 Chr.7: 17334980-17334980 on Build GRCh38
rs765339440 Chr.7: 17335029-17335029 on Build GRCh38
rs190667274 Chr.7: 17334971-17334971 on Build GRCh38
rs780863050 Chr.7: 17334955-17334955 on Build GRCh38
rs773968942 Chr.7: 17334996-17334996 on Build GRCh38
rs935503094 Chr.7: 17334736-17334736 on Build GRCh38
rs904271448 Chr.7: 17334790-17334790 on Build GRCh38
rs751838949 Chr.7: 17335039-17335039 on Build GRCh38
rs201806511 Chr.7: 17334929-17334929 on Build GRCh38
rs1018043425 Chr.7: 17334803-17334803 on Build GRCh38
rs766733705 Chr.7: 17334868-17334868 on Build GRCh38
rs976565624 Chr.7: 17334824-17334824 on Build GRCh38
rs181963856 Chr.7: 17334981-17334981 on Build GRCh38
rs376505668 Chr.7: 17334967-17334967 on Build GRCh38

View Full Product Details