Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 23643760-23644272 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC126
Entrez Gene ID
90693
Gene Name
coiled-coil domain containing 126
Gene Aliases
-
UniGene
Hs.232296
Gene Chromosome Location
Chr.7: 23597333-23644708 on Build GRCh38
Transcripts
NM_138771.3 XM_017012775.1 XM_017012776.1

SNP Detail

SNP ID Chromosome Location
rs571411786 Chr.7: 23644180-23644180 on Build GRCh38
rs1009203407 Chr.7: 23644139-23644139 on Build GRCh38
rs73287855 Chr.7: 23643948-23643948 on Build GRCh38
rs975716812 Chr.7: 23644164-23644164 on Build GRCh38
rs566993564 Chr.7: 23644227-23644227 on Build GRCh38
rs148774340 Chr.7: 23644139-23644140 on Build GRCh38
rs1048501001 Chr.7: 23643977-23643977 on Build GRCh38
rs192142129 Chr.7: 23644064-23644064 on Build GRCh38
rs1031999093 Chr.7: 23643869-23643869 on Build GRCh38
rs868747351 Chr.7: 23643784-23643784 on Build GRCh38
rs757337458 Chr.7: 23643834-23643834 on Build GRCh38
rs912779782 Chr.7: 23643901-23643901 on Build GRCh38
rs940116766 Chr.7: 23644233-23644233 on Build GRCh38
rs1038935762 Chr.7: 23644020-23644020 on Build GRCh38
rs78674075 Chr.7: 23643859-23643859 on Build GRCh38
rs547269668 Chr.7: 23644170-23644170 on Build GRCh38
rs887131671 Chr.7: 23643786-23643786 on Build GRCh38
rs184662722 Chr.7: 23644003-23644003 on Build GRCh38
rs1037018494 Chr.7: 23643957-23643957 on Build GRCh38
rs896373476 Chr.7: 23644057-23644057 on Build GRCh38
rs970244830 Chr.7: 23644083-23644083 on Build GRCh38
rs561401132 Chr.7: 23644048-23644048 on Build GRCh38
rs908592142 Chr.7: 23644196-23644196 on Build GRCh38
rs746117095 Chr.7: 23643943-23643943 on Build GRCh38
rs1001120919 Chr.7: 23644110-23644110 on Build GRCh38
rs758013764 Chr.7: 23644071-23644071 on Build GRCh38
rs1013797384 Chr.7: 23643851-23643851 on Build GRCh38
rs1004741450 Chr.7: 23643792-23643792 on Build GRCh38
rs905048576 Chr.7: 23644084-23644084 on Build GRCh38
rs559367277 Chr.7: 23643868-23643870 on Build GRCh38
rs890291025 Chr.7: 23644012-23644012 on Build GRCh38
rs570099135 Chr.7: 23644133-23644133 on Build GRCh38
rs550127326 Chr.7: 23644081-23644081 on Build GRCh38
rs777292410 Chr.7: 23644078-23644078 on Build GRCh38
rs986902905 Chr.7: 23644242-23644242 on Build GRCh38
rs201728451 Chr.7: 23643810-23643810 on Build GRCh38
rs910160084 Chr.7: 23643870-23643870 on Build GRCh38
rs1051626501 Chr.7: 23644004-23644004 on Build GRCh38
rs548018565 Chr.7: 23644117-23644124 on Build GRCh38
rs544569228 Chr.7: 23643785-23643785 on Build GRCh38
rs987824867 Chr.7: 23643886-23643886 on Build GRCh38
rs984711382 Chr.7: 23644178-23644178 on Build GRCh38
rs965629054 Chr.7: 23643919-23643919 on Build GRCh38
rs774898939 Chr.7: 23644193-23644193 on Build GRCh38
rs866280203 Chr.7: 23644089-23644089 on Build GRCh38
rs372643049 Chr.7: 23643972-23643972 on Build GRCh38
rs1007837077 Chr.7: 23644015-23644015 on Build GRCh38
rs2109348 Chr.7: 23644160-23644160 on Build GRCh38
rs749611363 Chr.7: 23644159-23644159 on Build GRCh38
rs918607174 Chr.7: 23643964-23643964 on Build GRCh38
rs1013965519 Chr.7: 23644067-23644067 on Build GRCh38
rs921418126 Chr.7: 23644166-23644166 on Build GRCh38
rs140730198 Chr.7: 23644046-23644046 on Build GRCh38
rs975981166 Chr.7: 23643930-23643930 on Build GRCh38
rs1000659301 Chr.7: 23643868-23643868 on Build GRCh38
rs753748972 Chr.7: 23644104-23644108 on Build GRCh38
rs559309125 Chr.7: 23644026-23644026 on Build GRCh38
rs190666864 Chr.7: 23644034-23644034 on Build GRCh38
rs1000131456 Chr.7: 23644038-23644038 on Build GRCh38
rs896422551 Chr.7: 23643821-23643821 on Build GRCh38
rs904473038 Chr.7: 23643907-23643907 on Build GRCh38

View Full Product Details