Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 116862307-116862802 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPZA2
Entrez Gene ID
830
Gene Name
capping actin protein of muscle Z-line alpha subunit 2
Gene Aliases
CAPPA2, CAPZ
UniGene
Hs.446123
Gene Chromosome Location
Chr.7: 116862478-116919261 on Build GRCh38
Transcripts
NM_006136.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6411341 c.16G>A p.E6K Chr.7: 116862627-116862627 on Build GRCh38
1084416 c.32A>T p.E11V Chr.7: 116862643-116862643 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1033136875 Chr.7: 116862540-116862540 on Build GRCh38
rs775528591 Chr.7: 116862589-116862589 on Build GRCh38
rs1001408638 Chr.7: 116862426-116862426 on Build GRCh38
rs913855535 Chr.7: 116862412-116862412 on Build GRCh38
rs1037327701 Chr.7: 116862368-116862368 on Build GRCh38
rs998363335 Chr.7: 116862374-116862374 on Build GRCh38
rs926202896 Chr.7: 116862760-116862760 on Build GRCh38
rs751753715 Chr.7: 116862599-116862599 on Build GRCh38
rs140352106 Chr.7: 116862767-116862767 on Build GRCh38
rs187133233 Chr.7: 116862382-116862382 on Build GRCh38
rs767575888 Chr.7: 116862617-116862617 on Build GRCh38
rs939739684 Chr.7: 116862494-116862494 on Build GRCh38
rs1049935972 Chr.7: 116862510-116862510 on Build GRCh38
rs926432903 Chr.7: 116862441-116862441 on Build GRCh38
rs909622322 Chr.7: 116862493-116862493 on Build GRCh38
rs959276129 Chr.7: 116862584-116862584 on Build GRCh38
rs370239082 Chr.7: 116862577-116862577 on Build GRCh38
rs1048168345 Chr.7: 116862476-116862476 on Build GRCh38
rs527866796 Chr.7: 116862682-116862682 on Build GRCh38
rs778810842 Chr.7: 116862575-116862577 on Build GRCh38
rs974985766 Chr.7: 116862544-116862544 on Build GRCh38
rs1025742376 Chr.7: 116862671-116862671 on Build GRCh38
rs557750264 Chr.7: 116862394-116862394 on Build GRCh38
rs1033497714 Chr.7: 116862390-116862390 on Build GRCh38
rs756390226 Chr.7: 116862659-116862659 on Build GRCh38
rs374586972 Chr.7: 116862660-116862660 on Build GRCh38
rs962395210 Chr.7: 116862474-116862474 on Build GRCh38
rs777582182 Chr.7: 116862575-116862580 on Build GRCh38
rs956087818 Chr.7: 116862393-116862393 on Build GRCh38
rs891898747 Chr.7: 116862392-116862392 on Build GRCh38
rs926228360 Chr.7: 116862563-116862563 on Build GRCh38
rs531999504 Chr.7: 116862712-116862712 on Build GRCh38
rs528189877 Chr.7: 116862332-116862332 on Build GRCh38
rs531781112 Chr.7: 116862375-116862375 on Build GRCh38
rs1036665984 Chr.7: 116862499-116862499 on Build GRCh38
rs564548452 Chr.7: 116862690-116862690 on Build GRCh38
rs907238787 Chr.7: 116862782-116862782 on Build GRCh38
rs955317114 Chr.7: 116862498-116862498 on Build GRCh38
rs753874591 Chr.7: 116862568-116862569 on Build GRCh38
rs937830589 Chr.7: 116862457-116862457 on Build GRCh38
rs763007525 Chr.7: 116862590-116862590 on Build GRCh38
rs774168216 Chr.7: 116862433-116862433 on Build GRCh38
rs539656548 Chr.7: 116862389-116862389 on Build GRCh38
rs970583029 Chr.7: 116862430-116862430 on Build GRCh38
rs1033540522 Chr.7: 116862580-116862580 on Build GRCh38
rs770542957 Chr.7: 116862339-116862339 on Build GRCh38
rs750662131 Chr.7: 116862643-116862643 on Build GRCh38
rs981911028 Chr.7: 116862439-116862439 on Build GRCh38
rs1054998240 Chr.7: 116862746-116862746 on Build GRCh38
rs988793810 Chr.7: 116862771-116862771 on Build GRCh38
rs1000257730 Chr.7: 116862386-116862386 on Build GRCh38
rs529356453 Chr.7: 116862776-116862776 on Build GRCh38
rs781401448 Chr.7: 116862574-116862575 on Build GRCh38
rs1024105635 Chr.7: 116862398-116862398 on Build GRCh38
rs115038026 Chr.7: 116862688-116862688 on Build GRCh38
rs954129635 Chr.7: 116862383-116862383 on Build GRCh38
rs1001489055 Chr.7: 116862783-116862783 on Build GRCh38
rs1050027648 Chr.7: 116862695-116862695 on Build GRCh38
rs936216319 Chr.7: 116862714-116862714 on Build GRCh38
rs918364496 Chr.7: 116862353-116862353 on Build GRCh38
rs1025371860 Chr.7: 116862495-116862495 on Build GRCh38
rs890031735 Chr.7: 116862704-116862704 on Build GRCh38
rs955114682 Chr.7: 116862681-116862681 on Build GRCh38
rs762112467 Chr.7: 116862601-116862601 on Build GRCh38
rs1000752982 Chr.7: 116862527-116862527 on Build GRCh38
rs764248285 Chr.7: 116862597-116862597 on Build GRCh38
rs754710280 Chr.7: 116862571-116862572 on Build GRCh38
rs901428234 Chr.7: 116862503-116862503 on Build GRCh38
rs550506901 Chr.7: 116862739-116862739 on Build GRCh38
rs192510686 Chr.7: 116862574-116862574 on Build GRCh38
rs535913703 Chr.7: 116862354-116862354 on Build GRCh38
rs969913836 Chr.7: 116862415-116862415 on Build GRCh38
rs576312341 Chr.7: 116862480-116862480 on Build GRCh38
rs573311102 Chr.7: 116862573-116862573 on Build GRCh38
rs559068359 Chr.7: 116862588-116862588 on Build GRCh38
rs929476903 Chr.7: 116862365-116862365 on Build GRCh38
rs891867383 Chr.7: 116862554-116862554 on Build GRCh38
rs998415674 Chr.7: 116862506-116862506 on Build GRCh38
rs374052696 Chr.7: 116862571-116862571 on Build GRCh38
rs1024272480 Chr.7: 116862780-116862780 on Build GRCh38
rs1010243577 Chr.7: 116862555-116862555 on Build GRCh38
rs889986793 Chr.7: 116862526-116862526 on Build GRCh38
rs901419913 Chr.7: 116862372-116862372 on Build GRCh38
rs922235320 Chr.7: 116862684-116862684 on Build GRCh38
rs543332166 Chr.7: 116862564-116862564 on Build GRCh38
rs913226323 Chr.7: 116862773-116862773 on Build GRCh38
rs11540705 Chr.7: 116862567-116862567 on Build GRCh38

View Full Product Details