Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 49029845-49030349 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TFE3
Entrez Gene ID
7030
Gene Name
transcription factor binding to IGHM enhancer 3
Gene Aliases
RCCP2, RCCX1, TFEA, bHLHe33
UniGene
Hs.730740
Gene Chromosome Location
Chr.X: 49028726-49043517 on Build GRCh38
Transcripts
NM_001282142.1 NM_006521.5

SNP Detail

SNP ID Chromosome Location
rs782641526 Chr.X: 49029963-49029963 on Build GRCh38
rs782274424 Chr.X: 49029948-49029948 on Build GRCh38
rs781800168 Chr.X: 49030069-49030069 on Build GRCh38
rs782366021 Chr.X: 49029871-49029871 on Build GRCh38
rs782528810 Chr.X: 49030071-49030071 on Build GRCh38
rs374323699 Chr.X: 49029977-49029977 on Build GRCh38
rs370070686 Chr.X: 49029914-49029914 on Build GRCh38
rs781959074 Chr.X: 49029956-49029956 on Build GRCh38
rs782782345 Chr.X: 49030192-49030192 on Build GRCh38
rs781999310 Chr.X: 49029876-49029876 on Build GRCh38
rs781858237 Chr.X: 49030074-49030074 on Build GRCh38
rs1020716932 Chr.X: 49030270-49030270 on Build GRCh38
rs782484357 Chr.X: 49029967-49029967 on Build GRCh38
rs782701013 Chr.X: 49030068-49030068 on Build GRCh38
rs782608594 Chr.X: 49030268-49030268 on Build GRCh38
rs781825487 Chr.X: 49030209-49030209 on Build GRCh38
rs782082061 Chr.X: 49029881-49029881 on Build GRCh38
rs782679964 Chr.X: 49030249-49030249 on Build GRCh38
rs782516347 Chr.X: 49030080-49030080 on Build GRCh38
rs782027549 Chr.X: 49029995-49029995 on Build GRCh38
rs782037153 Chr.X: 49030022-49030022 on Build GRCh38
rs782577298 Chr.X: 49029937-49029937 on Build GRCh38
rs782428510 Chr.X: 49030149-49030149 on Build GRCh38
rs782272958 Chr.X: 49030170-49030170 on Build GRCh38
rs782431595 Chr.X: 49029925-49029925 on Build GRCh38
rs781996328 Chr.X: 49030323-49030323 on Build GRCh38
rs781970950 Chr.X: 49029989-49029989 on Build GRCh38
rs782027002 Chr.X: 49030193-49030193 on Build GRCh38
rs377088709 Chr.X: 49029993-49029993 on Build GRCh38
rs782769489 Chr.X: 49030241-49030241 on Build GRCh38
rs782285732 Chr.X: 49030294-49030294 on Build GRCh38
rs782432988 Chr.X: 49030092-49030092 on Build GRCh38
rs782089605 Chr.X: 49030059-49030059 on Build GRCh38
rs782351636 Chr.X: 49030171-49030171 on Build GRCh38
rs782114663 Chr.X: 49030199-49030199 on Build GRCh38
rs377533245 Chr.X: 49030190-49030190 on Build GRCh38
rs782006875 Chr.X: 49030047-49030047 on Build GRCh38
rs782177459 Chr.X: 49029992-49029992 on Build GRCh38
rs782228992 Chr.X: 49030091-49030091 on Build GRCh38
rs371723359 Chr.X: 49029912-49029912 on Build GRCh38
rs985698346 Chr.X: 49029887-49029887 on Build GRCh38
rs11542392 Chr.X: 49030081-49030081 on Build GRCh38
rs782766621 Chr.X: 49030031-49030031 on Build GRCh38
rs367827397 Chr.X: 49029894-49029894 on Build GRCh38
rs782347207 Chr.X: 49029921-49029921 on Build GRCh38
rs373846907 Chr.X: 49030083-49030083 on Build GRCh38
rs782012535 Chr.X: 49030269-49030270 on Build GRCh38
rs782548608 Chr.X: 49030242-49030242 on Build GRCh38
rs781859773 Chr.X: 49030252-49030252 on Build GRCh38
rs782597619 Chr.X: 49030106-49030106 on Build GRCh38
rs782070258 Chr.X: 49029896-49029896 on Build GRCh38
rs782339241 Chr.X: 49029983-49029983 on Build GRCh38
rs373045428 Chr.X: 49030275-49030275 on Build GRCh38
rs376015528 Chr.X: 49029913-49029913 on Build GRCh38
rs782283460 Chr.X: 49030128-49030128 on Build GRCh38
rs782152448 Chr.X: 49030230-49030230 on Build GRCh38
rs201049883 Chr.X: 49030213-49030213 on Build GRCh38
rs782376412 Chr.X: 49030296-49030296 on Build GRCh38
rs782776048 Chr.X: 49030206-49030206 on Build GRCh38
rs782159878 Chr.X: 49029865-49029865 on Build GRCh38
rs782722360 Chr.X: 49030153-49030153 on Build GRCh38
rs782750132 Chr.X: 49029893-49029893 on Build GRCh38
rs781993470 Chr.X: 49030163-49030163 on Build GRCh38
rs782551260 Chr.X: 49029956-49029956 on Build GRCh38
rs782529128 Chr.X: 49030257-49030257 on Build GRCh38
rs782670973 Chr.X: 49030292-49030292 on Build GRCh38
rs782320148 Chr.X: 49030189-49030189 on Build GRCh38
rs782314136 Chr.X: 49030276-49030276 on Build GRCh38

View Full Product Details