Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 49030217-49030674 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TFE3
Entrez Gene ID
7030
Gene Name
transcription factor binding to IGHM enhancer 3
Gene Aliases
RCCP2, RCCX1, TFEA, bHLHe33
UniGene
Hs.730740
Gene Chromosome Location
Chr.X: 49028726-49043517 on Build GRCh38
Transcripts
NM_001282142.1 NM_006521.5

SNP Detail

SNP ID Chromosome Location
rs370693170 Chr.X: 49030548-49030548 on Build GRCh38
rs782018183 Chr.X: 49030466-49030466 on Build GRCh38
rs782402484 Chr.X: 49030637-49030637 on Build GRCh38
rs781929782 Chr.X: 49030521-49030521 on Build GRCh38
rs61742676 Chr.X: 49030436-49030436 on Build GRCh38
rs147974460 Chr.X: 49030478-49030478 on Build GRCh38
rs144431753 Chr.X: 49030460-49030460 on Build GRCh38
rs147100760 Chr.X: 49030435-49030435 on Build GRCh38
rs782342716 Chr.X: 49030606-49030606 on Build GRCh38
rs782085281 Chr.X: 49030400-49030400 on Build GRCh38
rs142370390 Chr.X: 49030549-49030549 on Build GRCh38
rs781870660 Chr.X: 49030449-49030449 on Build GRCh38
rs1020716932 Chr.X: 49030270-49030270 on Build GRCh38
rs376250395 Chr.X: 49030523-49030523 on Build GRCh38
rs782182621 Chr.X: 49030632-49030632 on Build GRCh38
rs781836515 Chr.X: 49030531-49030531 on Build GRCh38
rs141702671 Chr.X: 49030413-49030413 on Build GRCh38
rs971474665 Chr.X: 49030448-49030448 on Build GRCh38
rs927318052 Chr.X: 49030488-49030488 on Build GRCh38
rs3027472 Chr.X: 49030563-49030563 on Build GRCh38
rs782608594 Chr.X: 49030268-49030268 on Build GRCh38
rs181120439 Chr.X: 49030424-49030424 on Build GRCh38
rs139889590 Chr.X: 49030576-49030576 on Build GRCh38
rs782679964 Chr.X: 49030249-49030249 on Build GRCh38
rs782142662 Chr.X: 49030522-49030522 on Build GRCh38
rs782358848 Chr.X: 49030401-49030401 on Build GRCh38
rs781996328 Chr.X: 49030323-49030323 on Build GRCh38
rs376043279 Chr.X: 49030421-49030421 on Build GRCh38
rs782280641 Chr.X: 49030555-49030555 on Build GRCh38
rs782769489 Chr.X: 49030241-49030241 on Build GRCh38
rs782285732 Chr.X: 49030294-49030294 on Build GRCh38
rs782553054 Chr.X: 49030441-49030441 on Build GRCh38
rs782115854 Chr.X: 49030420-49030420 on Build GRCh38
rs782256627 Chr.X: 49030446-49030446 on Build GRCh38
rs374474638 Chr.X: 49030439-49030439 on Build GRCh38
rs374071577 Chr.X: 49030612-49030612 on Build GRCh38
rs782753958 Chr.X: 49030440-49030441 on Build GRCh38
rs782029710 Chr.X: 49030638-49030638 on Build GRCh38
rs782004079 Chr.X: 49030469-49030469 on Build GRCh38
rs782638809 Chr.X: 49030574-49030574 on Build GRCh38
rs782740611 Chr.X: 49030530-49030530 on Build GRCh38
rs781924132 Chr.X: 49030402-49030402 on Build GRCh38
rs782329572 Chr.X: 49030464-49030464 on Build GRCh38
rs782068672 Chr.X: 49030406-49030406 on Build GRCh38
rs782690477 Chr.X: 49030465-49030465 on Build GRCh38
rs946614302 Chr.X: 49030562-49030562 on Build GRCh38
rs782012535 Chr.X: 49030269-49030270 on Build GRCh38
rs782548608 Chr.X: 49030242-49030242 on Build GRCh38
rs782384434 Chr.X: 49030468-49030468 on Build GRCh38
rs781859773 Chr.X: 49030252-49030252 on Build GRCh38
rs782312780 Chr.X: 49030624-49030624 on Build GRCh38
rs782790681 Chr.X: 49030412-49030412 on Build GRCh38
rs373045428 Chr.X: 49030275-49030275 on Build GRCh38
rs1050863848 Chr.X: 49030626-49030626 on Build GRCh38
rs782164493 Chr.X: 49030467-49030467 on Build GRCh38
rs986155885 Chr.X: 49030429-49030429 on Build GRCh38
rs782509704 Chr.X: 49030525-49030525 on Build GRCh38
rs782619958 Chr.X: 49030444-49030444 on Build GRCh38
rs997854429 Chr.X: 49030361-49030361 on Build GRCh38
rs782376412 Chr.X: 49030296-49030296 on Build GRCh38
rs782743948 Chr.X: 49030519-49030519 on Build GRCh38
rs1047561 Chr.X: 49030524-49030524 on Build GRCh38
rs782529128 Chr.X: 49030257-49030257 on Build GRCh38
rs782670973 Chr.X: 49030292-49030292 on Build GRCh38
rs782314136 Chr.X: 49030276-49030276 on Build GRCh38

View Full Product Details