Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 228149635-228150136 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GJC2
Entrez Gene ID
57165
Gene Name
gap junction protein gamma 2
Gene Aliases
CX46.6, Cx47, GJA12, HLD2, LMPH1C, PMLDAR, SPG44
UniGene
Hs.100072
Gene Chromosome Location
Chr.1: 228149714-228159826 on Build GRCh38
Transcripts
NM_020435.3

SNP Detail

SNP ID Chromosome Location
rs879322091 Chr.1: 228149811-228149812 on Build GRCh38
rs369926757 Chr.1: 228150060-228150060 on Build GRCh38
rs538876106 Chr.1: 228150094-228150096 on Build GRCh38
rs587776888 Chr.1: 228149860-228149860 on Build GRCh38
rs914099187 Chr.1: 228149687-228149696 on Build GRCh38
rs887535785 Chr.1: 228150016-228150016 on Build GRCh38
rs1048862655 Chr.1: 228149982-228149982 on Build GRCh38
rs760991011 Chr.1: 228149871-228149871 on Build GRCh38
rs569587346 Chr.1: 228149802-228149802 on Build GRCh38
rs970998271 Chr.1: 228149750-228149750 on Build GRCh38
rs573670197 Chr.1: 228150062-228150062 on Build GRCh38
rs779687154 Chr.1: 228149733-228149733 on Build GRCh38
rs749284537 Chr.1: 228149754-228149754 on Build GRCh38
rs940432317 Chr.1: 228149911-228149911 on Build GRCh38
rs975383232 Chr.1: 228149851-228149851 on Build GRCh38
rs1031153234 Chr.1: 228149803-228149803 on Build GRCh38
rs558840197 Chr.1: 228149816-228149816 on Build GRCh38
rs965740911 Chr.1: 228150057-228150057 on Build GRCh38
rs796192430 Chr.1: 228149848-228149848 on Build GRCh38
rs764222633 Chr.1: 228150046-228150046 on Build GRCh38
rs868760509 Chr.1: 228149665-228149665 on Build GRCh38
rs898801887 Chr.1: 228149720-228149720 on Build GRCh38
rs1027368673 Chr.1: 228149772-228149772 on Build GRCh38
rs1052057100 Chr.1: 228149896-228149896 on Build GRCh38
rs538931806 Chr.1: 228149804-228149804 on Build GRCh38
rs1022376419 Chr.1: 228150102-228150102 on Build GRCh38
rs951789349 Chr.1: 228149780-228149780 on Build GRCh38
rs957026254 Chr.1: 228149806-228149806 on Build GRCh38
rs538228757 Chr.1: 228149881-228149881 on Build GRCh38
rs1045660773 Chr.1: 228149921-228149921 on Build GRCh38
rs906804096 Chr.1: 228149969-228149969 on Build GRCh38
rs1024159577 Chr.1: 228149726-228149726 on Build GRCh38
rs377273422 Chr.1: 228149724-228149724 on Build GRCh38
rs925380385 Chr.1: 228149850-228149850 on Build GRCh38
rs907676263 Chr.1: 228149905-228149905 on Build GRCh38
rs976354008 Chr.1: 228149819-228149819 on Build GRCh38
rs947739315 Chr.1: 228149942-228149942 on Build GRCh38
rs976260865 Chr.1: 228149687-228149691 on Build GRCh38
rs1037387701 Chr.1: 228149719-228149719 on Build GRCh38
rs577539759 Chr.1: 228149829-228149829 on Build GRCh38
rs945559027 Chr.1: 228149847-228149847 on Build GRCh38
rs933238670 Chr.1: 228149884-228149884 on Build GRCh38
rs587777496 Chr.1: 228149857-228149857 on Build GRCh38
rs897703165 Chr.1: 228149731-228149731 on Build GRCh38
rs34217351 Chr.1: 228150022-228150023 on Build GRCh38
rs1007337964 Chr.1: 228150034-228150034 on Build GRCh38
rs897712864 Chr.1: 228149939-228149939 on Build GRCh38
rs543591506 Chr.1: 228149796-228149796 on Build GRCh38
rs562648643 Chr.1: 228150109-228150109 on Build GRCh38
rs945508231 Chr.1: 228149696-228149696 on Build GRCh38
rs898833320 Chr.1: 228149926-228149926 on Build GRCh38
rs571475288 Chr.1: 228149714-228149715 on Build GRCh38
rs1043870610 Chr.1: 228149709-228149709 on Build GRCh38
rs1003777940 Chr.1: 228150041-228150041 on Build GRCh38
rs796458251 Chr.1: 228149723-228149723 on Build GRCh38
rs1051596189 Chr.1: 228149690-228149690 on Build GRCh38
rs886216084 Chr.1: 228149691-228149691 on Build GRCh38
rs751739961 Chr.1: 228149666-228149666 on Build GRCh38
rs1004616384 Chr.1: 228149716-228149716 on Build GRCh38
rs1010948982 Chr.1: 228150097-228150097 on Build GRCh38
rs533784158 Chr.1: 228149885-228149885 on Build GRCh38
rs989495059 Chr.1: 228149812-228149812 on Build GRCh38
rs116655303 Chr.1: 228150075-228150075 on Build GRCh38
rs938015304 Chr.1: 228149714-228149714 on Build GRCh38
rs747668732 Chr.1: 228149943-228149943 on Build GRCh38
rs556220396 Chr.1: 228149679-228149679 on Build GRCh38
rs1037418775 Chr.1: 228149925-228149925 on Build GRCh38
rs1017752221 Chr.1: 228150044-228150044 on Build GRCh38
rs963975430 Chr.1: 228149817-228149817 on Build GRCh38
rs553545487 Chr.1: 228149940-228149940 on Build GRCh38

View Full Product Details