Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 243125933-243126480 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP170
Entrez Gene ID
9859
Gene Name
centrosomal protein 170
Gene Aliases
FAM68A, KAB, KIAA0470
UniGene
Hs.533635
Gene Chromosome Location
Chr.1: 243124428-243255936 on Build GRCh38
Transcripts
NM_001042404.1 NM_001042405.1 NM_014812.2 XM_006711843.3 XM_011544334.2 XM_011544335.2 XM_011544336.2 XM_011544337.2 XM_011544338.2 XM_011544339.2 XM_011544340.2 XM_011544341.2 XM_011544342.2 XM_011544343.2 XM_011544344.2 XM_017002932.1 XM_017002933.1 XM_017002934.1 XM_017002935.1 XM_017002936.1 XM_017002937.1 XM_017002938.1 XM_017002939.1 XM_017002940.1 XM_017002941.1 XM_017002942.1 XM_017002943.1 XM_017002944.1 XM_017002945.1 XM_017002946.1 XM_017002947.1 XM_017002948.1 XM_017002949.1 XM_017002950.1 XM_017002951.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4030223 c.4357G>A p.E1453K Chr.1: 243126475-243126475 on Build GRCh38
679639 c.4729G>A p.E1577K Chr.1: 243126475-243126475 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs764646575 Chr.1: 243126410-243126410 on Build GRCh38
rs766675783 Chr.1: 243126444-243126444 on Build GRCh38
rs922941686 Chr.1: 243126147-243126148 on Build GRCh38
rs1003130996 Chr.1: 243126345-243126345 on Build GRCh38
rs906090593 Chr.1: 243126303-243126303 on Build GRCh38
rs979742675 Chr.1: 243126118-243126118 on Build GRCh38
rs929013580 Chr.1: 243126160-243126160 on Build GRCh38
rs537268220 Chr.1: 243126175-243126175 on Build GRCh38
rs748018548 Chr.1: 243126103-243126103 on Build GRCh38
rs758519781 Chr.1: 243126435-243126435 on Build GRCh38
rs910402462 Chr.1: 243126425-243126425 on Build GRCh38
rs748761097 Chr.1: 243126155-243126155 on Build GRCh38
rs1046118 Chr.1: 243126409-243126409 on Build GRCh38
rs117873748 Chr.1: 243126388-243126388 on Build GRCh38
rs769699981 Chr.1: 243126123-243126123 on Build GRCh38
rs558602683 Chr.1: 243126428-243126428 on Build GRCh38
rs979257044 Chr.1: 243126424-243126424 on Build GRCh38
rs764945080 Chr.1: 243126137-243126138 on Build GRCh38
rs548427911 Chr.1: 243126359-243126359 on Build GRCh38
rs887940053 Chr.1: 243126205-243126206 on Build GRCh38
rs2789204 Chr.1: 243126083-243126083 on Build GRCh38
rs761926589 Chr.1: 243126415-243126415 on Build GRCh38
rs913718731 Chr.1: 243126063-243126063 on Build GRCh38
rs763920300 Chr.1: 243126328-243126328 on Build GRCh38
rs16832383 Chr.1: 243126378-243126378 on Build GRCh38
rs754426798 Chr.1: 243126414-243126414 on Build GRCh38
rs180761051 Chr.1: 243126374-243126374 on Build GRCh38
rs956070957 Chr.1: 243126038-243126038 on Build GRCh38
rs765219814 Chr.1: 243126416-243126416 on Build GRCh38
rs544336517 Chr.1: 243125967-243125967 on Build GRCh38
rs571387922 Chr.1: 243126021-243126021 on Build GRCh38
rs557165265 Chr.1: 243126187-243126187 on Build GRCh38
rs567573593 Chr.1: 243126217-243126217 on Build GRCh38
rs1002846227 Chr.1: 243126002-243126002 on Build GRCh38
rs2787271 Chr.1: 243126286-243126286 on Build GRCh38
rs747485991 Chr.1: 243126426-243126426 on Build GRCh38
rs3125240 Chr.1: 243126454-243126454 on Build GRCh38
rs562556670 Chr.1: 243126044-243126046 on Build GRCh38
rs968271827 Chr.1: 243126404-243126404 on Build GRCh38
rs752389204 Chr.1: 243126446-243126446 on Build GRCh38
rs1047506461 Chr.1: 243126194-243126194 on Build GRCh38
rs536513084 Chr.1: 243126301-243126301 on Build GRCh38
rs397834422 Chr.1: 243126274-243126274 on Build GRCh38
rs755937398 Chr.1: 243126450-243126450 on Build GRCh38
rs775685042 Chr.1: 243126402-243126402 on Build GRCh38
rs529943881 Chr.1: 243126104-243126104 on Build GRCh38
rs777713532 Chr.1: 243126134-243126134 on Build GRCh38
rs943300507 Chr.1: 243126135-243126135 on Build GRCh38
rs956100953 Chr.1: 243126370-243126370 on Build GRCh38
rs750584859 Chr.1: 243126420-243126420 on Build GRCh38
rs2787270 Chr.1: 243126333-243126333 on Build GRCh38

View Full Product Details