Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 76324317-76324814 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C10orf11
Entrez Gene ID
83938
Gene Name
chromosome 10 open reading frame 11
Gene Aliases
CDA017
UniGene
Hs.118161
Gene Chromosome Location
Chr.10: 75431646-76559220 on Build GRCh38
Transcripts
NM_001305581.1 NM_032024.4 NR_131178.1 XM_011540256.2 XM_011540257.2 XM_017016759.1 XM_017016760.1 XM_017016761.1 XM_017016762.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3786725 c.475A>G p.T159A Chr.10: 76324443-76324443 on Build GRCh38
5985773 c.477G>A p.T159T Chr.10: 76324445-76324445 on Build GRCh38
7085585 c.437C>T p.S146F Chr.10: 76324405-76324405 on Build GRCh38
1965602 c.484C>T p.P162S Chr.10: 76324452-76324452 on Build GRCh38
1965604 c.499G>A p.E167K Chr.10: 76324467-76324467 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757418537 Chr.10: 76324532-76324532 on Build GRCh38
rs563008206 Chr.10: 76324416-76324416 on Build GRCh38
rs939416184 Chr.10: 76324571-76324571 on Build GRCh38
rs750204623 Chr.10: 76324380-76324380 on Build GRCh38
rs779505497 Chr.10: 76324537-76324537 on Build GRCh38
rs149792982 Chr.10: 76324435-76324435 on Build GRCh38
rs750668962 Chr.10: 76324341-76324341 on Build GRCh38
rs964373586 Chr.10: 76324747-76324747 on Build GRCh38
rs886496519 Chr.10: 76324768-76324768 on Build GRCh38
rs879521549 Chr.10: 76324342-76324343 on Build GRCh38
rs538978980 Chr.10: 76324698-76324698 on Build GRCh38
rs535508515 Chr.10: 76324366-76324366 on Build GRCh38
rs530880397 Chr.10: 76324575-76324575 on Build GRCh38
rs372941859 Chr.10: 76324342-76324343 on Build GRCh38
rs145036984 Chr.10: 76324659-76324659 on Build GRCh38
rs771772765 Chr.10: 76324623-76324623 on Build GRCh38
rs184988607 Chr.10: 76324607-76324607 on Build GRCh38
rs372764998 Chr.10: 76324471-76324471 on Build GRCh38
rs907963520 Chr.10: 76324570-76324570 on Build GRCh38
rs758254744 Chr.10: 76324382-76324382 on Build GRCh38
rs146076285 Chr.10: 76324429-76324429 on Build GRCh38
rs370235192 Chr.10: 76324538-76324538 on Build GRCh38
rs778869213 Chr.10: 76324376-76324376 on Build GRCh38
rs149082948 Chr.10: 76324702-76324702 on Build GRCh38
rs925688405 Chr.10: 76324621-76324621 on Build GRCh38
rs905879776 Chr.10: 76324701-76324701 on Build GRCh38
rs999707539 Chr.10: 76324629-76324629 on Build GRCh38
rs1322567 Chr.10: 76324343-76324343 on Build GRCh38
rs199621795 Chr.10: 76324523-76324523 on Build GRCh38
rs35349706 Chr.10: 76324426-76324426 on Build GRCh38
rs369831117 Chr.10: 76324434-76324434 on Build GRCh38
rs749383728 Chr.10: 76324530-76324530 on Build GRCh38
rs776235954 Chr.10: 76324731-76324731 on Build GRCh38
rs1042827569 Chr.10: 76324364-76324364 on Build GRCh38
rs917394507 Chr.10: 76324633-76324633 on Build GRCh38
rs976648232 Chr.10: 76324566-76324566 on Build GRCh38
rs774713218 Chr.10: 76324461-76324462 on Build GRCh38
rs896048025 Chr.10: 76324422-76324422 on Build GRCh38
rs754858971 Chr.10: 76324440-76324440 on Build GRCh38
rs904082077 Chr.10: 76324624-76324624 on Build GRCh38
rs750361989 Chr.10: 76324492-76324492 on Build GRCh38
rs761286170 Chr.10: 76324485-76324485 on Build GRCh38
rs1040936 Chr.10: 76324649-76324649 on Build GRCh38
rs775235354 Chr.10: 76324444-76324444 on Build GRCh38
rs548448260 Chr.10: 76324609-76324609 on Build GRCh38
rs142285984 Chr.10: 76324482-76324482 on Build GRCh38
rs778852931 Chr.10: 76324603-76324603 on Build GRCh38
rs760050260 Chr.10: 76324358-76324358 on Build GRCh38
rs766183670 Chr.10: 76324503-76324503 on Build GRCh38
rs200571917 Chr.10: 76324478-76324478 on Build GRCh38
rs150134041 Chr.10: 76324719-76324719 on Build GRCh38
rs781324694 Chr.10: 76324390-76324390 on Build GRCh38
rs763696041 Chr.10: 76324476-76324476 on Build GRCh38
rs1053784189 Chr.10: 76324421-76324421 on Build GRCh38
rs769710472 Chr.10: 76324419-76324419 on Build GRCh38
rs566813169 Chr.10: 76324674-76324674 on Build GRCh38
rs765153993 Chr.10: 76324490-76324490 on Build GRCh38
rs943037232 Chr.10: 76324466-76324466 on Build GRCh38
rs1322568 Chr.10: 76324342-76324342 on Build GRCh38
rs777904422 Chr.10: 76324525-76324525 on Build GRCh38
rs1005364463 Chr.10: 76324781-76324781 on Build GRCh38
rs757188769 Chr.10: 76324367-76324367 on Build GRCh38
rs775067979 Chr.10: 76324430-76324430 on Build GRCh38
rs974667522 Chr.10: 76324469-76324469 on Build GRCh38
rs974358853 Chr.10: 76324766-76324766 on Build GRCh38
rs763682531 Chr.10: 76324360-76324360 on Build GRCh38
rs763290904 Chr.10: 76324415-76324415 on Build GRCh38
rs376408399 Chr.10: 76324507-76324507 on Build GRCh38

View Full Product Details