Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 15630236-15630734 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDI2
Entrez Gene ID
84301
Gene Name
DNA damage inducible 1 homolog 2
Gene Aliases
-
UniGene
Hs.718857
Gene Chromosome Location
Chr.1: 15617458-15661057 on Build GRCh38
Transcripts
NM_032341.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4908459 c.382T>G p.S128A Chr.1: 15630438-15630438 on Build GRCh38
4891531 c.328C>A p.R110S Chr.1: 15630384-15630384 on Build GRCh38
310502 c.493A>T p.S165C Chr.1: 15630549-15630549 on Build GRCh38
3984373 c.288C>T p.F96F Chr.1: 15630344-15630344 on Build GRCh38
1319831 c.466A>T p.N156Y Chr.1: 15630522-15630522 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148633078 Chr.1: 15630385-15630385 on Build GRCh38
rs746374554 Chr.1: 15630406-15630406 on Build GRCh38
rs528246343 Chr.1: 15630642-15630642 on Build GRCh38
rs532223617 Chr.1: 15630412-15630412 on Build GRCh38
rs765246366 Chr.1: 15630269-15630269 on Build GRCh38
rs368189349 Chr.1: 15630523-15630523 on Build GRCh38
rs777823398 Chr.1: 15630278-15630278 on Build GRCh38
rs752896869 Chr.1: 15630295-15630295 on Build GRCh38
rs140092977 Chr.1: 15630379-15630379 on Build GRCh38
rs1000212771 Chr.1: 15630478-15630478 on Build GRCh38
rs748839324 Chr.1: 15630442-15630442 on Build GRCh38
rs777052167 Chr.1: 15630477-15630477 on Build GRCh38
rs754635922 Chr.1: 15630449-15630449 on Build GRCh38
rs748799263 Chr.1: 15630338-15630338 on Build GRCh38
rs766026806 Chr.1: 15630554-15630554 on Build GRCh38
rs151141668 Chr.1: 15630464-15630464 on Build GRCh38
rs919161439 Chr.1: 15630272-15630272 on Build GRCh38
rs892597058 Chr.1: 15630658-15630658 on Build GRCh38
rs755898821 Chr.1: 15630440-15630440 on Build GRCh38
rs943607068 Chr.1: 15630450-15630450 on Build GRCh38
rs141031535 Chr.1: 15630530-15630530 on Build GRCh38
rs777375861 Chr.1: 15630603-15630603 on Build GRCh38
rs962987072 Chr.1: 15630684-15630684 on Build GRCh38
rs745657430 Chr.1: 15630314-15630314 on Build GRCh38
rs58004141 Chr.1: 15630643-15630643 on Build GRCh38
rs1002511781 Chr.1: 15630676-15630677 on Build GRCh38
rs751503696 Chr.1: 15630284-15630284 on Build GRCh38
rs773869764 Chr.1: 15630355-15630355 on Build GRCh38
rs368388796 Chr.1: 15630321-15630321 on Build GRCh38
rs1053156819 Chr.1: 15630519-15630519 on Build GRCh38
rs754242472 Chr.1: 15630550-15630550 on Build GRCh38
rs746312819 Chr.1: 15630611-15630611 on Build GRCh38
rs767454817 Chr.1: 15630540-15630540 on Build GRCh38
rs746213909 Chr.1: 15630493-15630493 on Build GRCh38
rs748062998 Chr.1: 15630507-15630508 on Build GRCh38
rs757198316 Chr.1: 15630409-15630409 on Build GRCh38
rs371977898 Chr.1: 15630372-15630372 on Build GRCh38
rs564313462 Chr.1: 15630378-15630378 on Build GRCh38
rs753671312 Chr.1: 15630564-15630564 on Build GRCh38
rs113104115 Chr.1: 15630374-15630374 on Build GRCh38
rs201289209 Chr.1: 15630299-15630299 on Build GRCh38
rs970614716 Chr.1: 15630659-15630659 on Build GRCh38
rs948383742 Chr.1: 15630626-15630626 on Build GRCh38
rs768561475 Chr.1: 15630279-15630279 on Build GRCh38
rs140041751 Chr.1: 15630467-15630467 on Build GRCh38
rs72647120 Chr.1: 15630274-15630274 on Build GRCh38
rs778593288 Chr.1: 15630586-15630586 on Build GRCh38
rs865796587 Chr.1: 15630487-15630487 on Build GRCh38
rs200999837 Chr.1: 15630494-15630494 on Build GRCh38
rs530891266 Chr.1: 15630657-15630657 on Build GRCh38
rs1033574979 Chr.1: 15630677-15630677 on Build GRCh38
rs771412314 Chr.1: 15630472-15630472 on Build GRCh38
rs972192119 Chr.1: 15630501-15630501 on Build GRCh38
rs764221560 Chr.1: 15630400-15630400 on Build GRCh38
rs757729580 Chr.1: 15630384-15630384 on Build GRCh38
rs775946271 Chr.1: 15630503-15630503 on Build GRCh38
rs757945767 Chr.1: 15630598-15630598 on Build GRCh38
rs934384137 Chr.1: 15630655-15630655 on Build GRCh38
rs768110369 Chr.1: 15630351-15630351 on Build GRCh38
rs535427500 Chr.1: 15630343-15630343 on Build GRCh38
rs779927649 Chr.1: 15630441-15630441 on Build GRCh38
rs758544170 Chr.1: 15630308-15630308 on Build GRCh38
rs1007597309 Chr.1: 15630445-15630445 on Build GRCh38
rs915868530 Chr.1: 15630545-15630545 on Build GRCh38
rs779619064 Chr.1: 15630334-15630334 on Build GRCh38
rs1022554970 Chr.1: 15630413-15630413 on Build GRCh38
rs778915334 Chr.1: 15630299-15630301 on Build GRCh38
rs750222182 Chr.1: 15630415-15630415 on Build GRCh38
rs754621645 Chr.1: 15630579-15630579 on Build GRCh38
rs750285872 Chr.1: 15630549-15630549 on Build GRCh38
rs973045773 Chr.1: 15630698-15630698 on Build GRCh38
rs1011111346 Chr.1: 15630689-15630689 on Build GRCh38
rs764466841 Chr.1: 15630376-15630376 on Build GRCh38
rs755604718 Chr.1: 15630670-15630670 on Build GRCh38
rs763211597 Chr.1: 15630520-15630520 on Build GRCh38
rs948393633 Chr.1: 15630344-15630344 on Build GRCh38
rs752179773 Chr.1: 15630589-15630589 on Build GRCh38

View Full Product Details