Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 75904299-75904749 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO22
Entrez Gene ID
26263
Gene Name
F-box protein 22
Gene Aliases
FBX22, FISTC1
UniGene
Hs.591115
Gene Chromosome Location
Chr.15: 75903859-75937513 on Build GRCh38
Transcripts
NM_012170.3 NM_147188.2 NR_037623.1 XM_011521447.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5556762 c.246T>C p.H82H Chr.15: 75904596-75904596 on Build GRCh38
434337 c.154T>C p.W52R Chr.15: 75904504-75904504 on Build GRCh38
6662115 c.156G>C p.W52C Chr.15: 75904506-75904506 on Build GRCh38
5696024 c.225G>A p.E75E Chr.15: 75904575-75904575 on Build GRCh38
5798717 c.141-3C>T p.? Chr.15: 75904488-75904488 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs953887483 Chr.15: 75904692-75904693 on Build GRCh38
rs772384272 Chr.15: 75904442-75904442 on Build GRCh38
rs780286954 Chr.15: 75904520-75904520 on Build GRCh38
rs370914225 Chr.15: 75904497-75904497 on Build GRCh38
rs533410827 Chr.15: 75904498-75904498 on Build GRCh38
rs753868039 Chr.15: 75904522-75904522 on Build GRCh38
rs962923943 Chr.15: 75904337-75904337 on Build GRCh38
rs772888237 Chr.15: 75904512-75904513 on Build GRCh38
rs949616776 Chr.15: 75904424-75904424 on Build GRCh38
rs759307254 Chr.15: 75904448-75904448 on Build GRCh38
rs377674818 Chr.15: 75904491-75904491 on Build GRCh38
rs961055366 Chr.15: 75904365-75904365 on Build GRCh38
rs144326532 Chr.15: 75904701-75904701 on Build GRCh38
rs544773236 Chr.15: 75904399-75904399 on Build GRCh38
rs755071895 Chr.15: 75904643-75904643 on Build GRCh38
rs914212376 Chr.15: 75904716-75904719 on Build GRCh38
rs2255701 Chr.15: 75904368-75904368 on Build GRCh38
rs200646126 Chr.15: 75904447-75904447 on Build GRCh38
rs955744669 Chr.15: 75904414-75904414 on Build GRCh38
rs536690717 Chr.15: 75904370-75904370 on Build GRCh38
rs773149520 Chr.15: 75904674-75904674 on Build GRCh38
rs555824722 Chr.15: 75904334-75904334 on Build GRCh38
rs769405706 Chr.15: 75904350-75904353 on Build GRCh38
rs1055179733 Chr.15: 75904394-75904394 on Build GRCh38
rs988857126 Chr.15: 75904416-75904416 on Build GRCh38
rs768121710 Chr.15: 75904577-75904577 on Build GRCh38
rs114362346 Chr.15: 75904650-75904650 on Build GRCh38
rs754124691 Chr.15: 75904641-75904641 on Build GRCh38
rs138635967 Chr.15: 75904539-75904539 on Build GRCh38
rs762529485 Chr.15: 75904643-75904644 on Build GRCh38
rs191258743 Chr.15: 75904663-75904663 on Build GRCh38
rs771856550 Chr.15: 75904673-75904673 on Build GRCh38
rs374351360 Chr.15: 75904665-75904665 on Build GRCh38
rs775096776 Chr.15: 75904455-75904455 on Build GRCh38
rs916753271 Chr.15: 75904408-75904408 on Build GRCh38
rs201567790 Chr.15: 75904541-75904541 on Build GRCh38
rs773774511 Chr.15: 75904664-75904664 on Build GRCh38
rs370973879 Chr.15: 75904658-75904658 on Build GRCh38
rs74664624 Chr.15: 75904625-75904625 on Build GRCh38
rs747317087 Chr.15: 75904651-75904651 on Build GRCh38
rs762488598 Chr.15: 75904562-75904562 on Build GRCh38
rs953876789 Chr.15: 75904558-75904558 on Build GRCh38
rs548845213 Chr.15: 75904619-75904619 on Build GRCh38
rs571576794 Chr.15: 75904692-75904692 on Build GRCh38
rs779339421 Chr.15: 75904388-75904388 on Build GRCh38
rs912189188 Chr.15: 75904358-75904358 on Build GRCh38
rs758787214 Chr.15: 75904649-75904649 on Build GRCh38
rs544612400 Chr.15: 75904533-75904533 on Build GRCh38
rs1007985566 Chr.15: 75904693-75904693 on Build GRCh38
rs377482704 Chr.15: 75904653-75904653 on Build GRCh38
rs556573247 Chr.15: 75904714-75904714 on Build GRCh38
rs1028105609 Chr.15: 75904347-75904347 on Build GRCh38
rs761709471 Chr.15: 75904591-75904591 on Build GRCh38
rs769393904 Chr.15: 75904449-75904449 on Build GRCh38
rs562866418 Chr.15: 75904400-75904400 on Build GRCh38
rs764037110 Chr.15: 75904476-75904476 on Build GRCh38
rs150157752 Chr.15: 75904537-75904537 on Build GRCh38
rs558143960 Chr.15: 75904325-75904325 on Build GRCh38
rs757231580 Chr.15: 75904338-75904338 on Build GRCh38
rs750180280 Chr.15: 75904621-75904621 on Build GRCh38
rs988522200 Chr.15: 75904708-75904708 on Build GRCh38
rs1046615737 Chr.15: 75904430-75904430 on Build GRCh38
rs753213331 Chr.15: 75904644-75904644 on Build GRCh38
rs373567025 Chr.15: 75904484-75904484 on Build GRCh38
rs2455895 Chr.15: 75904333-75904333 on Build GRCh38
rs769518649 Chr.15: 75904554-75904554 on Build GRCh38
rs762572053 Chr.15: 75904458-75904458 on Build GRCh38
rs774335782 Chr.15: 75904584-75904584 on Build GRCh38
rs1047036149 Chr.15: 75904702-75904702 on Build GRCh38
rs778035636 Chr.15: 75904551-75904551 on Build GRCh38
rs766519487 Chr.15: 75904640-75904640 on Build GRCh38
rs912277975 Chr.15: 75904396-75904396 on Build GRCh38
rs761667976 Chr.15: 75904485-75904485 on Build GRCh38
rs373192132 Chr.15: 75904560-75904560 on Build GRCh38
rs527999497 Chr.15: 75904330-75904330 on Build GRCh38
rs772459909 Chr.15: 75904542-75904542 on Build GRCh38
rs778773523 Chr.15: 75904354-75904354 on Build GRCh38
rs1019514620 Chr.15: 75904638-75904638 on Build GRCh38

View Full Product Details