Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 70379586-70380107 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ST3GAL2
Entrez Gene ID
6483
Gene Name
ST3 beta-galactoside alpha-2,3-sialyltransferase 2
Gene Aliases
Gal-NAc6S, SIAT4B, ST3GALII, ST3GalA.2
UniGene
Hs.368611
Gene Chromosome Location
Chr.16: 70379435-70439088 on Build GRCh38
Transcripts
NM_006927.3

SNP Detail

SNP ID Chromosome Location
rs556124444 Chr.16: 70380066-70380066 on Build GRCh38
rs1013108479 Chr.16: 70379963-70379963 on Build GRCh38
rs912730904 Chr.16: 70379850-70379850 on Build GRCh38
rs1141941 Chr.16: 70379786-70379786 on Build GRCh38
rs550158602 Chr.16: 70379690-70379690 on Build GRCh38
rs1051706406 Chr.16: 70379795-70379795 on Build GRCh38
rs1141939 Chr.16: 70379733-70379733 on Build GRCh38
rs1035494944 Chr.16: 70379731-70379731 on Build GRCh38
rs546546769 Chr.16: 70379738-70379738 on Build GRCh38
rs918090407 Chr.16: 70379979-70379979 on Build GRCh38
rs1064758 Chr.16: 70379652-70379652 on Build GRCh38
rs71387521 Chr.16: 70379796-70379797 on Build GRCh38
rs893119929 Chr.16: 70379694-70379694 on Build GRCh38
rs552885801 Chr.16: 70379989-70379989 on Build GRCh38
rs537369209 Chr.16: 70379664-70379664 on Build GRCh38
rs907724132 Chr.16: 70379685-70379685 on Build GRCh38
rs912753138 Chr.16: 70380055-70380055 on Build GRCh38
rs532979074 Chr.16: 70379797-70379798 on Build GRCh38
rs112967319 Chr.16: 70379852-70379852 on Build GRCh38
rs781757164 Chr.16: 70380005-70380005 on Build GRCh38
rs935434648 Chr.16: 70379737-70379737 on Build GRCh38
rs887474097 Chr.16: 70380036-70380036 on Build GRCh38
rs1141940 Chr.16: 70379751-70379751 on Build GRCh38
rs545490054 Chr.16: 70379909-70379909 on Build GRCh38
rs901600006 Chr.16: 70380080-70380080 on Build GRCh38
rs1038569709 Chr.16: 70379609-70379609 on Build GRCh38
rs577030295 Chr.16: 70379885-70379885 on Build GRCh38
rs556994350 Chr.16: 70379755-70379755 on Build GRCh38
rs796138092 Chr.16: 70379971-70379974 on Build GRCh38
rs921924031 Chr.16: 70379722-70379722 on Build GRCh38
rs117171674 Chr.16: 70380056-70380056 on Build GRCh38
rs1036035556 Chr.16: 70379796-70379796 on Build GRCh38
rs991250857 Chr.16: 70379749-70379749 on Build GRCh38
rs770553537 Chr.16: 70380063-70380063 on Build GRCh38
rs11438171 Chr.16: 70379811-70379812 on Build GRCh38
rs568243308 Chr.16: 70379632-70379633 on Build GRCh38
rs1011530242 Chr.16: 70379707-70379707 on Build GRCh38
rs568006849 Chr.16: 70380070-70380070 on Build GRCh38
rs935415961 Chr.16: 70379974-70379974 on Build GRCh38
rs979406413 Chr.16: 70379961-70379962 on Build GRCh38
rs374948658 Chr.16: 70379980-70379989 on Build GRCh38
rs951673220 Chr.16: 70380011-70380011 on Build GRCh38
rs958734481 Chr.16: 70379734-70379734 on Build GRCh38
rs527510671 Chr.16: 70379947-70379947 on Build GRCh38
rs930186765 Chr.16: 70379789-70379789 on Build GRCh38
rs905456546 Chr.16: 70379797-70379802 on Build GRCh38
rs971705741 Chr.16: 70379972-70379972 on Build GRCh38
rs548228199 Chr.16: 70379907-70379907 on Build GRCh38
rs780238776 Chr.16: 70379723-70379723 on Build GRCh38
rs1027372271 Chr.16: 70379647-70379647 on Build GRCh38
rs971674884 Chr.16: 70379779-70379779 on Build GRCh38
rs908893107 Chr.16: 70379772-70379772 on Build GRCh38
rs150459098 Chr.16: 70379935-70379935 on Build GRCh38
rs1016027467 Chr.16: 70379968-70379968 on Build GRCh38
rs930155811 Chr.16: 70379632-70379632 on Build GRCh38
rs1014749927 Chr.16: 70379923-70379923 on Build GRCh38
rs893182950 Chr.16: 70379899-70379899 on Build GRCh38
rs529285597 Chr.16: 70379886-70379886 on Build GRCh38
rs146627322 Chr.16: 70380026-70380026 on Build GRCh38
rs768947562 Chr.16: 70379970-70379970 on Build GRCh38
rs376895920 Chr.16: 70379727-70379727 on Build GRCh38
rs1027751340 Chr.16: 70379879-70379879 on Build GRCh38
rs1045221538 Chr.16: 70380007-70380007 on Build GRCh38
rs1045329629 Chr.16: 70379797-70379798 on Build GRCh38
rs947420124 Chr.16: 70380076-70380076 on Build GRCh38
rs905427233 Chr.16: 70380008-70380008 on Build GRCh38
rs1141938 Chr.16: 70379688-70379688 on Build GRCh38
rs1041701102 Chr.16: 70379928-70379928 on Build GRCh38
rs537047464 Chr.16: 70380061-70380061 on Build GRCh38
rs918856142 Chr.16: 70379784-70379784 on Build GRCh38
rs891205580 Chr.16: 70379660-70379660 on Build GRCh38
rs1052983011 Chr.16: 70379976-70379979 on Build GRCh38
rs918782336 Chr.16: 70379617-70379617 on Build GRCh38
rs963000154 Chr.16: 70379774-70379774 on Build GRCh38
rs796266306 Chr.16: 70379943-70379943 on Build GRCh38
rs1002784021 Chr.16: 70379938-70379938 on Build GRCh38
rs79872848 Chr.16: 70379809-70379811 on Build GRCh38
rs551923074 Chr.16: 70379955-70379955 on Build GRCh38
rs538413806 Chr.16: 70380077-70380077 on Build GRCh38
rs1020226598 Chr.16: 70379716-70379716 on Build GRCh38
rs1001127554 Chr.16: 70379855-70379857 on Build GRCh38
rs983281314 Chr.16: 70379683-70379683 on Build GRCh38
rs1051180426 Chr.16: 70379639-70379639 on Build GRCh38

View Full Product Details