Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 35133070-35133562 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NLE1
Entrez Gene ID
54475
Gene Name
notchless homolog 1
Gene Aliases
Nle
UniGene
Hs.85570
Gene Chromosome Location
Chr.17: 35128755-35142315 on Build GRCh38
Transcripts
NM_001014445.2 NM_018096.4 XM_017024777.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4463275 c.1284C>T p.L428L Chr.17: 35133429-35133429 on Build GRCh38
5785696 c.1215-1G>A p.? Chr.17: 35133499-35133499 on Build GRCh38
5784814 c.1216T>C p.Y406H Chr.17: 35133497-35133497 on Build GRCh38
5852234 c.1382C>T p.A461V Chr.17: 35133234-35133234 on Build GRCh38
6846103 c.1369G>A p.D457N Chr.17: 35133344-35133344 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs867160344 Chr.17: 35133307-35133307 on Build GRCh38
rs1034211576 Chr.17: 35133514-35133514 on Build GRCh38
rs201814140 Chr.17: 35133328-35133328 on Build GRCh38
rs202004406 Chr.17: 35133150-35133150 on Build GRCh38
rs772719649 Chr.17: 35133521-35133521 on Build GRCh38
rs748945159 Chr.17: 35133473-35133473 on Build GRCh38
rs546214882 Chr.17: 35133170-35133170 on Build GRCh38
rs200586939 Chr.17: 35133138-35133138 on Build GRCh38
rs763414532 Chr.17: 35133327-35133327 on Build GRCh38
rs770972811 Chr.17: 35133132-35133132 on Build GRCh38
rs2306510 Chr.17: 35133156-35133156 on Build GRCh38
rs2306511 Chr.17: 35133348-35133348 on Build GRCh38
rs116857354 Chr.17: 35133194-35133194 on Build GRCh38
rs745751342 Chr.17: 35133456-35133456 on Build GRCh38
rs780135492 Chr.17: 35133275-35133275 on Build GRCh38
rs774141962 Chr.17: 35133142-35133142 on Build GRCh38
rs780791903 Chr.17: 35133442-35133442 on Build GRCh38
rs996410128 Chr.17: 35133106-35133106 on Build GRCh38
rs754100854 Chr.17: 35133434-35133434 on Build GRCh38
rs968524755 Chr.17: 35133179-35133179 on Build GRCh38
rs757096040 Chr.17: 35133451-35133451 on Build GRCh38
rs199746985 Chr.17: 35133278-35133278 on Build GRCh38
rs2306512 Chr.17: 35133412-35133412 on Build GRCh38
rs773803714 Chr.17: 35133481-35133481 on Build GRCh38
rs898736397 Chr.17: 35133183-35133183 on Build GRCh38
rs1028276023 Chr.17: 35133215-35133215 on Build GRCh38
rs554861899 Chr.17: 35133464-35133464 on Build GRCh38
rs762162494 Chr.17: 35133410-35133410 on Build GRCh38
rs775238574 Chr.17: 35133387-35133387 on Build GRCh38
rs2306513 Chr.17: 35133497-35133497 on Build GRCh38
rs148833526 Chr.17: 35133465-35133465 on Build GRCh38
rs759671268 Chr.17: 35133519-35133519 on Build GRCh38
rs369166012 Chr.17: 35133420-35133420 on Build GRCh38
rs1010761352 Chr.17: 35133518-35133518 on Build GRCh38
rs779269491 Chr.17: 35133448-35133448 on Build GRCh38
rs765521287 Chr.17: 35133173-35133173 on Build GRCh38
rs776211486 Chr.17: 35133311-35133311 on Build GRCh38
rs201248090 Chr.17: 35133171-35133171 on Build GRCh38
rs77153156 Chr.17: 35133124-35133124 on Build GRCh38
rs374843799 Chr.17: 35133339-35133339 on Build GRCh38
rs757704824 Chr.17: 35133345-35133345 on Build GRCh38
rs761041022 Chr.17: 35133487-35133487 on Build GRCh38
rs200490692 Chr.17: 35133222-35133222 on Build GRCh38
rs144117401 Chr.17: 35133377-35133377 on Build GRCh38
rs149698227 Chr.17: 35133419-35133419 on Build GRCh38
rs147757651 Chr.17: 35133480-35133480 on Build GRCh38
rs866559995 Chr.17: 35133469-35133469 on Build GRCh38
rs188481982 Chr.17: 35133178-35133178 on Build GRCh38
rs367759894 Chr.17: 35133133-35133133 on Build GRCh38
rs781229102 Chr.17: 35133455-35133455 on Build GRCh38
rs758302050 Chr.17: 35133357-35133357 on Build GRCh38
rs927216732 Chr.17: 35133268-35133268 on Build GRCh38
rs773979981 Chr.17: 35133477-35133477 on Build GRCh38
rs745431652 Chr.17: 35133382-35133382 on Build GRCh38
rs146002563 Chr.17: 35133393-35133393 on Build GRCh38
rs756293836 Chr.17: 35133257-35133257 on Build GRCh38
rs750583113 Chr.17: 35133347-35133347 on Build GRCh38
rs113578546 Chr.17: 35133162-35133162 on Build GRCh38
rs760697931 Chr.17: 35133306-35133307 on Build GRCh38
rs771055880 Chr.17: 35133364-35133364 on Build GRCh38
rs760851654 Chr.17: 35133424-35133424 on Build GRCh38
rs956336645 Chr.17: 35133318-35133318 on Build GRCh38
rs758676653 Chr.17: 35133175-35133175 on Build GRCh38
rs369573413 Chr.17: 35133474-35133474 on Build GRCh38
rs761664528 Chr.17: 35133145-35133145 on Build GRCh38
rs755181789 Chr.17: 35133439-35133439 on Build GRCh38
rs764070000 Chr.17: 35133420-35133441 on Build GRCh38
rs142500144 Chr.17: 35133479-35133479 on Build GRCh38
rs780376203 Chr.17: 35133353-35133353 on Build GRCh38
rs570801349 Chr.17: 35133320-35133320 on Build GRCh38
rs750904440 Chr.17: 35133386-35133386 on Build GRCh38
rs370055501 Chr.17: 35133332-35133332 on Build GRCh38
rs2306509 Chr.17: 35133141-35133141 on Build GRCh38
rs773324681 Chr.17: 35133282-35133282 on Build GRCh38
rs145494138 Chr.17: 35133450-35133450 on Build GRCh38
rs770575745 Chr.17: 35133298-35133298 on Build GRCh38
rs767590380 Chr.17: 35133346-35133346 on Build GRCh38
rs753930277 Chr.17: 35133516-35133516 on Build GRCh38
rs758960397 Chr.17: 35133313-35133313 on Build GRCh38
rs889815267 Chr.17: 35133266-35133266 on Build GRCh38
rs765958002 Chr.17: 35133165-35133165 on Build GRCh38
rs757356761 Chr.17: 35133197-35133197 on Build GRCh38
rs777651397 Chr.17: 35133362-35133362 on Build GRCh38
rs989558383 Chr.17: 35133168-35133168 on Build GRCh38
rs774983468 Chr.17: 35133325-35133325 on Build GRCh38
rs139404544 Chr.17: 35133354-35133354 on Build GRCh38
rs781201165 Chr.17: 35133203-35133203 on Build GRCh38
rs547620077 Chr.17: 35133297-35133297 on Build GRCh38
rs757377728 Chr.17: 35133176-35133176 on Build GRCh38
rs746944922 Chr.17: 35133363-35133363 on Build GRCh38
rs753565362 Chr.17: 35133166-35133166 on Build GRCh38
rs766331515 Chr.17: 35133433-35133433 on Build GRCh38
rs753168490 Chr.17: 35133174-35133174 on Build GRCh38
rs200716610 Chr.17: 35133402-35133402 on Build GRCh38
rs748037158 Chr.17: 35133276-35133276 on Build GRCh38

View Full Product Details