Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 35137100-35137589 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NLE1
Entrez Gene ID
54475
Gene Name
notchless homolog 1
Gene Aliases
Nle
UniGene
Hs.85570
Gene Chromosome Location
Chr.17: 35128755-35142315 on Build GRCh38
Transcripts
NM_001014445.2 NM_018096.4 XM_017024777.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5772608 c.671A>G p.K224R Chr.17: 35137158-35137158 on Build GRCh38
1382415 c.729C>T p.T243T Chr.17: 35137100-35137100 on Build GRCh38
4413501 c.710G>T p.R237L Chr.17: 35137119-35137119 on Build GRCh38
3181279 c.709C>T p.R237C Chr.17: 35137120-35137120 on Build GRCh38
1382416 c.718C>T p.R240C Chr.17: 35137111-35137111 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747370368 Chr.17: 35137496-35137496 on Build GRCh38
rs748945349 Chr.17: 35137124-35137124 on Build GRCh38
rs368961163 Chr.17: 35137143-35137143 on Build GRCh38
rs781772231 Chr.17: 35137567-35137567 on Build GRCh38
rs766355409 Chr.17: 35137540-35137540 on Build GRCh38
rs758133303 Chr.17: 35137551-35137551 on Build GRCh38
rs574114783 Chr.17: 35137553-35137553 on Build GRCh38
rs941246995 Chr.17: 35137488-35137488 on Build GRCh38
rs10153277 Chr.17: 35137235-35137235 on Build GRCh38
rs554232539 Chr.17: 35137526-35137526 on Build GRCh38
rs1032296569 Chr.17: 35137386-35137386 on Build GRCh38
rs141917940 Chr.17: 35137181-35137181 on Build GRCh38
rs144949018 Chr.17: 35137543-35137543 on Build GRCh38
rs780109376 Chr.17: 35137473-35137473 on Build GRCh38
rs533571855 Chr.17: 35137563-35137563 on Build GRCh38
rs147760459 Chr.17: 35137473-35137473 on Build GRCh38
rs78469973 Chr.17: 35137216-35137216 on Build GRCh38
rs924199282 Chr.17: 35137305-35137305 on Build GRCh38
rs184366452 Chr.17: 35137515-35137515 on Build GRCh38
rs769876469 Chr.17: 35137517-35137517 on Build GRCh38
rs750341201 Chr.17: 35137213-35137213 on Build GRCh38
rs995979871 Chr.17: 35137532-35137532 on Build GRCh38
rs1029186079 Chr.17: 35137255-35137255 on Build GRCh38
rs1045854791 Chr.17: 35137159-35137159 on Build GRCh38
rs774482384 Chr.17: 35137536-35137537 on Build GRCh38
rs938520441 Chr.17: 35137475-35137475 on Build GRCh38
rs987757854 Chr.17: 35137287-35137287 on Build GRCh38
rs375266862 Chr.17: 35137151-35137151 on Build GRCh38
rs928683976 Chr.17: 35137426-35137426 on Build GRCh38
rs372211951 Chr.17: 35137393-35137393 on Build GRCh38
rs771622078 Chr.17: 35137500-35137500 on Build GRCh38
rs539954745 Chr.17: 35137120-35137120 on Build GRCh38
rs377605042 Chr.17: 35137241-35137241 on Build GRCh38
rs376313441 Chr.17: 35137528-35137528 on Build GRCh38
rs370419699 Chr.17: 35137229-35137229 on Build GRCh38
rs550657514 Chr.17: 35137257-35137257 on Build GRCh38
rs755644419 Chr.17: 35137119-35137119 on Build GRCh38
rs569399812 Chr.17: 35137477-35137477 on Build GRCh38
rs758032173 Chr.17: 35137351-35137351 on Build GRCh38
rs373263269 Chr.17: 35137419-35137419 on Build GRCh38
rs530944709 Chr.17: 35137197-35137197 on Build GRCh38
rs915285297 Chr.17: 35137410-35137410 on Build GRCh38
rs1056937150 Chr.17: 35137498-35137498 on Build GRCh38
rs764758756 Chr.17: 35137548-35137548 on Build GRCh38
rs771008788 Chr.17: 35137196-35137200 on Build GRCh38
rs752359094 Chr.17: 35137238-35137238 on Build GRCh38
rs970067118 Chr.17: 35137402-35137402 on Build GRCh38
rs776915676 Chr.17: 35137179-35137179 on Build GRCh38
rs748512588 Chr.17: 35137144-35137144 on Build GRCh38
rs955126822 Chr.17: 35137268-35137268 on Build GRCh38
rs1009074576 Chr.17: 35137561-35137561 on Build GRCh38
rs539223670 Chr.17: 35137430-35137430 on Build GRCh38
rs761543719 Chr.17: 35137196-35137196 on Build GRCh38
rs530185110 Chr.17: 35137317-35137317 on Build GRCh38
rs1044615654 Chr.17: 35137459-35137459 on Build GRCh38
rs760451454 Chr.17: 35137530-35137530 on Build GRCh38
rs996455964 Chr.17: 35137550-35137550 on Build GRCh38
rs948183496 Chr.17: 35137458-35137458 on Build GRCh38
rs989501956 Chr.17: 35137394-35137394 on Build GRCh38
rs771768674 Chr.17: 35137527-35137527 on Build GRCh38
rs775597300 Chr.17: 35137523-35137523 on Build GRCh38
rs752292980 Chr.17: 35137549-35137549 on Build GRCh38
rs75185184 Chr.17: 35137242-35137242 on Build GRCh38
rs975302820 Chr.17: 35137254-35137254 on Build GRCh38
rs781758551 Chr.17: 35137503-35137503 on Build GRCh38
rs567003591 Chr.17: 35137399-35137399 on Build GRCh38
rs771135738 Chr.17: 35137177-35137177 on Build GRCh38
rs1029380860 Chr.17: 35137249-35137249 on Build GRCh38
rs147564941 Chr.17: 35137180-35137180 on Build GRCh38
rs905754383 Chr.17: 35137139-35137139 on Build GRCh38
rs773627447 Chr.17: 35137152-35137152 on Build GRCh38
rs370612629 Chr.17: 35137258-35137258 on Build GRCh38
rs367653256 Chr.17: 35137127-35137127 on Build GRCh38
rs147948223 Chr.17: 35137566-35137566 on Build GRCh38
rs561346717 Chr.17: 35137315-35137315 on Build GRCh38
rs764357289 Chr.17: 35137447-35137447 on Build GRCh38
rs759003078 Chr.17: 35137544-35137544 on Build GRCh38
rs747457974 Chr.17: 35137154-35137154 on Build GRCh38
rs552781748 Chr.17: 35137472-35137472 on Build GRCh38

View Full Product Details