Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 38361631-38362099 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SOCS7
Entrez Gene ID
30837
Gene Name
suppressor of cytokine signaling 7
Gene Aliases
NAP4, NCKAP4
UniGene
Hs.514132
Gene Chromosome Location
Chr.17: 38351876-38405593 on Build GRCh38
Transcripts
NM_014598.3 XM_017024551.1 XM_017024552.1

SNP Detail

SNP ID Chromosome Location
rs775357497 Chr.17: 38361786-38361786 on Build GRCh38
rs751652753 Chr.17: 38361659-38361659 on Build GRCh38
rs4795269 Chr.17: 38361946-38361946 on Build GRCh38
rs370641464 Chr.17: 38361820-38361820 on Build GRCh38
rs587767206 Chr.17: 38361819-38361819 on Build GRCh38
rs1027946130 Chr.17: 38362076-38362076 on Build GRCh38
rs779085383 Chr.17: 38361749-38361749 on Build GRCh38
rs762677439 Chr.17: 38361660-38361660 on Build GRCh38
rs780311034 Chr.17: 38361704-38361704 on Build GRCh38
rs763524178 Chr.17: 38361669-38361669 on Build GRCh38
rs112843196 Chr.17: 38361708-38361708 on Build GRCh38
rs191830301 Chr.17: 38361681-38361681 on Build GRCh38
rs950149505 Chr.17: 38361849-38361849 on Build GRCh38
rs983418202 Chr.17: 38361847-38361847 on Build GRCh38
rs76549267 Chr.17: 38361731-38361731 on Build GRCh38
rs1028770540 Chr.17: 38361875-38361875 on Build GRCh38
rs996032279 Chr.17: 38362047-38362047 on Build GRCh38
rs961815130 Chr.17: 38361943-38361943 on Build GRCh38
rs139493404 Chr.17: 38361748-38361748 on Build GRCh38
rs751103727 Chr.17: 38361670-38361670 on Build GRCh38
rs1043247217 Chr.17: 38361797-38361797 on Build GRCh38
rs748287819 Chr.17: 38361762-38361762 on Build GRCh38
rs1055152851 Chr.17: 38361907-38361907 on Build GRCh38
rs975089551 Chr.17: 38361807-38361807 on Build GRCh38
rs750755724 Chr.17: 38361686-38361686 on Build GRCh38
rs587677016 Chr.17: 38361680-38361680 on Build GRCh38
rs1020684484 Chr.17: 38361683-38361683 on Build GRCh38
rs769415677 Chr.17: 38361782-38361782 on Build GRCh38
rs773833001 Chr.17: 38361816-38361816 on Build GRCh38
rs753951167 Chr.17: 38361723-38361723 on Build GRCh38
rs902237908 Chr.17: 38362004-38362004 on Build GRCh38
rs780198546 Chr.17: 38361658-38361658 on Build GRCh38
rs140087603 Chr.17: 38361961-38361961 on Build GRCh38
rs41470848 Chr.17: 38361999-38361999 on Build GRCh38
rs938659377 Chr.17: 38361672-38361672 on Build GRCh38
rs1055899035 Chr.17: 38361687-38361687 on Build GRCh38
rs745658058 Chr.17: 38361778-38361778 on Build GRCh38
rs762476001 Chr.17: 38361793-38361793 on Build GRCh38
rs376589426 Chr.17: 38361703-38361703 on Build GRCh38
rs75312910 Chr.17: 38361851-38361851 on Build GRCh38
rs781341947 Chr.17: 38361766-38361766 on Build GRCh38
rs893402581 Chr.17: 38361926-38361926 on Build GRCh38
rs947446583 Chr.17: 38362006-38362006 on Build GRCh38
rs41439448 Chr.17: 38361891-38361891 on Build GRCh38
rs368071528 Chr.17: 38361813-38361813 on Build GRCh38
rs900372979 Chr.17: 38362015-38362015 on Build GRCh38
rs587773460 Chr.17: 38361666-38361666 on Build GRCh38
rs1018762632 Chr.17: 38362013-38362013 on Build GRCh38
rs755086156 Chr.17: 38361821-38361821 on Build GRCh38
rs368047299 Chr.17: 38361750-38361750 on Build GRCh38
rs587720972 Chr.17: 38361890-38361890 on Build GRCh38
rs770957993 Chr.17: 38361848-38361848 on Build GRCh38
rs749141561 Chr.17: 38361811-38361811 on Build GRCh38
rs1009093392 Chr.17: 38362007-38362007 on Build GRCh38
rs999114356 Chr.17: 38361830-38361830 on Build GRCh38
rs748839943 Chr.17: 38361815-38361816 on Build GRCh38
rs75445121 Chr.17: 38361730-38361730 on Build GRCh38
rs868357356 Chr.17: 38361724-38361724 on Build GRCh38
rs770960071 Chr.17: 38361763-38361763 on Build GRCh38
rs937602869 Chr.17: 38361895-38361895 on Build GRCh38

View Full Product Details