Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 38365221-38365721 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SOCS7
Entrez Gene ID
30837
Gene Name
suppressor of cytokine signaling 7
Gene Aliases
NAP4, NCKAP4
UniGene
Hs.514132
Gene Chromosome Location
Chr.17: 38351876-38405593 on Build GRCh38
Transcripts
NM_014598.3 XM_017024551.1 XM_017024552.1

SNP Detail

SNP ID Chromosome Location
rs764428404 Chr.17: 38365299-38365299 on Build GRCh38
rs1051980558 Chr.17: 38365495-38365495 on Build GRCh38
rs763453184 Chr.17: 38365418-38365418 on Build GRCh38
rs769131073 Chr.17: 38365420-38365420 on Build GRCh38
rs754916142 Chr.17: 38365344-38365344 on Build GRCh38
rs777548712 Chr.17: 38365357-38365357 on Build GRCh38
rs769962522 Chr.17: 38365399-38365399 on Build GRCh38
rs34825831 Chr.17: 38365623-38365624 on Build GRCh38
rs746131710 Chr.17: 38365691-38365691 on Build GRCh38
rs912944752 Chr.17: 38365662-38365662 on Build GRCh38
rs587621728 Chr.17: 38365665-38365665 on Build GRCh38
rs891547163 Chr.17: 38365339-38365339 on Build GRCh38
rs142786789 Chr.17: 38365375-38365375 on Build GRCh38
rs916776483 Chr.17: 38365505-38365505 on Build GRCh38
rs56345414 Chr.17: 38365610-38365610 on Build GRCh38
rs587679464 Chr.17: 38365303-38365303 on Build GRCh38
rs751564657 Chr.17: 38365323-38365323 on Build GRCh38
rs754037792 Chr.17: 38365265-38365265 on Build GRCh38
rs762023559 Chr.17: 38365324-38365324 on Build GRCh38
rs752535505 Chr.17: 38365350-38365350 on Build GRCh38
rs989544493 Chr.17: 38365692-38365692 on Build GRCh38
rs1024865419 Chr.17: 38365605-38365605 on Build GRCh38
rs375787710 Chr.17: 38365424-38365424 on Build GRCh38
rs144493134 Chr.17: 38365312-38365312 on Build GRCh38
rs35470022 Chr.17: 38365639-38365640 on Build GRCh38
rs376607083 Chr.17: 38365636-38365636 on Build GRCh38
rs757432053 Chr.17: 38365381-38365381 on Build GRCh38
rs776025243 Chr.17: 38365279-38365279 on Build GRCh38
rs145376452 Chr.17: 38365386-38365386 on Build GRCh38
rs951208325 Chr.17: 38365598-38365598 on Build GRCh38
rs763215543 Chr.17: 38365294-38365294 on Build GRCh38
rs774696887 Chr.17: 38365426-38365426 on Build GRCh38
rs750535304 Chr.17: 38365433-38365433 on Build GRCh38
rs980751883 Chr.17: 38365622-38365622 on Build GRCh38
rs780181262 Chr.17: 38365589-38365589 on Build GRCh38
rs778640611 Chr.17: 38365347-38365347 on Build GRCh38
rs765197104 Chr.17: 38365442-38365442 on Build GRCh38
rs193161497 Chr.17: 38365480-38365480 on Build GRCh38
rs371505316 Chr.17: 38365390-38365390 on Build GRCh38
rs933785244 Chr.17: 38365679-38365679 on Build GRCh38
rs757955135 Chr.17: 38365351-38365351 on Build GRCh38
rs1052805913 Chr.17: 38365643-38365643 on Build GRCh38
rs587697165 Chr.17: 38365668-38365668 on Build GRCh38
rs746488553 Chr.17: 38365368-38365368 on Build GRCh38
rs899573221 Chr.17: 38365568-38365568 on Build GRCh38
rs759423035 Chr.17: 38365439-38365439 on Build GRCh38
rs587716644 Chr.17: 38365253-38365253 on Build GRCh38
rs945625242 Chr.17: 38365490-38365490 on Build GRCh38
rs878967809 Chr.17: 38365380-38365380 on Build GRCh38
rs936849924 Chr.17: 38365435-38365435 on Build GRCh38
rs587607111 Chr.17: 38365528-38365528 on Build GRCh38
rs752484188 Chr.17: 38365450-38365450 on Build GRCh38
rs587664942 Chr.17: 38365594-38365594 on Build GRCh38
rs761954805 Chr.17: 38365427-38365427 on Build GRCh38
rs750293452 Chr.17: 38365456-38365456 on Build GRCh38
rs149190855 Chr.17: 38365387-38365387 on Build GRCh38
rs935731750 Chr.17: 38365283-38365283 on Build GRCh38
rs1039456835 Chr.17: 38365567-38365567 on Build GRCh38
rs925536650 Chr.17: 38365596-38365596 on Build GRCh38
rs995333954 Chr.17: 38365572-38365572 on Build GRCh38
rs774103071 Chr.17: 38365357-38365358 on Build GRCh38
rs753718768 Chr.17: 38365333-38365333 on Build GRCh38
rs766214695 Chr.17: 38365328-38365328 on Build GRCh38
rs767568846 Chr.17: 38365432-38365432 on Build GRCh38
rs758265244 Chr.17: 38365544-38365544 on Build GRCh38
rs960833089 Chr.17: 38365464-38365464 on Build GRCh38
rs141734345 Chr.17: 38365604-38365604 on Build GRCh38
rs117759581 Chr.17: 38365254-38365254 on Build GRCh38
rs745480490 Chr.17: 38365416-38365420 on Build GRCh38
rs970749171 Chr.17: 38365621-38365621 on Build GRCh38

View Full Product Details