Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 38366001-38366499 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SOCS7
Entrez Gene ID
30837
Gene Name
suppressor of cytokine signaling 7
Gene Aliases
NAP4, NCKAP4
UniGene
Hs.514132
Gene Chromosome Location
Chr.17: 38351876-38405593 on Build GRCh38
Transcripts
NM_014598.3 XM_017024551.1 XM_017024552.1

SNP Detail

SNP ID Chromosome Location
rs770931713 Chr.17: 38366384-38366384 on Build GRCh38
rs779728689 Chr.17: 38366370-38366370 on Build GRCh38
rs773479428 Chr.17: 38366380-38366380 on Build GRCh38
rs774584907 Chr.17: 38366296-38366296 on Build GRCh38
rs867418668 Chr.17: 38366308-38366308 on Build GRCh38
rs776845960 Chr.17: 38366256-38366258 on Build GRCh38
rs762800266 Chr.17: 38366327-38366327 on Build GRCh38
rs761582484 Chr.17: 38366442-38366442 on Build GRCh38
rs372664557 Chr.17: 38366374-38366374 on Build GRCh38
rs745417727 Chr.17: 38366269-38366269 on Build GRCh38
rs369677064 Chr.17: 38366326-38366326 on Build GRCh38
rs201354583 Chr.17: 38366299-38366299 on Build GRCh38
rs587726426 Chr.17: 38366126-38366126 on Build GRCh38
rs763906134 Chr.17: 38366330-38366330 on Build GRCh38
rs905298612 Chr.17: 38366164-38366164 on Build GRCh38
rs955027083 Chr.17: 38366225-38366225 on Build GRCh38
rs765145698 Chr.17: 38366316-38366316 on Build GRCh38
rs370033423 Chr.17: 38366371-38366371 on Build GRCh38
rs930967328 Chr.17: 38366230-38366230 on Build GRCh38
rs749951792 Chr.17: 38366350-38366350 on Build GRCh38
rs748512376 Chr.17: 38366376-38366376 on Build GRCh38
rs199820589 Chr.17: 38366298-38366298 on Build GRCh38
rs943632418 Chr.17: 38366132-38366132 on Build GRCh38
rs1047501101 Chr.17: 38366096-38366096 on Build GRCh38
rs370524031 Chr.17: 38366462-38366462 on Build GRCh38
rs774162486 Chr.17: 38366422-38366422 on Build GRCh38
rs770396005 Chr.17: 38366275-38366275 on Build GRCh38
rs910942690 Chr.17: 38366242-38366242 on Build GRCh38
rs760809198 Chr.17: 38366305-38366305 on Build GRCh38
rs893823473 Chr.17: 38366433-38366433 on Build GRCh38
rs587638606 Chr.17: 38366112-38366112 on Build GRCh38
rs751329713 Chr.17: 38366331-38366331 on Build GRCh38
rs202212370 Chr.17: 38366260-38366260 on Build GRCh38
rs200305110 Chr.17: 38366354-38366354 on Build GRCh38
rs587759910 Chr.17: 38366127-38366127 on Build GRCh38
rs753313415 Chr.17: 38366458-38366458 on Build GRCh38
rs587682412 Chr.17: 38366421-38366421 on Build GRCh38
rs202135022 Chr.17: 38366445-38366445 on Build GRCh38
rs988451605 Chr.17: 38366202-38366202 on Build GRCh38
rs776719849 Chr.17: 38366398-38366398 on Build GRCh38
rs749606877 Chr.17: 38366287-38366287 on Build GRCh38
rs957160616 Chr.17: 38366191-38366191 on Build GRCh38
rs185027645 Chr.17: 38366181-38366181 on Build GRCh38
rs587709547 Chr.17: 38366218-38366218 on Build GRCh38
rs373112910 Chr.17: 38366344-38366344 on Build GRCh38
rs929932181 Chr.17: 38366072-38366072 on Build GRCh38
rs765951629 Chr.17: 38366451-38366451 on Build GRCh38
rs769010821 Chr.17: 38366295-38366295 on Build GRCh38
rs751153119 Chr.17: 38366247-38366247 on Build GRCh38
rs896521475 Chr.17: 38366098-38366098 on Build GRCh38
rs773243550 Chr.17: 38366300-38366300 on Build GRCh38
rs189924642 Chr.17: 38366236-38366236 on Build GRCh38
rs772275000 Chr.17: 38366378-38366378 on Build GRCh38
rs762896332 Chr.17: 38366409-38366409 on Build GRCh38
rs747244252 Chr.17: 38366381-38366381 on Build GRCh38
rs780776053 Chr.17: 38366283-38366283 on Build GRCh38
rs767266190 Chr.17: 38366443-38366443 on Build GRCh38
rs769431831 Chr.17: 38366081-38366081 on Build GRCh38
rs1001006268 Chr.17: 38366172-38366172 on Build GRCh38
rs762676494 Chr.17: 38366178-38366178 on Build GRCh38
rs1023689250 Chr.17: 38366135-38366135 on Build GRCh38
rs745733821 Chr.17: 38366257-38366257 on Build GRCh38
rs747763614 Chr.17: 38366064-38366064 on Build GRCh38
rs767321219 Chr.17: 38366345-38366345 on Build GRCh38
rs912184708 Chr.17: 38366101-38366101 on Build GRCh38
rs766616981 Chr.17: 38366113-38366113 on Build GRCh38

View Full Product Details