Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 38367801-38368311 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SOCS7
Entrez Gene ID
30837
Gene Name
suppressor of cytokine signaling 7
Gene Aliases
NAP4, NCKAP4
UniGene
Hs.514132
Gene Chromosome Location
Chr.17: 38351876-38405593 on Build GRCh38
Transcripts
NM_014598.3 XM_017024551.1 XM_017024552.1

SNP Detail

SNP ID Chromosome Location
rs1018924761 Chr.17: 38368013-38368013 on Build GRCh38
rs772162015 Chr.17: 38368098-38368098 on Build GRCh38
rs763231234 Chr.17: 38368100-38368100 on Build GRCh38
rs373718693 Chr.17: 38367872-38367872 on Build GRCh38
rs1054840869 Chr.17: 38368205-38368205 on Build GRCh38
rs112592220 Chr.17: 38368195-38368195 on Build GRCh38
rs950351606 Chr.17: 38368171-38368171 on Build GRCh38
rs771926869 Chr.17: 38367879-38367879 on Build GRCh38
rs773590397 Chr.17: 38367932-38367932 on Build GRCh38
rs937667795 Chr.17: 38368200-38368200 on Build GRCh38
rs367552501 Chr.17: 38368080-38368080 on Build GRCh38
rs375597476 Chr.17: 38367856-38367856 on Build GRCh38
rs781333545 Chr.17: 38368077-38368077 on Build GRCh38
rs758930667 Chr.17: 38368010-38368010 on Build GRCh38
rs908105242 Chr.17: 38368152-38368152 on Build GRCh38
rs551383615 Chr.17: 38367980-38367980 on Build GRCh38
rs762105624 Chr.17: 38368059-38368060 on Build GRCh38
rs200851298 Chr.17: 38367854-38367854 on Build GRCh38
rs780188054 Chr.17: 38368085-38368085 on Build GRCh38
rs778111088 Chr.17: 38367849-38367849 on Build GRCh38
rs377748962 Chr.17: 38367871-38367871 on Build GRCh38
rs766186754 Chr.17: 38367940-38367940 on Build GRCh38
rs41487952 Chr.17: 38368206-38368206 on Build GRCh38
rs753075845 Chr.17: 38368005-38368005 on Build GRCh38
rs182048323 Chr.17: 38368120-38368120 on Build GRCh38
rs773176788 Chr.17: 38367893-38367893 on Build GRCh38
rs1054956414 Chr.17: 38367829-38367829 on Build GRCh38
rs587650322 Chr.17: 38367944-38367944 on Build GRCh38
rs148327709 Chr.17: 38368047-38368047 on Build GRCh38
rs758701482 Chr.17: 38367848-38367848 on Build GRCh38
rs41473346 Chr.17: 38367831-38367831 on Build GRCh38
rs909333396 Chr.17: 38368086-38368086 on Build GRCh38
rs761382101 Chr.17: 38368098-38368098 on Build GRCh38
rs953430501 Chr.17: 38368012-38368012 on Build GRCh38
rs587737643 Chr.17: 38367857-38367857 on Build GRCh38
rs760358428 Chr.17: 38367899-38367899 on Build GRCh38
rs756367254 Chr.17: 38368225-38368225 on Build GRCh38
rs764813391 Chr.17: 38367981-38367981 on Build GRCh38
rs778062040 Chr.17: 38368014-38368014 on Build GRCh38
rs370780030 Chr.17: 38368178-38368178 on Build GRCh38
rs780106672 Chr.17: 38368215-38368215 on Build GRCh38
rs757438431 Chr.17: 38368063-38368063 on Build GRCh38
rs974834439 Chr.17: 38368111-38368111 on Build GRCh38
rs769706559 Chr.17: 38368084-38368084 on Build GRCh38
rs587661886 Chr.17: 38368129-38368129 on Build GRCh38
rs186643475 Chr.17: 38368196-38368196 on Build GRCh38
rs41473152 Chr.17: 38367844-38367844 on Build GRCh38
rs143317326 Chr.17: 38367951-38367951 on Build GRCh38
rs766285056 Chr.17: 38368133-38368133 on Build GRCh38
rs748059025 Chr.17: 38368096-38368096 on Build GRCh38
rs587595023 Chr.17: 38367873-38367873 on Build GRCh38

View Full Product Details