Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 38395757-38396259 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SOCS7
Entrez Gene ID
30837
Gene Name
suppressor of cytokine signaling 7
Gene Aliases
NAP4, NCKAP4
UniGene
Hs.514132
Gene Chromosome Location
Chr.17: 38351876-38405593 on Build GRCh38
Transcripts
NM_014598.3 XM_017024551.1 XM_017024552.1

SNP Detail

SNP ID Chromosome Location
rs41322144 Chr.17: 38395822-38395822 on Build GRCh38
rs763089506 Chr.17: 38395980-38395980 on Build GRCh38
rs962467499 Chr.17: 38395881-38395881 on Build GRCh38
rs777101615 Chr.17: 38395965-38395965 on Build GRCh38
rs111451958 Chr.17: 38395829-38395829 on Build GRCh38
rs775721509 Chr.17: 38395971-38395971 on Build GRCh38
rs41524844 Chr.17: 38396142-38396142 on Build GRCh38
rs3890580 Chr.17: 38396203-38396203 on Build GRCh38
rs989250396 Chr.17: 38396166-38396166 on Build GRCh38
rs138060923 Chr.17: 38395923-38395923 on Build GRCh38
rs935412544 Chr.17: 38396224-38396224 on Build GRCh38
rs55687042 Chr.17: 38395937-38395937 on Build GRCh38
rs202233136 Chr.17: 38396002-38396002 on Build GRCh38
rs557542624 Chr.17: 38395922-38395922 on Build GRCh38
rs1051749345 Chr.17: 38396207-38396207 on Build GRCh38
rs543750640 Chr.17: 38396094-38396094 on Build GRCh38
rs34475821 Chr.17: 38396169-38396170 on Build GRCh38
rs371444577 Chr.17: 38395830-38395830 on Build GRCh38
rs752372086 Chr.17: 38395921-38395921 on Build GRCh38
rs970924105 Chr.17: 38396137-38396137 on Build GRCh38
rs747499302 Chr.17: 38395963-38395963 on Build GRCh38
rs565803028 Chr.17: 38396156-38396156 on Build GRCh38
rs147429718 Chr.17: 38395863-38395863 on Build GRCh38
rs371514542 Chr.17: 38395844-38395844 on Build GRCh38
rs750310007 Chr.17: 38395889-38395889 on Build GRCh38
rs760043583 Chr.17: 38395969-38395969 on Build GRCh38
rs187079008 Chr.17: 38396105-38396105 on Build GRCh38
rs200707695 Chr.17: 38395914-38395914 on Build GRCh38
rs995385017 Chr.17: 38396005-38396005 on Build GRCh38
rs150808963 Chr.17: 38396192-38396192 on Build GRCh38
rs764093296 Chr.17: 38395883-38395883 on Build GRCh38
rs1024637733 Chr.17: 38396104-38396104 on Build GRCh38
rs369848227 Chr.17: 38395925-38395925 on Build GRCh38
rs771610215 Chr.17: 38395964-38395964 on Build GRCh38
rs947974755 Chr.17: 38396181-38396181 on Build GRCh38
rs753683915 Chr.17: 38395918-38395918 on Build GRCh38
rs564777155 Chr.17: 38396070-38396070 on Build GRCh38
rs533729479 Chr.17: 38396077-38396077 on Build GRCh38
rs763149434 Chr.17: 38395875-38395875 on Build GRCh38
rs370631916 Chr.17: 38396208-38396208 on Build GRCh38
rs751615957 Chr.17: 38396009-38396009 on Build GRCh38
rs200758439 Chr.17: 38396163-38396163 on Build GRCh38
rs536855102 Chr.17: 38395820-38395820 on Build GRCh38
rs373701577 Chr.17: 38396184-38396187 on Build GRCh38
rs41326148 Chr.17: 38395824-38395824 on Build GRCh38
rs756077906 Chr.17: 38395900-38395900 on Build GRCh38
rs765595674 Chr.17: 38395868-38395868 on Build GRCh38
rs781241002 Chr.17: 38396225-38396225 on Build GRCh38
rs770126346 Chr.17: 38395970-38395970 on Build GRCh38
rs141149459 Chr.17: 38396133-38396133 on Build GRCh38
rs936533525 Chr.17: 38396151-38396151 on Build GRCh38
rs926334214 Chr.17: 38396143-38396143 on Build GRCh38
rs982472060 Chr.17: 38396089-38396089 on Build GRCh38
rs752714486 Chr.17: 38395869-38395869 on Build GRCh38

View Full Product Details