Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 16689555-16690096 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BZW2
Entrez Gene ID
28969
Gene Name
basic leucine zipper and W2 domains 2
Gene Aliases
HSPC028, MST017, MSTP017
UniGene
Hs.487635
Gene Chromosome Location
Chr.7: 16646134-16706523 on Build GRCh38
Transcripts
NM_001159767.1 NM_014038.2 NR_027624.1 XM_006715706.1 XM_006715707.1 XM_006715708.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4788903 c.551C>T p.A184V Chr.7: 16689806-16689806 on Build GRCh38
5574444 c.595G>A p.D199N Chr.7: 16689850-16689850 on Build GRCh38
1088072 c.649C>T p.L217F Chr.7: 16689904-16689904 on Build GRCh38
5872378 c.623G>A p.R208K Chr.7: 16689878-16689878 on Build GRCh38
3636911 c.554C>T p.S185L Chr.7: 16689809-16689809 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771398715 Chr.7: 16689916-16689916 on Build GRCh38
rs150344952 Chr.7: 16689744-16689744 on Build GRCh38
rs758537627 Chr.7: 16689894-16689894 on Build GRCh38
rs73679836 Chr.7: 16689747-16689747 on Build GRCh38
rs910593080 Chr.7: 16689914-16689914 on Build GRCh38
rs1037916165 Chr.7: 16689934-16689934 on Build GRCh38
rs944901518 Chr.7: 16689583-16689583 on Build GRCh38
rs6950993 Chr.7: 16689627-16689627 on Build GRCh38
rs772280528 Chr.7: 16689776-16689776 on Build GRCh38
rs757211845 Chr.7: 16689858-16689858 on Build GRCh38
rs183361343 Chr.7: 16690048-16690048 on Build GRCh38
rs773709644 Chr.7: 16689778-16689778 on Build GRCh38
rs946085593 Chr.7: 16690012-16690012 on Build GRCh38
rs773230749 Chr.7: 16689896-16689897 on Build GRCh38
rs922094848 Chr.7: 16689932-16689932 on Build GRCh38
rs746163227 Chr.7: 16689931-16689931 on Build GRCh38
rs572468645 Chr.7: 16689803-16689803 on Build GRCh38
rs1054876443 Chr.7: 16689629-16689629 on Build GRCh38
rs771321407 Chr.7: 16689800-16689800 on Build GRCh38
rs145957585 Chr.7: 16689805-16689805 on Build GRCh38
rs757022539 Chr.7: 16689748-16689748 on Build GRCh38
rs377010139 Chr.7: 16689761-16689761 on Build GRCh38
rs963810343 Chr.7: 16689927-16689927 on Build GRCh38
rs936912842 Chr.7: 16689607-16689607 on Build GRCh38
rs373727005 Chr.7: 16689819-16689819 on Build GRCh38
rs554968663 Chr.7: 16689760-16689760 on Build GRCh38
rs988109445 Chr.7: 16689624-16689624 on Build GRCh38
rs556729599 Chr.7: 16689745-16689745 on Build GRCh38
rs775156427 Chr.7: 16689806-16689806 on Build GRCh38
rs768240963 Chr.7: 16690014-16690014 on Build GRCh38
rs990322934 Chr.7: 16689966-16689966 on Build GRCh38
rs899059014 Chr.7: 16689730-16689730 on Build GRCh38
rs770173639 Chr.7: 16689944-16689944 on Build GRCh38
rs552136026 Chr.7: 16689636-16689636 on Build GRCh38
rs1032523070 Chr.7: 16689832-16689832 on Build GRCh38
rs377414985 Chr.7: 16689768-16689768 on Build GRCh38
rs114116855 Chr.7: 16689712-16689712 on Build GRCh38
rs141321905 Chr.7: 16689904-16689904 on Build GRCh38
rs747527030 Chr.7: 16689784-16689784 on Build GRCh38
rs375628464 Chr.7: 16689869-16689869 on Build GRCh38
rs928737665 Chr.7: 16690024-16690024 on Build GRCh38
rs988421846 Chr.7: 16689862-16689862 on Build GRCh38
rs987495176 Chr.7: 16689911-16689911 on Build GRCh38
rs557364963 Chr.7: 16689664-16689664 on Build GRCh38
rs370591549 Chr.7: 16689807-16689807 on Build GRCh38
rs533960818 Chr.7: 16689666-16689666 on Build GRCh38
rs780911184 Chr.7: 16689749-16689749 on Build GRCh38
rs925485321 Chr.7: 16689584-16689584 on Build GRCh38
rs929210211 Chr.7: 16689994-16689994 on Build GRCh38
rs566848858 Chr.7: 16689644-16689644 on Build GRCh38
rs907289651 Chr.7: 16689706-16689706 on Build GRCh38
rs138783404 Chr.7: 16689870-16689870 on Build GRCh38
rs899028032 Chr.7: 16689962-16689962 on Build GRCh38
rs913243636 Chr.7: 16689976-16689976 on Build GRCh38
rs759167537 Chr.7: 16689804-16689804 on Build GRCh38
rs748392463 Chr.7: 16689887-16689887 on Build GRCh38
rs760256408 Chr.7: 16689897-16689897 on Build GRCh38
rs957875151 Chr.7: 16689951-16689951 on Build GRCh38
rs752449068 Chr.7: 16689793-16689793 on Build GRCh38
rs1001816377 Chr.7: 16689711-16689711 on Build GRCh38
rs555791953 Chr.7: 16689680-16689680 on Build GRCh38
rs771171381 Chr.7: 16689684-16689684 on Build GRCh38
rs550690348 Chr.7: 16689924-16689924 on Build GRCh38
rs888954879 Chr.7: 16689662-16689662 on Build GRCh38
rs771945976 Chr.7: 16689781-16689782 on Build GRCh38
rs57937778 Chr.7: 16689843-16689843 on Build GRCh38
rs933380276 Chr.7: 16689633-16689633 on Build GRCh38
rs993395168 Chr.7: 16689742-16689742 on Build GRCh38
rs528975449 Chr.7: 16689889-16689889 on Build GRCh38
rs767317583 Chr.7: 16689863-16689863 on Build GRCh38

View Full Product Details