Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 43335606-43336038 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAL
Entrez Gene ID
161823
Gene Name
adenosine deaminase like
Gene Aliases
-
UniGene
Hs.533913
Gene Chromosome Location
Chr.15: 43330351-43354555 on Build GRCh38
Transcripts
NM_001012969.3 NM_001159280.2 NM_001324364.1 NM_001324365.1 NM_001324366.1 NM_001324367.1 NM_001324368.1 XM_011521291.2 XM_017021964.1 XM_017021965.1 XM_017021966.1 XM_017021967.1 XM_017021968.1 XM_017021969.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5895400 c.64-36_64-2del35 p.? Chr.15: 43335660-43335694 on Build GRCh38
1587861 c.81G>T p.L27F Chr.15: 43335713-43335713 on Build GRCh38
1135660 c.176A>G p.K59R Chr.15: 43335808-43335808 on Build GRCh38
5500791 c.70C>T p.H24Y Chr.15: 43335702-43335702 on Build GRCh38
7097388 c.177G>T p.K59N Chr.15: 43335809-43335809 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774542683 Chr.15: 43335669-43335669 on Build GRCh38
rs201110280 Chr.15: 43335751-43335751 on Build GRCh38
rs750034798 Chr.15: 43335706-43335706 on Build GRCh38
rs759739442 Chr.15: 43335673-43335673 on Build GRCh38
rs533606745 Chr.15: 43335924-43335924 on Build GRCh38
rs770061685 Chr.15: 43335647-43335647 on Build GRCh38
rs769806846 Chr.15: 43335846-43335846 on Build GRCh38
rs762559206 Chr.15: 43335708-43335708 on Build GRCh38
rs774001812 Chr.15: 43335868-43335868 on Build GRCh38
rs745875165 Chr.15: 43335822-43335822 on Build GRCh38
rs772056678 Chr.15: 43335827-43335827 on Build GRCh38
rs749710717 Chr.15: 43335791-43335791 on Build GRCh38
rs754554268 Chr.15: 43335748-43335748 on Build GRCh38
rs562539706 Chr.15: 43335691-43335691 on Build GRCh38
rs779344931 Chr.15: 43335804-43335804 on Build GRCh38
rs765553055 Chr.15: 43335986-43335986 on Build GRCh38
rs959068796 Chr.15: 43335655-43335655 on Build GRCh38
rs748064693 Chr.15: 43335834-43335834 on Build GRCh38
rs983366592 Chr.15: 43335837-43335837 on Build GRCh38
rs567819657 Chr.15: 43335883-43335883 on Build GRCh38
rs201697344 Chr.15: 43335814-43335814 on Build GRCh38
rs1049745807 Chr.15: 43335978-43335978 on Build GRCh38
rs762791153 Chr.15: 43335863-43335863 on Build GRCh38
rs570531972 Chr.15: 43335984-43335984 on Build GRCh38
rs753161723 Chr.15: 43335771-43335771 on Build GRCh38
rs202037235 Chr.15: 43335686-43335686 on Build GRCh38
rs778031712 Chr.15: 43335785-43335785 on Build GRCh38
rs762226014 Chr.15: 43335704-43335704 on Build GRCh38
rs918048313 Chr.15: 43335710-43335710 on Build GRCh38
rs907705856 Chr.15: 43335845-43335845 on Build GRCh38
rs768005001 Chr.15: 43335750-43335750 on Build GRCh38
rs146713520 Chr.15: 43335779-43335779 on Build GRCh38
rs766066483 Chr.15: 43335720-43335720 on Build GRCh38
rs926989786 Chr.15: 43335888-43335888 on Build GRCh38
rs757531267 Chr.15: 43335792-43335792 on Build GRCh38
rs770531552 Chr.15: 43335777-43335777 on Build GRCh38
rs777995732 Chr.15: 43335649-43335649 on Build GRCh38
rs751052393 Chr.15: 43335727-43335727 on Build GRCh38
rs917859191 Chr.15: 43335799-43335799 on Build GRCh38
rs766019671 Chr.15: 43335865-43335865 on Build GRCh38
rs776436865 Chr.15: 43335679-43335679 on Build GRCh38
rs749546014 Chr.15: 43335654-43335654 on Build GRCh38
rs768634420 Chr.15: 43335678-43335678 on Build GRCh38
rs536630633 Chr.15: 43335857-43335857 on Build GRCh38
rs191087126 Chr.15: 43335963-43335963 on Build GRCh38
rs932663199 Chr.15: 43335951-43335951 on Build GRCh38
rs1549525 Chr.15: 43335657-43335657 on Build GRCh38
rs772965678 Chr.15: 43335850-43335850 on Build GRCh38
rs1054245924 Chr.15: 43335925-43335925 on Build GRCh38
rs1038895369 Chr.15: 43335918-43335918 on Build GRCh38
rs776580151 Chr.15: 43335831-43335831 on Build GRCh38
rs970634005 Chr.15: 43335825-43335825 on Build GRCh38

View Full Product Details