Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 49033340-49033938 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TFE3
Entrez Gene ID
7030
Gene Name
transcription factor binding to IGHM enhancer 3
Gene Aliases
RCCP2, RCCX1, TFEA, bHLHe33
UniGene
Hs.730740
Gene Chromosome Location
Chr.X: 49028726-49043517 on Build GRCh38
Transcripts
NM_001282142.1 NM_006521.5

SNP Detail

SNP ID Chromosome Location
rs369475777 Chr.X: 49033711-49033711 on Build GRCh38
rs982554029 Chr.X: 49033382-49033382 on Build GRCh38
rs782358989 Chr.X: 49033793-49033793 on Build GRCh38
rs367560398 Chr.X: 49033435-49033435 on Build GRCh38
rs781929039 Chr.X: 49033796-49033796 on Build GRCh38
rs201808352 Chr.X: 49033512-49033512 on Build GRCh38
rs960063584 Chr.X: 49033837-49033837 on Build GRCh38
rs369698146 Chr.X: 49033588-49033588 on Build GRCh38
rs781835191 Chr.X: 49033830-49033830 on Build GRCh38
rs377187631 Chr.X: 49033583-49033583 on Build GRCh38
rs782115411 Chr.X: 49033778-49033778 on Build GRCh38
rs890548874 Chr.X: 49033655-49033655 on Build GRCh38
rs782737875 Chr.X: 49033371-49033371 on Build GRCh38
rs782076934 Chr.X: 49033712-49033712 on Build GRCh38
rs782034465 Chr.X: 49033768-49033768 on Build GRCh38
rs185587819 Chr.X: 49033705-49033705 on Build GRCh38
rs201168885 Chr.X: 49033699-49033699 on Build GRCh38
rs782004627 Chr.X: 49033777-49033777 on Build GRCh38
rs782445226 Chr.X: 49033418-49033418 on Build GRCh38
rs41305381 Chr.X: 49033443-49033443 on Build GRCh38
rs189222789 Chr.X: 49033825-49033825 on Build GRCh38
rs782369351 Chr.X: 49033766-49033766 on Build GRCh38
rs782379337 Chr.X: 49033720-49033720 on Build GRCh38
rs782697975 Chr.X: 49033558-49033558 on Build GRCh38
rs782656388 Chr.X: 49033709-49033709 on Build GRCh38
rs781846888 Chr.X: 49033824-49033824 on Build GRCh38
rs782342097 Chr.X: 49033600-49033600 on Build GRCh38
rs781863465 Chr.X: 49033693-49033693 on Build GRCh38
rs782784061 Chr.X: 49033484-49033484 on Build GRCh38
rs991221441 Chr.X: 49033863-49033863 on Build GRCh38
rs372314452 Chr.X: 49033694-49033694 on Build GRCh38
rs782635252 Chr.X: 49033688-49033688 on Build GRCh38
rs3027471 Chr.X: 49033821-49033821 on Build GRCh38
rs782613018 Chr.X: 49033698-49033698 on Build GRCh38
rs782316450 Chr.X: 49033606-49033606 on Build GRCh38
rs886969467 Chr.X: 49033786-49033786 on Build GRCh38
rs781845261 Chr.X: 49033388-49033388 on Build GRCh38
rs782535970 Chr.X: 49033591-49033591 on Build GRCh38
rs782716693 Chr.X: 49033826-49033826 on Build GRCh38
rs368263592 Chr.X: 49033447-49033447 on Build GRCh38
rs782120251 Chr.X: 49033640-49033640 on Build GRCh38
rs782106588 Chr.X: 49033546-49033546 on Build GRCh38
rs782075514 Chr.X: 49033816-49033816 on Build GRCh38
rs199560694 Chr.X: 49033419-49033419 on Build GRCh38
rs1051885706 Chr.X: 49033638-49033638 on Build GRCh38
rs781963537 Chr.X: 49033612-49033612 on Build GRCh38

View Full Product Details