Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 58196000-58196528 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IPMK
Entrez Gene ID
253430
Gene Name
inositol polyphosphate multikinase
Gene Aliases
-
UniGene
Hs.30280
Gene Chromosome Location
Chr.10: 58191517-58269837 on Build GRCh38
Transcripts
NM_152230.4 XM_011539565.2 XM_017016013.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
386277 c.824A>T p.E275V Chr.10: 58196503-58196503 on Build GRCh38
7137530 c.809A>G p.D270G Chr.10: 58196518-58196518 on Build GRCh38
5969850 c.820G>A p.A274T Chr.10: 58196507-58196507 on Build GRCh38
3807382 c.906T>G p.A302A Chr.10: 58196421-58196421 on Build GRCh38
7295722 c.1085T>C p.L362P Chr.10: 58196242-58196242 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs541806725 Chr.10: 58196401-58196401 on Build GRCh38
rs375383787 Chr.10: 58196371-58196371 on Build GRCh38
rs775609595 Chr.10: 58196219-58196219 on Build GRCh38
rs764957765 Chr.10: 58196228-58196228 on Build GRCh38
rs201334665 Chr.10: 58196189-58196189 on Build GRCh38
rs774063548 Chr.10: 58196481-58196481 on Build GRCh38
rs780067236 Chr.10: 58196444-58196444 on Build GRCh38
rs755724483 Chr.10: 58196284-58196284 on Build GRCh38
rs761486525 Chr.10: 58196099-58196099 on Build GRCh38
rs199822749 Chr.10: 58196224-58196224 on Build GRCh38
rs762947078 Chr.10: 58196238-58196238 on Build GRCh38
rs1044344662 Chr.10: 58196411-58196411 on Build GRCh38
rs776881750 Chr.10: 58196462-58196462 on Build GRCh38
rs759410890 Chr.10: 58196369-58196369 on Build GRCh38
rs770946028 Chr.10: 58196325-58196325 on Build GRCh38
rs754683419 Chr.10: 58196164-58196164 on Build GRCh38
rs766290279 Chr.10: 58196244-58196244 on Build GRCh38
rs148537566 Chr.10: 58196488-58196488 on Build GRCh38
rs772305007 Chr.10: 58196338-58196338 on Build GRCh38
rs768810591 Chr.10: 58196457-58196457 on Build GRCh38
rs772940182 Chr.10: 58196353-58196353 on Build GRCh38
rs750168956 Chr.10: 58196233-58196233 on Build GRCh38
rs142574341 Chr.10: 58196406-58196406 on Build GRCh38
rs535168447 Chr.10: 58196368-58196368 on Build GRCh38
rs762715336 Chr.10: 58196363-58196363 on Build GRCh38
rs772459420 Chr.10: 58196216-58196216 on Build GRCh38
rs371083086 Chr.10: 58196252-58196252 on Build GRCh38
rs779545983 Chr.10: 58196129-58196129 on Build GRCh38
rs748319721 Chr.10: 58196465-58196465 on Build GRCh38
rs750353387 Chr.10: 58196112-58196112 on Build GRCh38
rs766938208 Chr.10: 58196370-58196370 on Build GRCh38
rs749762196 Chr.10: 58196184-58196184 on Build GRCh38
rs185185082 Chr.10: 58196339-58196339 on Build GRCh38
rs913891219 Chr.10: 58196400-58196400 on Build GRCh38
rs144385789 Chr.10: 58196186-58196186 on Build GRCh38
rs773621587 Chr.10: 58196068-58196068 on Build GRCh38
rs755776415 Chr.10: 58196398-58196398 on Build GRCh38
rs747797228 Chr.10: 58196180-58196180 on Build GRCh38
rs368295594 Chr.10: 58196425-58196425 on Build GRCh38
rs1025935796 Chr.10: 58196218-58196218 on Build GRCh38
rs761503472 Chr.10: 58196501-58196501 on Build GRCh38
rs748448778 Chr.10: 58196046-58196046 on Build GRCh38
rs781604678 Chr.10: 58196041-58196041 on Build GRCh38
rs771146097 Chr.10: 58196089-58196089 on Build GRCh38
rs749648025 Chr.10: 58196292-58196292 on Build GRCh38
rs754291508 Chr.10: 58196413-58196413 on Build GRCh38
rs146319184 Chr.10: 58196431-58196431 on Build GRCh38
rs548900003 Chr.10: 58196113-58196113 on Build GRCh38
rs373653986 Chr.10: 58196102-58196102 on Build GRCh38
rs2275443 Chr.10: 58196280-58196280 on Build GRCh38
rs770372872 Chr.10: 58196059-58196059 on Build GRCh38
rs765751052 Chr.10: 58196343-58196343 on Build GRCh38
rs760196498 Chr.10: 58196494-58196494 on Build GRCh38
rs780801664 Chr.10: 58196170-58196170 on Build GRCh38
rs747099599 Chr.10: 58196452-58196452 on Build GRCh38
rs746395205 Chr.10: 58196428-58196428 on Build GRCh38
rs759662679 Chr.10: 58196206-58196206 on Build GRCh38
rs774909915 Chr.10: 58196493-58196493 on Build GRCh38
rs76733713 Chr.10: 58196455-58196455 on Build GRCh38
rs774628464 Chr.10: 58196203-58196203 on Build GRCh38
rs150092014 Chr.10: 58196249-58196249 on Build GRCh38
rs757723741 Chr.10: 58196415-58196415 on Build GRCh38
rs759900712 Chr.10: 58196095-58196095 on Build GRCh38
rs746340310 Chr.10: 58196324-58196324 on Build GRCh38
rs201911489 Chr.10: 58196182-58196182 on Build GRCh38
rs751066669 Chr.10: 58196155-58196155 on Build GRCh38
rs770725627 Chr.10: 58196472-58196472 on Build GRCh38
rs766345094 Chr.10: 58196365-58196365 on Build GRCh38
rs763467528 Chr.10: 58196073-58196073 on Build GRCh38
rs530321996 Chr.10: 58196036-58196036 on Build GRCh38
rs149331771 Chr.10: 58196282-58196282 on Build GRCh38
rs779527924 Chr.10: 58196309-58196309 on Build GRCh38
rs767834995 Chr.10: 58196096-58196096 on Build GRCh38
rs146242677 Chr.10: 58196296-58196296 on Build GRCh38
rs774503755 Chr.10: 58196090-58196090 on Build GRCh38
rs753571526 Chr.10: 58196499-58196499 on Build GRCh38
rs778062653 Chr.10: 58196288-58196288 on Build GRCh38

View Full Product Details