Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 69632908-69633341 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C10orf35
Entrez Gene ID
219738
Gene Name
chromosome 10 open reading frame 35
Gene Aliases
-
UniGene
Hs.522992
Gene Chromosome Location
Chr.10: 69630241-69633599 on Build GRCh38
Transcripts
NM_145306.2 XM_005269606.1 XM_005269608.3 XM_011539455.2 XM_017015836.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3751916 c.243T>C p.L81L Chr.10: 69632936-69632936 on Build GRCh38
1348810 c.264G>A p.P88P Chr.10: 69632957-69632957 on Build GRCh38
6443961 c.276C>G p.I92M Chr.10: 69632969-69632969 on Build GRCh38
1348811 c.316C>A p.L106I Chr.10: 69633009-69633009 on Build GRCh38
1246349 c.322C>A p.L108M Chr.10: 69633015-69633015 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765510575 Chr.10: 69632957-69632957 on Build GRCh38
rs891273986 Chr.10: 69633268-69633268 on Build GRCh38
rs554299114 Chr.10: 69633162-69633162 on Build GRCh38
rs554097533 Chr.10: 69633074-69633074 on Build GRCh38
rs145356439 Chr.10: 69632945-69632945 on Build GRCh38
rs762044523 Chr.10: 69632956-69632956 on Build GRCh38
rs41305047 Chr.10: 69633224-69633224 on Build GRCh38
rs1030535176 Chr.10: 69632978-69632978 on Build GRCh38
rs187756447 Chr.10: 69633208-69633208 on Build GRCh38
rs762295272 Chr.10: 69633079-69633079 on Build GRCh38
rs753261942 Chr.10: 69633063-69633063 on Build GRCh38
rs546464168 Chr.10: 69633184-69633184 on Build GRCh38
rs756140050 Chr.10: 69632983-69632983 on Build GRCh38
rs1016187687 Chr.10: 69632941-69632941 on Build GRCh38
rs900631432 Chr.10: 69633278-69633278 on Build GRCh38
rs996133353 Chr.10: 69633290-69633290 on Build GRCh38
rs529746502 Chr.10: 69633319-69633319 on Build GRCh38
rs995512880 Chr.10: 69632952-69632952 on Build GRCh38
rs979224245 Chr.10: 69633070-69633070 on Build GRCh38
rs572536264 Chr.10: 69633174-69633174 on Build GRCh38
rs763060886 Chr.10: 69633109-69633109 on Build GRCh38
rs374963949 Chr.10: 69633129-69633129 on Build GRCh38
rs764492900 Chr.10: 69632937-69632937 on Build GRCh38
rs745505897 Chr.10: 69633056-69633056 on Build GRCh38
rs1051304080 Chr.10: 69633230-69633238 on Build GRCh38
rs989345702 Chr.10: 69633014-69633014 on Build GRCh38
rs750596351 Chr.10: 69632960-69632960 on Build GRCh38
rs764060126 Chr.10: 69633044-69633045 on Build GRCh38
rs765626119 Chr.10: 69633179-69633179 on Build GRCh38
rs1052152 Chr.10: 69632936-69632936 on Build GRCh38
rs758570192 Chr.10: 69632963-69632963 on Build GRCh38
rs774485331 Chr.10: 69633075-69633075 on Build GRCh38
rs773521857 Chr.10: 69633107-69633107 on Build GRCh38
rs756947920 Chr.10: 69633032-69633032 on Build GRCh38
rs184130654 Chr.10: 69633157-69633157 on Build GRCh38
rs765513352 Chr.10: 69633097-69633097 on Build GRCh38
rs374261908 Chr.10: 69633004-69633004 on Build GRCh38
rs752728246 Chr.10: 69632982-69632982 on Build GRCh38
rs544192606 Chr.10: 69633264-69633264 on Build GRCh38
rs141324225 Chr.10: 69632969-69632969 on Build GRCh38
rs966834024 Chr.10: 69633066-69633066 on Build GRCh38
rs556176739 Chr.10: 69633125-69633125 on Build GRCh38
rs774960137 Chr.10: 69633064-69633064 on Build GRCh38
rs769181998 Chr.10: 69633076-69633076 on Build GRCh38
rs777808950 Chr.10: 69632991-69632991 on Build GRCh38
rs917837048 Chr.10: 69633151-69633151 on Build GRCh38
rs777064129 Chr.10: 69633078-69633078 on Build GRCh38
rs11598407 Chr.10: 69633281-69633281 on Build GRCh38
rs981956319 Chr.10: 69633039-69633039 on Build GRCh38
rs1132075 Chr.10: 69633269-69633269 on Build GRCh38
rs761611484 Chr.10: 69632984-69632984 on Build GRCh38
rs778688304 Chr.10: 69633052-69633052 on Build GRCh38
rs960835830 Chr.10: 69633087-69633087 on Build GRCh38

View Full Product Details