Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 206766532-206767068 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FASTKD2
Entrez Gene ID
22868
Gene Name
FAST kinase domains 2
Gene Aliases
KIAA0971
UniGene
Hs.5930
Gene Chromosome Location
Chr.2: 206765388-206868284 on Build GRCh38
Transcripts
NM_001136193.1 NM_001136194.1 NM_014929.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1015493 c.184A>C p.N62H Chr.2: 206766877-206766877 on Build GRCh38
4418873 c.44G>A p.S15N Chr.2: 206766737-206766737 on Build GRCh38
2714311 c.26G>T p.G9V Chr.2: 206766719-206766719 on Build GRCh38
2714310 c.25G>T p.G9* Chr.2: 206766718-206766718 on Build GRCh38
3576880 c.152C>T p.P51L Chr.2: 206766845-206766845 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143223304 Chr.2: 206766746-206766746 on Build GRCh38
rs898028998 Chr.2: 206766812-206766812 on Build GRCh38
rs757770737 Chr.2: 206767042-206767042 on Build GRCh38
rs769183701 Chr.2: 206766684-206766684 on Build GRCh38
rs763480439 Chr.2: 206767010-206767014 on Build GRCh38
rs374830420 Chr.2: 206766918-206766918 on Build GRCh38
rs111996157 Chr.2: 206766685-206766685 on Build GRCh38
rs768025505 Chr.2: 206766778-206766779 on Build GRCh38
rs577645130 Chr.2: 206766838-206766838 on Build GRCh38
rs1052476894 Chr.2: 206766853-206766853 on Build GRCh38
rs1040247354 Chr.2: 206766764-206766764 on Build GRCh38
rs547620965 Chr.2: 206766847-206766847 on Build GRCh38
rs756353957 Chr.2: 206766779-206766779 on Build GRCh38
rs752757043 Chr.2: 206767019-206767019 on Build GRCh38
rs779536604 Chr.2: 206766808-206766808 on Build GRCh38
rs901836718 Chr.2: 206766561-206766561 on Build GRCh38
rs781534706 Chr.2: 206766658-206766658 on Build GRCh38
rs747511083 Chr.2: 206767030-206767030 on Build GRCh38
rs763693414 Chr.2: 206766870-206766870 on Build GRCh38
rs765204520 Chr.2: 206766657-206766657 on Build GRCh38
rs1017533007 Chr.2: 206766620-206766620 on Build GRCh38
rs778865889 Chr.2: 206766903-206766903 on Build GRCh38
rs750218375 Chr.2: 206766886-206766886 on Build GRCh38
rs771403858 Chr.2: 206766976-206766976 on Build GRCh38
rs568472244 Chr.2: 206766809-206766809 on Build GRCh38
rs1017559758 Chr.2: 206766887-206766887 on Build GRCh38
rs549677418 Chr.2: 206766755-206766755 on Build GRCh38
rs866591177 Chr.2: 206766591-206766591 on Build GRCh38
rs956260223 Chr.2: 206766735-206766736 on Build GRCh38
rs767395981 Chr.2: 206767006-206767006 on Build GRCh38
rs752284309 Chr.2: 206766736-206766736 on Build GRCh38
rs886055505 Chr.2: 206766679-206766679 on Build GRCh38
rs199972302 Chr.2: 206766723-206766723 on Build GRCh38
rs756555125 Chr.2: 206766892-206766893 on Build GRCh38
rs372274271 Chr.2: 206766911-206766911 on Build GRCh38
rs772567572 Chr.2: 206766814-206766814 on Build GRCh38
rs747423460 Chr.2: 206766928-206766928 on Build GRCh38
rs765453133 Chr.2: 206767016-206767016 on Build GRCh38
rs778197337 Chr.2: 206767029-206767029 on Build GRCh38
rs1012121296 Chr.2: 206766620-206766620 on Build GRCh38
rs3762568 Chr.2: 206766737-206766737 on Build GRCh38
rs150457139 Chr.2: 206766885-206766885 on Build GRCh38
rs996238032 Chr.2: 206766562-206766562 on Build GRCh38
rs768295259 Chr.2: 206766985-206766985 on Build GRCh38
rs749318348 Chr.2: 206766681-206766681 on Build GRCh38
rs757378301 Chr.2: 206766908-206766908 on Build GRCh38
rs773112768 Chr.2: 206767007-206767007 on Build GRCh38
rs977268504 Chr.2: 206766950-206766950 on Build GRCh38
rs570728838 Chr.2: 206766570-206766570 on Build GRCh38
rs764930961 Chr.2: 206766733-206766733 on Build GRCh38
rs954735024 Chr.2: 206766584-206766584 on Build GRCh38
rs773465848 Chr.2: 206766819-206766819 on Build GRCh38
rs775258027 Chr.2: 206766843-206766843 on Build GRCh38
rs989538082 Chr.2: 206766641-206766641 on Build GRCh38
rs112551007 Chr.2: 206766683-206766683 on Build GRCh38
rs140026989 Chr.2: 206766567-206766567 on Build GRCh38
rs368791899 Chr.2: 206766863-206766863 on Build GRCh38
rs766162714 Chr.2: 206767014-206767014 on Build GRCh38
rs144137525 Chr.2: 206766900-206766900 on Build GRCh38
rs1050537783 Chr.2: 206766807-206766807 on Build GRCh38
rs758479510 Chr.2: 206767021-206767021 on Build GRCh38
rs753566860 Chr.2: 206767015-206767015 on Build GRCh38
rs1030582122 Chr.2: 206766583-206766583 on Build GRCh38
rs201816518 Chr.2: 206766756-206766756 on Build GRCh38
rs766637816 Chr.2: 206766891-206766891 on Build GRCh38
rs773754189 Chr.2: 206766991-206766991 on Build GRCh38
rs140324512 Chr.2: 206766983-206766983 on Build GRCh38
rs139855985 Chr.2: 206767001-206767001 on Build GRCh38
rs371471616 Chr.2: 206766688-206766688 on Build GRCh38
rs1012286338 Chr.2: 206766940-206766940 on Build GRCh38
rs961847801 Chr.2: 206767041-206767041 on Build GRCh38
rs756692703 Chr.2: 206766974-206766979 on Build GRCh38
rs754050859 Chr.2: 206766893-206766893 on Build GRCh38
rs866874741 Chr.2: 206766759-206766759 on Build GRCh38
rs863223961 Chr.2: 206766954-206766954 on Build GRCh38
rs1020554490 Chr.2: 206766629-206766629 on Build GRCh38
rs145438423 Chr.2: 206766664-206766664 on Build GRCh38
rs141447598 Chr.2: 206766842-206766842 on Build GRCh38
rs750173012 Chr.2: 206766775-206766775 on Build GRCh38
rs886044127 Chr.2: 206766927-206766927 on Build GRCh38
rs768701270 Chr.2: 206766792-206766792 on Build GRCh38
rs368286687 Chr.2: 206766876-206766876 on Build GRCh38
rs863223960 Chr.2: 206766875-206766875 on Build GRCh38
rs751406725 Chr.2: 206766654-206766654 on Build GRCh38
rs777810805 Chr.2: 206766653-206766653 on Build GRCh38
rs1003116172 Chr.2: 206766660-206766660 on Build GRCh38
rs751014977 Chr.2: 206766871-206766871 on Build GRCh38
rs760522658 Chr.2: 206767012-206767012 on Build GRCh38
rs771505646 Chr.2: 206766902-206766902 on Build GRCh38
rs751798516 Chr.2: 206766907-206766907 on Build GRCh38
rs977779943 Chr.2: 206766670-206766670 on Build GRCh38
rs775348698 Chr.2: 206766981-206766981 on Build GRCh38
rs749411560 Chr.2: 206766783-206766783 on Build GRCh38
rs755873619 Chr.2: 206766890-206766890 on Build GRCh38
rs984543218 Chr.2: 206766994-206766994 on Build GRCh38
rs748602309 Chr.2: 206766813-206766813 on Build GRCh38
rs556217379 Chr.2: 206766686-206766686 on Build GRCh38
rs772314275 Chr.2: 206766697-206766697 on Build GRCh38
rs780122499 Chr.2: 206766673-206766673 on Build GRCh38
rs201275973 Chr.2: 206766699-206766699 on Build GRCh38
rs931974124 Chr.2: 206766806-206766806 on Build GRCh38
rs536180346 Chr.2: 206766769-206766769 on Build GRCh38
rs147727753 Chr.2: 206766722-206766722 on Build GRCh38

View Full Product Details