Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 49034003-49034484 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TFE3
Entrez Gene ID
7030
Gene Name
transcription factor binding to IGHM enhancer 3
Gene Aliases
RCCP2, RCCX1, TFEA, bHLHe33
UniGene
Hs.730740
Gene Chromosome Location
Chr.X: 49028726-49043517 on Build GRCh38
Transcripts
NM_001282142.1 NM_006521.5

SNP Detail

SNP ID Chromosome Location
rs782436702 Chr.X: 49034092-49034092 on Build GRCh38
rs1031274197 Chr.X: 49034388-49034388 on Build GRCh38
rs781936583 Chr.X: 49034216-49034216 on Build GRCh38
rs782089868 Chr.X: 49034209-49034209 on Build GRCh38
rs781842431 Chr.X: 49034296-49034296 on Build GRCh38
rs782531998 Chr.X: 49034282-49034282 on Build GRCh38
rs782812641 Chr.X: 49034222-49034222 on Build GRCh38
rs781927976 Chr.X: 49034459-49034459 on Build GRCh38
rs782207856 Chr.X: 49034373-49034373 on Build GRCh38
rs782188306 Chr.X: 49034145-49034145 on Build GRCh38
rs782261224 Chr.X: 49034122-49034122 on Build GRCh38
rs912087174 Chr.X: 49034458-49034458 on Build GRCh38
rs781822962 Chr.X: 49034119-49034119 on Build GRCh38
rs368895179 Chr.X: 49034256-49034256 on Build GRCh38
rs782370157 Chr.X: 49034446-49034446 on Build GRCh38
rs113854587 Chr.X: 49034292-49034293 on Build GRCh38
rs782793485 Chr.X: 49034034-49034034 on Build GRCh38
rs150779978 Chr.X: 49034195-49034195 on Build GRCh38
rs782614531 Chr.X: 49034126-49034126 on Build GRCh38
rs782625054 Chr.X: 49034435-49034435 on Build GRCh38
rs782593756 Chr.X: 49034082-49034082 on Build GRCh38
rs55909867 Chr.X: 49034439-49034439 on Build GRCh38
rs1013118349 Chr.X: 49034355-49034355 on Build GRCh38
rs781949099 Chr.X: 49034207-49034207 on Build GRCh38
rs782228674 Chr.X: 49034114-49034114 on Build GRCh38
rs912024196 Chr.X: 49034031-49034031 on Build GRCh38
rs1022649074 Chr.X: 49034358-49034358 on Build GRCh38
rs782239289 Chr.X: 49034197-49034197 on Build GRCh38
rs782521367 Chr.X: 49034079-49034079 on Build GRCh38
rs782555413 Chr.X: 49034113-49034113 on Build GRCh38
rs956491286 Chr.X: 49034396-49034396 on Build GRCh38
rs781807680 Chr.X: 49034428-49034428 on Build GRCh38
rs978408589 Chr.X: 49034372-49034372 on Build GRCh38
rs782148893 Chr.X: 49034217-49034217 on Build GRCh38
rs1004095655 Chr.X: 49034318-49034318 on Build GRCh38
rs988063122 Chr.X: 49034026-49034026 on Build GRCh38
rs782538136 Chr.X: 49034095-49034096 on Build GRCh38
rs782333221 Chr.X: 49034153-49034153 on Build GRCh38
rs782346414 Chr.X: 49034125-49034125 on Build GRCh38
rs141452086 Chr.X: 49034338-49034338 on Build GRCh38
rs781902060 Chr.X: 49034362-49034363 on Build GRCh38
rs895643717 Chr.X: 49034349-49034349 on Build GRCh38
rs1047731769 Chr.X: 49034206-49034206 on Build GRCh38
rs899189335 Chr.X: 49034146-49034146 on Build GRCh38
rs782725136 Chr.X: 49034295-49034295 on Build GRCh38
rs139188133 Chr.X: 49034214-49034214 on Build GRCh38
rs782437066 Chr.X: 49034297-49034297 on Build GRCh38
rs782584600 Chr.X: 49034093-49034093 on Build GRCh38
rs3027470 Chr.X: 49034200-49034200 on Build GRCh38
rs968382643 Chr.X: 49034365-49034365 on Build GRCh38
rs781888101 Chr.X: 49034097-49034097 on Build GRCh38
rs782377455 Chr.X: 49034203-49034203 on Build GRCh38

View Full Product Details