Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 112488335-112488516 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTPN11
Entrez Gene ID
5781
Gene Name
protein tyrosine phosphatase, non-receptor type 11
Gene Aliases
BPTP3, CFC, JMML, METCDS, NS1, PTP-1D, PTP2C, SH-PTP2, SH-PTP3, SHP2
UniGene
Hs.506852
Gene Chromosome Location
Chr.12: 112418732-112509913 on Build GRCh38
Transcripts
NM_002834.3 XM_006719526.2 XM_011538613.2 XM_017019722.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
170715 c.1403C>T p.T468M Chr.12: 112488466-112488466 on Build GRCh38
3935746 c.1420A>G p.I474V Chr.12: 112488483-112488483 on Build GRCh38
935283 c.1387A>C p.I463L Chr.12: 112488450-112488450 on Build GRCh38
5611360 c.1390_1391GG>AA p.G464N Chr.12: 112488453-112488454 on Build GRCh38
6986171 c.1390G>T p.G464C Chr.12: 112488453-112488453 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs916103771 Chr.12: 112488388-112488388 on Build GRCh38
rs886039711 Chr.12: 112488465-112488466 on Build GRCh38
rs121918469 Chr.12: 112488454-112488454 on Build GRCh38
rs757361438 Chr.12: 112488420-112488420 on Build GRCh38
rs930734273 Chr.12: 112488364-112488364 on Build GRCh38
rs121918457 Chr.12: 112488466-112488466 on Build GRCh38
rs587781130 Chr.12: 112488467-112488467 on Build GRCh38
rs397509344 Chr.12: 112488445-112488445 on Build GRCh38
rs193922657 Chr.12: 112488429-112488429 on Build GRCh38
rs187807440 Chr.12: 112488384-112488384 on Build GRCh38
rs779978269 Chr.12: 112488471-112488471 on Build GRCh38
rs984859449 Chr.12: 112488378-112488378 on Build GRCh38
rs121918468 Chr.12: 112488444-112488444 on Build GRCh38
rs554148931 Chr.12: 112488361-112488361 on Build GRCh38
rs750640531 Chr.12: 112488439-112488439 on Build GRCh38
rs765455455 Chr.12: 112488438-112488438 on Build GRCh38
rs397507537 Chr.12: 112488465-112488465 on Build GRCh38
rs746719818 Chr.12: 112488488-112488488 on Build GRCh38

View Full Product Details