Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 72108642-72109130 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANAPC15
Entrez Gene ID
25906
Gene Name
anaphase promoting complex subunit 15
Gene Aliases
APC15, C11orf51, HSPC020
UniGene
Hs.38044
Gene Chromosome Location
Chr.11: 72106254-72112793 on Build GRCh38
Transcripts
XM_017017488.1 XM_017017489.1 XM_017017490.1 XM_017017491.1 XM_017017492.1 XM_017017493.1 XM_017017494.1 XM_017017495.1 NM_001145308.4 NM_001145309.3 NM_001145310.3 XM_017017356.1 XM_017017357.1 XM_017017358.1

Gene Symbol
LRTOMT
Entrez Gene ID
220074
Gene Name
leucine rich transmembrane and O-methyltransferase domain containing
Gene Aliases
CFAP111, DFNB63, LRRC51
UniGene
Hs.317243
Gene Chromosome Location
Chr.11: 72079982-72110782 on Build GRCh38
Transcripts
XM_017017488.1 XM_017017489.1 XM_017017490.1 XM_017017491.1 XM_017017492.1 XM_017017493.1 XM_017017494.1 XM_017017495.1 NM_001145308.4 NM_001145309.3 NM_001145310.3 XM_017017356.1 XM_017017357.1 XM_017017358.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3416210 c.755G>A p.R252H Chr.11: 72108804-72108804 on Build GRCh38
1585998 c.622C>T p.R208W Chr.11: 72108671-72108671 on Build GRCh38
4965260 c.641T>C p.L214P Chr.11: 72108690-72108690 on Build GRCh38
6348998 c.791G>T p.R264L Chr.11: 72108840-72108840 on Build GRCh38
6519333 c.650G>A p.R217Q Chr.11: 72108699-72108699 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs991850714 Chr.11: 72108694-72108694 on Build GRCh38
rs370362087 Chr.11: 72108689-72108689 on Build GRCh38
rs886044040 Chr.11: 72108852-72108852 on Build GRCh38
rs571179861 Chr.11: 72109081-72109081 on Build GRCh38
rs933277974 Chr.11: 72109090-72109090 on Build GRCh38
rs1003700981 Chr.11: 72108810-72108810 on Build GRCh38
rs372524977 Chr.11: 72108699-72108699 on Build GRCh38
rs948059983 Chr.11: 72109013-72109013 on Build GRCh38
rs878981648 Chr.11: 72108839-72108839 on Build GRCh38
rs376514789 Chr.11: 72109027-72109027 on Build GRCh38
rs892451020 Chr.11: 72108836-72108836 on Build GRCh38
rs1025935979 Chr.11: 72108829-72108829 on Build GRCh38
rs375590805 Chr.11: 72108787-72108787 on Build GRCh38
rs370168246 Chr.11: 72108803-72108803 on Build GRCh38
rs553247286 Chr.11: 72109071-72109071 on Build GRCh38
rs797044907 Chr.11: 72108662-72108663 on Build GRCh38
rs933205686 Chr.11: 72108805-72108805 on Build GRCh38
rs948147816 Chr.11: 72108773-72108773 on Build GRCh38
rs923848175 Chr.11: 72108704-72108704 on Build GRCh38
rs534644428 Chr.11: 72109048-72109048 on Build GRCh38
rs887354138 Chr.11: 72108833-72108833 on Build GRCh38
rs1016024999 Chr.11: 72109005-72109005 on Build GRCh38
rs773983684 Chr.11: 72108930-72108930 on Build GRCh38
rs928062227 Chr.11: 72109060-72109060 on Build GRCh38
rs540371915 Chr.11: 72108744-72108744 on Build GRCh38
rs397516627 Chr.11: 72108721-72108721 on Build GRCh38
rs935371141 Chr.11: 72108741-72108741 on Build GRCh38
rs72953778 Chr.11: 72108698-72108698 on Build GRCh38
rs943224181 Chr.11: 72108751-72108751 on Build GRCh38
rs770337351 Chr.11: 72108863-72108863 on Build GRCh38
rs371308729 Chr.11: 72108887-72108887 on Build GRCh38
rs968187301 Chr.11: 72108664-72108664 on Build GRCh38
rs987315987 Chr.11: 72108875-72108875 on Build GRCh38
rs974485290 Chr.11: 72108671-72108671 on Build GRCh38
rs727503153 Chr.11: 72108893-72108893 on Build GRCh38
rs762334109 Chr.11: 72108960-72108960 on Build GRCh38
rs950354642 Chr.11: 72108961-72108961 on Build GRCh38
rs766936141 Chr.11: 72108733-72108733 on Build GRCh38
rs876657863 Chr.11: 72108778-72108778 on Build GRCh38
rs984422120 Chr.11: 72108965-72108965 on Build GRCh38
rs61741195 Chr.11: 72108672-72108672 on Build GRCh38
rs779971909 Chr.11: 72108767-72108767 on Build GRCh38
rs1026023983 Chr.11: 72108933-72108933 on Build GRCh38
rs541690359 Chr.11: 72108737-72108737 on Build GRCh38
rs759862161 Chr.11: 72108954-72108959 on Build GRCh38
rs548947474 Chr.11: 72109010-72109010 on Build GRCh38
rs776760828 Chr.11: 72108846-72108846 on Build GRCh38
rs368590279 Chr.11: 72108845-72108845 on Build GRCh38
rs963049398 Chr.11: 72108840-72108840 on Build GRCh38
rs1005768038 Chr.11: 72108834-72108834 on Build GRCh38
rs775803008 Chr.11: 72108869-72108869 on Build GRCh38
rs886048633 Chr.11: 72108829-72108829 on Build GRCh38
rs567452783 Chr.11: 72109044-72109044 on Build GRCh38
rs984536181 Chr.11: 72108715-72108715 on Build GRCh38
rs886048635 Chr.11: 72108909-72108909 on Build GRCh38
rs754759504 Chr.11: 72108804-72108804 on Build GRCh38
rs886048634 Chr.11: 72108833-72108833 on Build GRCh38
rs747210285 Chr.11: 72108844-72108844 on Build GRCh38
rs752640844 Chr.11: 72108917-72108917 on Build GRCh38
rs530987072 Chr.11: 72108910-72108910 on Build GRCh38
rs781173860 Chr.11: 72108914-72108914 on Build GRCh38
rs956628252 Chr.11: 72108908-72108908 on Build GRCh38
rs886048636 Chr.11: 72108921-72108921 on Build GRCh38
rs551775464 Chr.11: 72108832-72108832 on Build GRCh38
rs915259470 Chr.11: 72108924-72108924 on Build GRCh38
rs71469421 Chr.11: 72108962-72108963 on Build GRCh38
rs570166217 Chr.11: 72108851-72108851 on Build GRCh38

View Full Product Details