Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 38206057-38206559 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DSCR10
Entrez Gene ID
259234
Gene Name
Down syndrome critical region 10 (non-protein coding)
Gene Aliases
-
UniGene
Hs.147695
Gene Chromosome Location
Chr.21: 38206156-38208644 on Build GRCh38
Transcripts
NR_027695.1

SNP Detail

SNP ID Chromosome Location
rs868761101 Chr.21: 38206109-38206109 on Build GRCh38
rs1041516879 Chr.21: 38206275-38206275 on Build GRCh38
rs1038502994 Chr.21: 38206391-38206391 on Build GRCh38
rs1045990253 Chr.21: 38206315-38206315 on Build GRCh38
rs896024087 Chr.21: 38206270-38206270 on Build GRCh38
rs1018714931 Chr.21: 38206435-38206435 on Build GRCh38
rs1033355403 Chr.21: 38206116-38206116 on Build GRCh38
rs965431388 Chr.21: 38206128-38206128 on Build GRCh38
rs950329607 Chr.21: 38206311-38206311 on Build GRCh38
rs902820108 Chr.21: 38206305-38206305 on Build GRCh38
rs141478447 Chr.21: 38206122-38206122 on Build GRCh38
rs936102243 Chr.21: 38206280-38206280 on Build GRCh38
rs767629973 Chr.21: 38206291-38206291 on Build GRCh38
rs1573271 Chr.21: 38206127-38206127 on Build GRCh38
rs34460077 Chr.21: 38206465-38206466 on Build GRCh38
rs965880055 Chr.21: 38206461-38206461 on Build GRCh38
rs189807996 Chr.21: 38206144-38206144 on Build GRCh38
rs75090952 Chr.21: 38206232-38206232 on Build GRCh38
rs1045871706 Chr.21: 38206082-38206082 on Build GRCh38
rs181414152 Chr.21: 38206239-38206239 on Build GRCh38
rs78557137 Chr.21: 38206123-38206123 on Build GRCh38
rs994628500 Chr.21: 38206466-38206466 on Build GRCh38
rs889024188 Chr.21: 38206393-38206393 on Build GRCh38
rs1055963297 Chr.21: 38206267-38206267 on Build GRCh38
rs987427296 Chr.21: 38206184-38206184 on Build GRCh38
rs145283188 Chr.21: 38206340-38206340 on Build GRCh38
rs77422882 Chr.21: 38206355-38206355 on Build GRCh38
rs1007471784 Chr.21: 38206397-38206397 on Build GRCh38
rs760640425 Chr.21: 38206428-38206428 on Build GRCh38
rs910130590 Chr.21: 38206189-38206189 on Build GRCh38
rs117522226 Chr.21: 38206112-38206112 on Build GRCh38
rs929982757 Chr.21: 38206185-38206185 on Build GRCh38
rs1010442391 Chr.21: 38206113-38206113 on Build GRCh38
rs540854357 Chr.21: 38206465-38206465 on Build GRCh38
rs570816567 Chr.21: 38206151-38206151 on Build GRCh38
rs898874280 Chr.21: 38206419-38206419 on Build GRCh38
rs753118391 Chr.21: 38206195-38206195 on Build GRCh38
rs984367720 Chr.21: 38206187-38206187 on Build GRCh38
rs530024395 Chr.21: 38206491-38206491 on Build GRCh38
rs886131585 Chr.21: 38206502-38206502 on Build GRCh38
rs949762908 Chr.21: 38206271-38206271 on Build GRCh38
rs796397140 Chr.21: 38206092-38206092 on Build GRCh38
rs926004488 Chr.21: 38206279-38206279 on Build GRCh38
rs368617974 Chr.21: 38206492-38206492 on Build GRCh38
rs766073314 Chr.21: 38206146-38206146 on Build GRCh38

View Full Product Details