Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 38285978-38286490 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EGFLAM
Entrez Gene ID
133584
Gene Name
EGF like, fibronectin type III and laminin G domains
Gene Aliases
AGRINL, AGRNL, PIKA
UniGene
Hs.20103
Gene Chromosome Location
Chr.5: 38258409-38465480 on Build GRCh38
Transcripts
NM_001205301.1 NM_152403.3 NR_046219.1

Gene Symbol
EGFLAM-AS4
Entrez Gene ID
100852408
Gene Name
EGFLAM antisense RNA 4
Gene Aliases
-
UniGene
Hs.570954
Gene Chromosome Location
Chr.5: 38282269-38290986 on Build GRCh38
Transcripts
NM_001205301.1 NM_152403.3 NR_046219.1

SNP Detail

SNP ID Chromosome Location
rs1039235763 Chr.5: 38286018-38286018 on Build GRCh38
rs928515391 Chr.5: 38286218-38286218 on Build GRCh38
rs950754738 Chr.5: 38286264-38286264 on Build GRCh38
rs796728732 Chr.5: 38286327-38286327 on Build GRCh38
rs1037842828 Chr.5: 38286298-38286298 on Build GRCh38
rs867790922 Chr.5: 38286181-38286181 on Build GRCh38
rs1012068859 Chr.5: 38286456-38286456 on Build GRCh38
rs973398400 Chr.5: 38286207-38286207 on Build GRCh38
rs1042049033 Chr.5: 38286398-38286398 on Build GRCh38
rs147136320 Chr.5: 38286455-38286455 on Build GRCh38
rs1030510853 Chr.5: 38286019-38286019 on Build GRCh38
rs758416742 Chr.5: 38286094-38286094 on Build GRCh38
rs1003739500 Chr.5: 38286079-38286079 on Build GRCh38
rs183413790 Chr.5: 38286061-38286061 on Build GRCh38
rs76173990 Chr.5: 38286440-38286440 on Build GRCh38
rs557320903 Chr.5: 38286433-38286433 on Build GRCh38
rs188811386 Chr.5: 38286326-38286326 on Build GRCh38
rs943514584 Chr.5: 38286293-38286293 on Build GRCh38
rs568317448 Chr.5: 38286296-38286296 on Build GRCh38
rs989969423 Chr.5: 38286149-38286149 on Build GRCh38
rs959950973 Chr.5: 38286220-38286220 on Build GRCh38
rs1012105961 Chr.5: 38286115-38286115 on Build GRCh38
rs114112547 Chr.5: 38286325-38286325 on Build GRCh38
rs796752630 Chr.5: 38286118-38286118 on Build GRCh38
rs958265521 Chr.5: 38286129-38286129 on Build GRCh38
rs890674382 Chr.5: 38286428-38286428 on Build GRCh38
rs531797960 Chr.5: 38286075-38286075 on Build GRCh38
rs550509706 Chr.5: 38286329-38286329 on Build GRCh38
rs57879643 Chr.5: 38286108-38286108 on Build GRCh38
rs76076333 Chr.5: 38286107-38286107 on Build GRCh38
rs1003213958 Chr.5: 38286421-38286421 on Build GRCh38
rs374978667 Chr.5: 38286201-38286201 on Build GRCh38
rs943540981 Chr.5: 38286003-38286003 on Build GRCh38
rs1042063139 Chr.5: 38286038-38286038 on Build GRCh38
rs919284351 Chr.5: 38286233-38286233 on Build GRCh38
rs143448662 Chr.5: 38286087-38286087 on Build GRCh38
rs76549731 Chr.5: 38286108-38286108 on Build GRCh38
rs1021063377 Chr.5: 38286187-38286189 on Build GRCh38
rs537357506 Chr.5: 38286031-38286031 on Build GRCh38
rs902143870 Chr.5: 38286411-38286411 on Build GRCh38
rs548166678 Chr.5: 38286202-38286202 on Build GRCh38
rs559111133 Chr.5: 38286037-38286037 on Build GRCh38
rs911996402 Chr.5: 38286274-38286274 on Build GRCh38
rs987918810 Chr.5: 38286273-38286273 on Build GRCh38

View Full Product Details