Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 154392243-154392789 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALNT10
Entrez Gene ID
55568
Gene Name
polypeptide N-acetylgalactosaminyltransferase 10
Gene Aliases
GALNACT10, PPGALNACT10, PPGANTASE10
UniGene
Hs.631797
Gene Chromosome Location
Chr.5: 154190735-154420984 on Build GRCh38
Transcripts
NM_198321.3 NR_037897.1

Gene Symbol
SAP30L-AS1
Entrez Gene ID
386627
Gene Name
SAP30L antisense RNA 1 (head to head)
Gene Aliases
GALNT10-AS1
UniGene
Hs.652021
Gene Chromosome Location
Chr.5: 154389769-154445822 on Build GRCh38
Transcripts
NM_198321.3 NR_037897.1

SNP Detail

SNP ID Chromosome Location
rs546543868 Chr.5: 154392483-154392483 on Build GRCh38
rs10079389 Chr.5: 154392708-154392708 on Build GRCh38
rs749705413 Chr.5: 154392467-154392467 on Build GRCh38
rs111738868 Chr.5: 154392550-154392550 on Build GRCh38
rs903597973 Chr.5: 154392394-154392394 on Build GRCh38
rs931153532 Chr.5: 154392506-154392506 on Build GRCh38
rs544240864 Chr.5: 154392595-154392595 on Build GRCh38
rs967496192 Chr.5: 154392459-154392459 on Build GRCh38
rs972549429 Chr.5: 154392503-154392503 on Build GRCh38
rs906562786 Chr.5: 154392367-154392367 on Build GRCh38
rs931415064 Chr.5: 154392280-154392280 on Build GRCh38
rs944016051 Chr.5: 154392525-154392525 on Build GRCh38
rs545705253 Chr.5: 154392397-154392397 on Build GRCh38
rs952492599 Chr.5: 154392661-154392661 on Build GRCh38
rs10064551 Chr.5: 154392647-154392647 on Build GRCh38
rs1049739106 Chr.5: 154392283-154392283 on Build GRCh38
rs182409648 Chr.5: 154392360-154392360 on Build GRCh38
rs919781113 Chr.5: 154392505-154392505 on Build GRCh38
rs117199475 Chr.5: 154392629-154392629 on Build GRCh38
rs965064868 Chr.5: 154392518-154392518 on Build GRCh38
rs10064410 Chr.5: 154392601-154392601 on Build GRCh38
rs562114732 Chr.5: 154392689-154392689 on Build GRCh38
rs1034260542 Chr.5: 154392378-154392378 on Build GRCh38
rs751051880 Chr.5: 154392642-154392642 on Build GRCh38
rs577569401 Chr.5: 154392377-154392377 on Build GRCh38
rs78463228 Chr.5: 154392728-154392728 on Build GRCh38
rs952995438 Chr.5: 154392531-154392531 on Build GRCh38
rs11746823 Chr.5: 154392265-154392265 on Build GRCh38
rs142220285 Chr.5: 154392645-154392645 on Build GRCh38
rs189433165 Chr.5: 154392644-154392644 on Build GRCh38
rs1046490891 Chr.5: 154392598-154392598 on Build GRCh38
rs1011498789 Chr.5: 154392610-154392610 on Build GRCh38
rs1033407934 Chr.5: 154392413-154392413 on Build GRCh38
rs10064618 Chr.5: 154392729-154392729 on Build GRCh38
rs11750554 Chr.5: 154392615-154392615 on Build GRCh38
rs760824238 Chr.5: 154392639-154392640 on Build GRCh38
rs546431941 Chr.5: 154392604-154392604 on Build GRCh38
rs992241670 Chr.5: 154392482-154392482 on Build GRCh38
rs757087525 Chr.5: 154392628-154392628 on Build GRCh38
rs913244422 Chr.5: 154392715-154392715 on Build GRCh38
rs985296830 Chr.5: 154392688-154392688 on Build GRCh38
rs115179451 Chr.5: 154392605-154392605 on Build GRCh38
rs894302666 Chr.5: 154392386-154392386 on Build GRCh38
rs946319202 Chr.5: 154392576-154392576 on Build GRCh38
rs944152328 Chr.5: 154392690-154392690 on Build GRCh38
rs369869893 Chr.5: 154392753-154392753 on Build GRCh38
rs940128028 Chr.5: 154392365-154392365 on Build GRCh38
rs200757775 Chr.5: 154392679-154392680 on Build GRCh38
rs1002728966 Chr.5: 154392370-154392370 on Build GRCh38
rs959041599 Chr.5: 154392441-154392441 on Build GRCh38
rs575288002 Chr.5: 154392551-154392551 on Build GRCh38
rs1041267589 Chr.5: 154392321-154392321 on Build GRCh38
rs908686293 Chr.5: 154392720-154392720 on Build GRCh38
rs1011865584 Chr.5: 154392395-154392395 on Build GRCh38
rs780521685 Chr.5: 154392369-154392369 on Build GRCh38
rs889768157 Chr.5: 154392286-154392286 on Build GRCh38
rs552039233 Chr.5: 154392626-154392626 on Build GRCh38
rs1021459193 Chr.5: 154392434-154392434 on Build GRCh38
rs764722770 Chr.5: 154392366-154392366 on Build GRCh38
rs186589192 Chr.5: 154392596-154392596 on Build GRCh38
rs961626154 Chr.5: 154392501-154392501 on Build GRCh38
rs944051526 Chr.5: 154392315-154392315 on Build GRCh38
rs10072289 Chr.5: 154392521-154392521 on Build GRCh38
rs189838980 Chr.5: 154392324-154392324 on Build GRCh38
rs531235790 Chr.5: 154392716-154392716 on Build GRCh38
rs911306167 Chr.5: 154392523-154392523 on Build GRCh38

View Full Product Details