Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 112237148-112237280 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSMD3
Entrez Gene ID
114788
Gene Name
CUB and Sushi multiple domains 3
Gene Aliases
-
UniGene
Hs.91381
Gene Chromosome Location
Chr.8: 112222928-113437499 on Build GRCh38
Transcripts
NM_052900.2 NM_198123.1 NM_198124.1 XM_011516815.2 XM_011516816.2 XM_017013008.1 XM_017013009.1 XM_017013010.1 XM_017013011.1 XM_017013012.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1173367 c.10463T>G p.V3488G Chr.8: 112237234-112237234 on Build GRCh38
25453 c.10537C>T p.P3513S Chr.8: 112237280-112237280 on Build GRCh38
5652490 c.10576G>T p.G3526W Chr.8: 112237241-112237241 on Build GRCh38
1330371 c.10538C>A p.P3513H Chr.8: 112237279-112237279 on Build GRCh38
6179423 c.10589C>A p.A3530E Chr.8: 112237228-112237228 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139899414 Chr.8: 112237199-112237199 on Build GRCh38
rs753532523 Chr.8: 112237230-112237230 on Build GRCh38
rs148694393 Chr.8: 112237246-112237246 on Build GRCh38
rs751594020 Chr.8: 112237209-112237209 on Build GRCh38
rs779362635 Chr.8: 112237239-112237239 on Build GRCh38
rs780355829 Chr.8: 112237232-112237232 on Build GRCh38
rs775077628 Chr.8: 112237185-112237185 on Build GRCh38
rs759168754 Chr.8: 112237182-112237182 on Build GRCh38
rs533896354 Chr.8: 112237184-112237184 on Build GRCh38
rs145347948 Chr.8: 112237202-112237202 on Build GRCh38
rs757412233 Chr.8: 112237215-112237215 on Build GRCh38
rs749606037 Chr.8: 112237237-112237237 on Build GRCh38
rs147861313 Chr.8: 112237238-112237238 on Build GRCh38
rs750533886 Chr.8: 112237229-112237229 on Build GRCh38
rs748720200 Chr.8: 112237249-112237249 on Build GRCh38
rs770460606 Chr.8: 112237252-112237252 on Build GRCh38
rs767553384 Chr.8: 112237226-112237226 on Build GRCh38

View Full Product Details