Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 49033393-49033624 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TFE3
Entrez Gene ID
7030
Gene Name
transcription factor binding to IGHM enhancer 3
Gene Aliases
RCCP2, RCCX1, TFEA, bHLHe33
UniGene
Hs.730740
Gene Chromosome Location
Chr.X: 49028726-49043517 on Build GRCh38
Transcripts
NM_001282142.1 NM_006521.5

SNP Detail

SNP ID Chromosome Location
rs782784061 Chr.X: 49033484-49033484 on Build GRCh38
rs782342097 Chr.X: 49033600-49033600 on Build GRCh38
rs367560398 Chr.X: 49033435-49033435 on Build GRCh38
rs201808352 Chr.X: 49033512-49033512 on Build GRCh38
rs369698146 Chr.X: 49033588-49033588 on Build GRCh38
rs377187631 Chr.X: 49033583-49033583 on Build GRCh38
rs782535970 Chr.X: 49033591-49033591 on Build GRCh38
rs782445226 Chr.X: 49033418-49033418 on Build GRCh38
rs368263592 Chr.X: 49033447-49033447 on Build GRCh38
rs41305381 Chr.X: 49033443-49033443 on Build GRCh38
rs782697975 Chr.X: 49033558-49033558 on Build GRCh38
rs782106588 Chr.X: 49033546-49033546 on Build GRCh38
rs199560694 Chr.X: 49033419-49033419 on Build GRCh38

View Full Product Details